Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 87413748-87414283 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CFAP206
Entrez Gene ID
154313
Gene Name
cilia and flagella associated protein 206
Gene Aliases
C6orf165, dJ382I10.1
UniGene
Hs.82921
Gene Chromosome Location
Chr.6: 87407972-87464473 on Build GRCh38
Transcripts
NM_001031743.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1198931 c.201G>C p.M67I Chr.6: 87413818-87413818 on Build GRCh38
4938735 c.206G>T p.R69L Chr.6: 87413823-87413823 on Build GRCh38
1198930 c.201G>C p.M67I Chr.6: 87413818-87413818 on Build GRCh38
3177738 c.276G>A p.T92T Chr.6: 87413893-87413893 on Build GRCh38
3177732 c.227C>T p.P76L Chr.6: 87413844-87413844 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs549525189 Chr.6: 87413883-87413883 on Build GRCh38
rs554234477 Chr.6: 87414175-87414175 on Build GRCh38
rs112448795 Chr.6: 87414080-87414080 on Build GRCh38
rs576109113 Chr.6: 87414245-87414245 on Build GRCh38
rs965359824 Chr.6: 87414098-87414098 on Build GRCh38
rs753028170 Chr.6: 87413836-87413836 on Build GRCh38
rs947158483 Chr.6: 87413774-87413774 on Build GRCh38
rs979585448 Chr.6: 87413786-87413786 on Build GRCh38
rs958288127 Chr.6: 87414176-87414176 on Build GRCh38
rs975348739 Chr.6: 87414151-87414151 on Build GRCh38
rs750829085 Chr.6: 87414194-87414194 on Build GRCh38
rs1021712068 Chr.6: 87414101-87414101 on Build GRCh38
rs138677801 Chr.6: 87413892-87413892 on Build GRCh38
rs67411803 Chr.6: 87414044-87414045 on Build GRCh38
rs377042908 Chr.6: 87413879-87413879 on Build GRCh38
rs774364253 Chr.6: 87413791-87413791 on Build GRCh38
rs767989524 Chr.6: 87413908-87413908 on Build GRCh38
rs958640391 Chr.6: 87414027-87414027 on Build GRCh38
rs777957063 Chr.6: 87413856-87413856 on Build GRCh38
rs756054604 Chr.6: 87414151-87414153 on Build GRCh38
rs753261037 Chr.6: 87413910-87413910 on Build GRCh38
rs780757723 Chr.6: 87414248-87414249 on Build GRCh38
rs1026134589 Chr.6: 87414087-87414087 on Build GRCh38
rs368228971 Chr.6: 87413933-87413933 on Build GRCh38
rs769544970 Chr.6: 87413807-87413807 on Build GRCh38
rs965639371 Chr.6: 87413823-87413823 on Build GRCh38
rs763730094 Chr.6: 87414134-87414134 on Build GRCh38
rs983218483 Chr.6: 87413810-87413810 on Build GRCh38
rs35438647 Chr.6: 87413831-87413831 on Build GRCh38
rs76111328 Chr.6: 87414043-87414043 on Build GRCh38
rs138459969 Chr.6: 87413891-87413891 on Build GRCh38
rs1019608010 Chr.6: 87414094-87414094 on Build GRCh38
rs761521740 Chr.6: 87413801-87413801 on Build GRCh38
rs201184978 Chr.6: 87414038-87414039 on Build GRCh38
rs143377893 Chr.6: 87414041-87414042 on Build GRCh38
rs762359523 Chr.6: 87413816-87413816 on Build GRCh38
rs761021569 Chr.6: 87413918-87413918 on Build GRCh38
rs370690253 Chr.6: 87413897-87413897 on Build GRCh38
rs566911911 Chr.6: 87413865-87413865 on Build GRCh38
rs751951635 Chr.6: 87413832-87413832 on Build GRCh38
rs977588624 Chr.6: 87414224-87414224 on Build GRCh38
rs142290940 Chr.6: 87413814-87413814 on Build GRCh38
rs768745897 Chr.6: 87413882-87413882 on Build GRCh38
rs188721494 Chr.6: 87413893-87413893 on Build GRCh38
rs532439821 Chr.6: 87413837-87413837 on Build GRCh38
rs760106614 Chr.6: 87413833-87413833 on Build GRCh38
rs192846814 Chr.6: 87414009-87414009 on Build GRCh38
rs968463991 Chr.6: 87414127-87414127 on Build GRCh38
rs542201942 Chr.6: 87414173-87414173 on Build GRCh38
rs16667 Chr.6: 87414043-87414044 on Build GRCh38
rs899057027 Chr.6: 87413901-87413901 on Build GRCh38
rs370028866 Chr.6: 87413889-87413889 on Build GRCh38
rs571737593 Chr.6: 87414168-87414168 on Build GRCh38
rs538989180 Chr.6: 87414090-87414090 on Build GRCh38
rs554042927 Chr.6: 87414091-87414091 on Build GRCh38
rs140621996 Chr.6: 87413863-87413863 on Build GRCh38
rs773938782 Chr.6: 87413898-87413898 on Build GRCh38
rs758332898 Chr.6: 87413928-87413929 on Build GRCh38
rs769771629 Chr.6: 87413888-87413888 on Build GRCh38
rs1002767244 Chr.6: 87413939-87413939 on Build GRCh38
rs748077718 Chr.6: 87413836-87413837 on Build GRCh38
rs117489265 Chr.6: 87413878-87413878 on Build GRCh38
rs767825733 Chr.6: 87413835-87413835 on Build GRCh38
rs776590872 Chr.6: 87413792-87413792 on Build GRCh38
rs1027502058 Chr.6: 87413781-87413781 on Build GRCh38

View Full Product Details