Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 87428331-87428840 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CFAP206
Entrez Gene ID
154313
Gene Name
cilia and flagella associated protein 206
Gene Aliases
C6orf165, dJ382I10.1
UniGene
Hs.82921
Gene Chromosome Location
Chr.6: 87407972-87464473 on Build GRCh38
Transcripts
NM_001031743.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4586963 c.1005C>T p.D335D Chr.6: 87428670-87428670 on Build GRCh38
1131784 c.992C>G p.T331S Chr.6: 87428657-87428657 on Build GRCh38
3703080 c.1049T>G p.I350S Chr.6: 87428714-87428714 on Build GRCh38
5527791 c.1105C>T p.H369Y Chr.6: 87428770-87428770 on Build GRCh38
3949520 c.1062G>C p.L354F Chr.6: 87428727-87428727 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs767554033 Chr.6: 87428726-87428726 on Build GRCh38
rs1038191152 Chr.6: 87428590-87428590 on Build GRCh38
rs936783060 Chr.6: 87428437-87428437 on Build GRCh38
rs776856828 Chr.6: 87428700-87428700 on Build GRCh38
rs915980269 Chr.6: 87428392-87428392 on Build GRCh38
rs756870628 Chr.6: 87428770-87428770 on Build GRCh38
rs191910901 Chr.6: 87428564-87428564 on Build GRCh38
rs966745567 Chr.6: 87428555-87428555 on Build GRCh38
rs761097527 Chr.6: 87428684-87428684 on Build GRCh38
rs112263505 Chr.6: 87428423-87428423 on Build GRCh38
rs752577387 Chr.6: 87428733-87428733 on Build GRCh38
rs745368981 Chr.6: 87428783-87428783 on Build GRCh38
rs943418794 Chr.6: 87428561-87428561 on Build GRCh38
rs565388337 Chr.6: 87428575-87428575 on Build GRCh38
rs762038475 Chr.6: 87428749-87428749 on Build GRCh38
rs563231792 Chr.6: 87428701-87428701 on Build GRCh38
rs769018531 Chr.6: 87428802-87428802 on Build GRCh38
rs776034016 Chr.6: 87428683-87428683 on Build GRCh38
rs750503458 Chr.6: 87428718-87428718 on Build GRCh38
rs755925712 Chr.6: 87428631-87428631 on Build GRCh38
rs368000058 Chr.6: 87428481-87428481 on Build GRCh38
rs765370275 Chr.6: 87428713-87428713 on Build GRCh38
rs553208689 Chr.6: 87428408-87428408 on Build GRCh38
rs373776261 Chr.6: 87428641-87428641 on Build GRCh38
rs753487433 Chr.6: 87428754-87428754 on Build GRCh38
rs901354710 Chr.6: 87428465-87428465 on Build GRCh38
rs765422738 Chr.6: 87428594-87428594 on Build GRCh38
rs889104082 Chr.6: 87428543-87428543 on Build GRCh38
rs959172942 Chr.6: 87428368-87428369 on Build GRCh38
rs760817966 Chr.6: 87428590-87428591 on Build GRCh38
rs1052612547 Chr.6: 87428466-87428466 on Build GRCh38
rs745420473 Chr.6: 87428671-87428671 on Build GRCh38
rs781321836 Chr.6: 87428626-87428626 on Build GRCh38
rs922830524 Chr.6: 87428753-87428753 on Build GRCh38
rs780543947 Chr.6: 87428788-87428788 on Build GRCh38
rs915542030 Chr.6: 87428400-87428400 on Build GRCh38
rs1041284448 Chr.6: 87428446-87428446 on Build GRCh38
rs1011029912 Chr.6: 87428540-87428540 on Build GRCh38
rs145724678 Chr.6: 87428748-87428748 on Build GRCh38
rs768907065 Chr.6: 87428687-87428687 on Build GRCh38
rs151002888 Chr.6: 87428670-87428670 on Build GRCh38
rs747486023 Chr.6: 87428790-87428790 on Build GRCh38
rs891769274 Chr.6: 87428415-87428429 on Build GRCh38
rs754999878 Chr.6: 87428604-87428604 on Build GRCh38
rs552297180 Chr.6: 87428743-87428743 on Build GRCh38
rs374985737 Chr.6: 87428720-87428720 on Build GRCh38
rs74507171 Chr.6: 87428500-87428500 on Build GRCh38
rs770376098 Chr.6: 87428650-87428650 on Build GRCh38
rs183010552 Chr.6: 87428784-87428784 on Build GRCh38
rs541397229 Chr.6: 87428589-87428589 on Build GRCh38
rs1033139947 Chr.6: 87428740-87428740 on Build GRCh38
rs576137367 Chr.6: 87428523-87428523 on Build GRCh38
rs76730959 Chr.6: 87428384-87428384 on Build GRCh38
rs778585023 Chr.6: 87428771-87428771 on Build GRCh38
rs988284828 Chr.6: 87428766-87428766 on Build GRCh38
rs370949345 Chr.6: 87428638-87428638 on Build GRCh38
rs893549660 Chr.6: 87428527-87428527 on Build GRCh38
rs978453075 Chr.6: 87428739-87428739 on Build GRCh38
rs1003431054 Chr.6: 87428571-87428571 on Build GRCh38
rs772545515 Chr.6: 87428673-87428673 on Build GRCh38
rs914128360 Chr.6: 87428486-87428486 on Build GRCh38
rs751495559 Chr.6: 87428602-87428602 on Build GRCh38
rs34294674 Chr.6: 87428409-87428409 on Build GRCh38
rs932840036 Chr.6: 87428764-87428764 on Build GRCh38
rs79821877 Chr.6: 87428615-87428615 on Build GRCh38
rs776892905 Chr.6: 87428806-87428806 on Build GRCh38
rs866475168 Chr.6: 87428536-87428536 on Build GRCh38
rs577348374 Chr.6: 87428576-87428576 on Build GRCh38
rs34588434 Chr.6: 87428736-87428736 on Build GRCh38
rs1020621475 Chr.6: 87428545-87428545 on Build GRCh38
rs777448285 Chr.6: 87428737-87428737 on Build GRCh38
rs376498041 Chr.6: 87428629-87428629 on Build GRCh38
rs952822289 Chr.6: 87428729-87428729 on Build GRCh38
rs1052048087 Chr.6: 87428541-87428541 on Build GRCh38
rs914054858 Chr.6: 87428767-87428767 on Build GRCh38

View Full Product Details