Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 116253713-116254209 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DSE
Entrez Gene ID
29940
Gene Name
dermatan sulfate epimerase
Gene Aliases
DS-epi1, DSEP, DSEPI, EDSMC2, SART-2, SART2
UniGene
Hs.458358
Gene Chromosome Location
Chr.6: 116254152-116441261 on Build GRCh38
Transcripts
NM_001322937.1 NM_001322938.1 NM_001322940.1 NR_136520.1 NM_021648.4

Gene Symbol
TSPYL4
Entrez Gene ID
23270
Gene Name
TSPY like 4
Gene Aliases
dJ486I3.2
UniGene
Hs.284141
Gene Chromosome Location
Chr.6: 116249964-116254098 on Build GRCh38
Transcripts
NM_001322937.1 NM_001322938.1 NM_001322940.1 NR_136520.1 NM_021648.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6810705 c.182G>A p.G61D Chr.6: 116253827-116253827 on Build GRCh38
5737735 c.160C>A p.L54M Chr.6: 116253849-116253849 on Build GRCh38
6566189 c.24C>T p.N8N Chr.6: 116253985-116253985 on Build GRCh38
6514305 c.27G>T p.K9N Chr.6: 116253982-116253982 on Build GRCh38
6514304 c.109C>T p.R37C Chr.6: 116253900-116253900 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs756388718 Chr.6: 116253955-116253955 on Build GRCh38
rs17524614 Chr.6: 116253953-116253953 on Build GRCh38
rs756121275 Chr.6: 116253905-116253905 on Build GRCh38
rs1029564649 Chr.6: 116254071-116254071 on Build GRCh38
rs17553296 Chr.6: 116253992-116253992 on Build GRCh38
rs766150650 Chr.6: 116254015-116254015 on Build GRCh38
rs910391 Chr.6: 116254181-116254181 on Build GRCh38
rs1010042375 Chr.6: 116254183-116254183 on Build GRCh38
rs760585826 Chr.6: 116254017-116254018 on Build GRCh38
rs972407276 Chr.6: 116254065-116254065 on Build GRCh38
rs753768829 Chr.6: 116253868-116253868 on Build GRCh38
rs757305705 Chr.6: 116253959-116253959 on Build GRCh38
rs200293371 Chr.6: 116253832-116253832 on Build GRCh38
rs773546657 Chr.6: 116253767-116253767 on Build GRCh38
rs983913017 Chr.6: 116253864-116253864 on Build GRCh38
rs192760666 Chr.6: 116253925-116253925 on Build GRCh38
rs187133419 Chr.6: 116253814-116253814 on Build GRCh38
rs760221449 Chr.6: 116253858-116253858 on Build GRCh38
rs775215098 Chr.6: 116253924-116253924 on Build GRCh38
rs761864202 Chr.6: 116253878-116253878 on Build GRCh38
rs201531864 Chr.6: 116253831-116253831 on Build GRCh38
rs1049908114 Chr.6: 116253893-116253893 on Build GRCh38
rs75652959 Chr.6: 116253921-116253921 on Build GRCh38
rs149060725 Chr.6: 116253777-116253777 on Build GRCh38
rs770330799 Chr.6: 116253762-116253762 on Build GRCh38
rs748653080 Chr.6: 116253761-116253761 on Build GRCh38
rs368304091 Chr.6: 116253820-116253820 on Build GRCh38
rs995297376 Chr.6: 116254106-116254106 on Build GRCh38
rs771536035 Chr.6: 116253915-116253915 on Build GRCh38
rs769221538 Chr.6: 116253996-116253996 on Build GRCh38
rs757292974 Chr.6: 116254036-116254036 on Build GRCh38
rs761621515 Chr.6: 116253938-116253938 on Build GRCh38
rs1053696232 Chr.6: 116253763-116253763 on Build GRCh38
rs777410337 Chr.6: 116254050-116254052 on Build GRCh38
rs905567471 Chr.6: 116254114-116254114 on Build GRCh38
rs767139495 Chr.6: 116253826-116253838 on Build GRCh38
rs375496363 Chr.6: 116253888-116253888 on Build GRCh38
rs376996291 Chr.6: 116254048-116254048 on Build GRCh38
rs774837804 Chr.6: 116253845-116253845 on Build GRCh38
rs370141135 Chr.6: 116254002-116254002 on Build GRCh38
rs531814242 Chr.6: 116254081-116254081 on Build GRCh38
rs755420805 Chr.6: 116254050-116254050 on Build GRCh38
rs2232470 Chr.6: 116253920-116253920 on Build GRCh38
rs140380512 Chr.6: 116254152-116254152 on Build GRCh38
rs979822761 Chr.6: 116254003-116254003 on Build GRCh38
rs762915452 Chr.6: 116253912-116253912 on Build GRCh38
rs764251448 Chr.6: 116254031-116254031 on Build GRCh38
rs544194039 Chr.6: 116254163-116254163 on Build GRCh38
rs766923864 Chr.6: 116253782-116253782 on Build GRCh38
rs41289922 Chr.6: 116253885-116253885 on Build GRCh38
rs527681800 Chr.6: 116254004-116254004 on Build GRCh38
rs763699582 Chr.6: 116253862-116253862 on Build GRCh38
rs959365088 Chr.6: 116253850-116253850 on Build GRCh38
rs774198411 Chr.6: 116254020-116254020 on Build GRCh38
rs552398310 Chr.6: 116253901-116253901 on Build GRCh38
rs773050437 Chr.6: 116253948-116253948 on Build GRCh38
rs759107370 Chr.6: 116253849-116253849 on Build GRCh38
rs765271762 Chr.6: 116253946-116253946 on Build GRCh38
rs960999930 Chr.6: 116254052-116254052 on Build GRCh38
rs752294732 Chr.6: 116253786-116253786 on Build GRCh38
rs760667007 Chr.6: 116253795-116253795 on Build GRCh38
rs368916895 Chr.6: 116253892-116253892 on Build GRCh38
rs750925816 Chr.6: 116254039-116254039 on Build GRCh38
rs138535555 Chr.6: 116254075-116254075 on Build GRCh38
rs749745938 Chr.6: 116253910-116253910 on Build GRCh38
rs756931755 Chr.6: 116253805-116253805 on Build GRCh38
rs571471665 Chr.6: 116254179-116254179 on Build GRCh38
rs912693090 Chr.6: 116254042-116254042 on Build GRCh38
rs773811221 Chr.6: 116253778-116253778 on Build GRCh38
rs1042588624 Chr.6: 116254029-116254029 on Build GRCh38
rs760856147 Chr.6: 116254023-116254023 on Build GRCh38
rs775985772 Chr.6: 116253990-116253990 on Build GRCh38
rs775303353 Chr.6: 116253857-116253857 on Build GRCh38
rs771805876 Chr.6: 116253851-116253851 on Build GRCh38
rs375497924 Chr.6: 116253989-116253989 on Build GRCh38
rs776826016 Chr.6: 116253743-116253743 on Build GRCh38
rs375653462 Chr.6: 116253834-116253834 on Build GRCh38
rs891332085 Chr.6: 116253899-116253899 on Build GRCh38
rs911037818 Chr.6: 116254091-116254092 on Build GRCh38
rs776343865 Chr.6: 116253930-116253930 on Build GRCh38
rs765244525 Chr.6: 116254038-116254038 on Build GRCh38
rs779434331 Chr.6: 116253974-116253974 on Build GRCh38
rs773138804 Chr.6: 116254000-116254000 on Build GRCh38
rs2232468 Chr.6: 116253988-116253988 on Build GRCh38
rs2232466 Chr.6: 116254113-116254113 on Build GRCh38
rs200711366 Chr.6: 116253922-116253922 on Build GRCh38
rs199748766 Chr.6: 116253926-116253926 on Build GRCh38
rs199711865 Chr.6: 116253952-116253952 on Build GRCh38
rs750275527 Chr.6: 116253889-116253889 on Build GRCh38
rs572966412 Chr.6: 116254049-116254049 on Build GRCh38
rs759240410 Chr.6: 116253781-116253781 on Build GRCh38
rs369213899 Chr.6: 116253981-116253981 on Build GRCh38
rs768557241 Chr.6: 116253741-116253741 on Build GRCh38
rs542187191 Chr.6: 116254041-116254041 on Build GRCh38
rs749890763 Chr.6: 116253906-116253907 on Build GRCh38
rs896783100 Chr.6: 116253932-116253932 on Build GRCh38
rs753855600 Chr.6: 116253796-116253796 on Build GRCh38
rs985423502 Chr.6: 116254090-116254090 on Build GRCh38
rs751714101 Chr.6: 116253815-116253815 on Build GRCh38
rs763257455 Chr.6: 116253775-116253775 on Build GRCh38
rs771144968 Chr.6: 116253840-116253840 on Build GRCh38
rs2232469 Chr.6: 116253958-116253958 on Build GRCh38
rs959868856 Chr.6: 116253977-116253977 on Build GRCh38
rs2232467 Chr.6: 116254047-116254047 on Build GRCh38
rs754037847 Chr.6: 116254034-116254034 on Build GRCh38

View Full Product Details