Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 116496410-116496926 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TRAPPC3L
Entrez Gene ID
100128327
Gene Name
trafficking protein particle complex 3 like
Gene Aliases
BET3L, bA259P20.2
UniGene
Hs.134795
Gene Chromosome Location
Chr.6: 116496483-116545610 on Build GRCh38
Transcripts
NM_001139444.2

SNP Detail

SNP ID Chromosome Location
rs752415039 Chr.6: 116496719-116496719 on Build GRCh38
rs1028233843 Chr.6: 116496566-116496566 on Build GRCh38
rs949908277 Chr.6: 116496718-116496718 on Build GRCh38
rs905675664 Chr.6: 116496750-116496750 on Build GRCh38
rs188929422 Chr.6: 116496656-116496656 on Build GRCh38
rs955424665 Chr.6: 116496567-116496567 on Build GRCh38
rs751266363 Chr.6: 116496862-116496862 on Build GRCh38
rs536796525 Chr.6: 116496456-116496456 on Build GRCh38
rs140201245 Chr.6: 116496528-116496528 on Build GRCh38
rs181681581 Chr.6: 116496772-116496772 on Build GRCh38
rs370448856 Chr.6: 116496481-116496481 on Build GRCh38
rs1027413693 Chr.6: 116496529-116496529 on Build GRCh38
rs568119568 Chr.6: 116496502-116496502 on Build GRCh38
rs921039655 Chr.6: 116496596-116496596 on Build GRCh38
rs986613819 Chr.6: 116496578-116496578 on Build GRCh38
rs1028661368 Chr.6: 116496888-116496888 on Build GRCh38
rs140962692 Chr.6: 116496790-116496791 on Build GRCh38
rs1002251450 Chr.6: 116496771-116496771 on Build GRCh38
rs555195730 Chr.6: 116496486-116496486 on Build GRCh38
rs892184208 Chr.6: 116496497-116496497 on Build GRCh38
rs560120430 Chr.6: 116496583-116496584 on Build GRCh38
rs574588986 Chr.6: 116496854-116496854 on Build GRCh38
rs762406312 Chr.6: 116496775-116496775 on Build GRCh38
rs371022711 Chr.6: 116496715-116496715 on Build GRCh38
rs150335649 Chr.6: 116496717-116496717 on Build GRCh38
rs1056605697 Chr.6: 116496732-116496732 on Build GRCh38
rs976905947 Chr.6: 116496584-116496584 on Build GRCh38
rs911985766 Chr.6: 116496629-116496629 on Build GRCh38
rs1025041488 Chr.6: 116496494-116496494 on Build GRCh38
rs151079190 Chr.6: 116496436-116496436 on Build GRCh38
rs920621535 Chr.6: 116496660-116496660 on Build GRCh38
rs1045764707 Chr.6: 116496691-116496691 on Build GRCh38
rs945131013 Chr.6: 116496796-116496796 on Build GRCh38
rs528932338 Chr.6: 116496646-116496646 on Build GRCh38
rs2637667 Chr.6: 116496450-116496450 on Build GRCh38
rs1044225939 Chr.6: 116496730-116496730 on Build GRCh38
rs952708633 Chr.6: 116496543-116496543 on Build GRCh38
rs200993473 Chr.6: 116496791-116496791 on Build GRCh38
rs903555740 Chr.6: 116496812-116496812 on Build GRCh38
rs965964468 Chr.6: 116496571-116496571 on Build GRCh38
rs983440587 Chr.6: 116496549-116496549 on Build GRCh38
rs1043481901 Chr.6: 116496800-116496800 on Build GRCh38
rs967687861 Chr.6: 116496525-116496525 on Build GRCh38
rs777640751 Chr.6: 116496859-116496859 on Build GRCh38
rs1040241633 Chr.6: 116496740-116496740 on Build GRCh38
rs895718637 Chr.6: 116496713-116496713 on Build GRCh38
rs537327187 Chr.6: 116496607-116496607 on Build GRCh38
rs1009249085 Chr.6: 116496499-116496499 on Build GRCh38
rs3832400 Chr.6: 116496887-116496888 on Build GRCh38
rs577131240 Chr.6: 116496662-116496662 on Build GRCh38
rs962075835 Chr.6: 116496654-116496654 on Build GRCh38
rs932411091 Chr.6: 116496642-116496642 on Build GRCh38
rs1015845529 Chr.6: 116496562-116496562 on Build GRCh38
rs34071551 Chr.6: 116496641-116496641 on Build GRCh38
rs1016289916 Chr.6: 116496808-116496808 on Build GRCh38
rs965601542 Chr.6: 116496876-116496876 on Build GRCh38

View Full Product Details