Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 121448006-121448516 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GJA1
Entrez Gene ID
2697
Gene Name
gap junction protein alpha 1
Gene Aliases
AVSD3, CMDR, CX43, EKVP, GJAL, HLHS1, HSS, ODDD, PPKCA
UniGene
Hs.700699
Gene Chromosome Location
Chr.6: 121435577-121449744 on Build GRCh38
Transcripts
NM_000165.4

SNP Detail

SNP ID Chromosome Location
rs976584595 Chr.6: 121448054-121448054 on Build GRCh38
rs72548749 Chr.6: 121448297-121448297 on Build GRCh38
rs997989542 Chr.6: 121448301-121448301 on Build GRCh38
rs971813926 Chr.6: 121448435-121448435 on Build GRCh38
rs886061010 Chr.6: 121448043-121448043 on Build GRCh38
rs910808442 Chr.6: 121448465-121448465 on Build GRCh38
rs67721882 Chr.6: 121448085-121448085 on Build GRCh38
rs1007597789 Chr.6: 121448261-121448261 on Build GRCh38
rs1036503910 Chr.6: 121448187-121448187 on Build GRCh38
rs36054859 Chr.6: 121448163-121448164 on Build GRCh38
rs967301127 Chr.6: 121448100-121448100 on Build GRCh38
rs571457341 Chr.6: 121448327-121448327 on Build GRCh38
rs72548747 Chr.6: 121448268-121448268 on Build GRCh38
rs943332195 Chr.6: 121448468-121448468 on Build GRCh38
rs72548748 Chr.6: 121448281-121448281 on Build GRCh38
rs772093340 Chr.6: 121448481-121448481 on Build GRCh38
rs919415571 Chr.6: 121448440-121448440 on Build GRCh38
rs554145953 Chr.6: 121448361-121448361 on Build GRCh38
rs549064986 Chr.6: 121448210-121448210 on Build GRCh38
rs778313354 Chr.6: 121448195-121448195 on Build GRCh38
rs533993385 Chr.6: 121448352-121448352 on Build GRCh38
rs45516992 Chr.6: 121448349-121448349 on Build GRCh38
rs949823229 Chr.6: 121448412-121448412 on Build GRCh38
rs772242482 Chr.6: 121448198-121448199 on Build GRCh38
rs72548746 Chr.6: 121448267-121448267 on Build GRCh38
rs886061011 Chr.6: 121448159-121448159 on Build GRCh38
rs916959201 Chr.6: 121448395-121448395 on Build GRCh38
rs777396732 Chr.6: 121448040-121448040 on Build GRCh38
rs113758463 Chr.6: 121448368-121448368 on Build GRCh38
rs775142598 Chr.6: 121448040-121448041 on Build GRCh38
rs561937205 Chr.6: 121448098-121448098 on Build GRCh38
rs139128953 Chr.6: 121448239-121448239 on Build GRCh38
rs889107668 Chr.6: 121448241-121448241 on Build GRCh38
rs930880001 Chr.6: 121448222-121448222 on Build GRCh38
rs748395995 Chr.6: 121448160-121448160 on Build GRCh38
rs746134652 Chr.6: 121448042-121448042 on Build GRCh38
rs1049776092 Chr.6: 121448449-121448449 on Build GRCh38
rs758644469 Chr.6: 121448048-121448048 on Build GRCh38
rs538969867 Chr.6: 121448446-121448447 on Build GRCh38
rs72548745 Chr.6: 121448237-121448237 on Build GRCh38
rs868021594 Chr.6: 121448084-121448084 on Build GRCh38
rs967227385 Chr.6: 121448358-121448358 on Build GRCh38
rs544396539 Chr.6: 121448119-121448119 on Build GRCh38
rs771042540 Chr.6: 121448294-121448294 on Build GRCh38
rs886061012 Chr.6: 121448187-121448187 on Build GRCh38
rs573918353 Chr.6: 121448369-121448369 on Build GRCh38
rs72548742 Chr.6: 121448115-121448115 on Build GRCh38
rs138314962 Chr.6: 121448161-121448161 on Build GRCh38
rs72548744 Chr.6: 121448169-121448169 on Build GRCh38
rs776536702 Chr.6: 121448338-121448338 on Build GRCh38
rs555726181 Chr.6: 121448450-121448450 on Build GRCh38
rs774503143 Chr.6: 121448298-121448298 on Build GRCh38
rs749664900 Chr.6: 121448272-121448272 on Build GRCh38
rs978398566 Chr.6: 121448101-121448101 on Build GRCh38
rs768996173 Chr.6: 121448279-121448279 on Build GRCh38
rs542131377 Chr.6: 121448387-121448387 on Build GRCh38
rs72548743 Chr.6: 121448150-121448150 on Build GRCh38
rs755999137 Chr.6: 121448032-121448032 on Build GRCh38

View Full Product Details