Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 131164964-131165485 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AKAP7
Entrez Gene ID
9465
Gene Name
A-kinase anchoring protein 7
Gene Aliases
AKAP15, AKAP18
UniGene
Hs.486483
Gene Chromosome Location
Chr.6: 131125698-131283535 on Build GRCh38
Transcripts
NM_016377.3 XM_005267228.1 XM_005267229.4 XM_006715614.2 XM_011536262.1 XM_017011506.1 XM_017011507.1 XM_017011508.1 XM_017011509.1 XM_017011510.1 XM_017011511.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6847438 c.328G>T p.V110L Chr.6: 131165183-131165183 on Build GRCh38
3173045 c.294G>A p.M98I Chr.6: 131165149-131165149 on Build GRCh38
6597788 c.289G>A p.A97T Chr.6: 131165144-131165144 on Build GRCh38
1073234 c.330G>A p.V110V Chr.6: 131165185-131165185 on Build GRCh38
3858257 c.334C>T p.Q112* Chr.6: 131165189-131165189 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs367995839 Chr.6: 131165136-131165136 on Build GRCh38
rs770883703 Chr.6: 131165035-131165035 on Build GRCh38
rs138197745 Chr.6: 131165190-131165190 on Build GRCh38
rs893619194 Chr.6: 131165282-131165282 on Build GRCh38
rs779533383 Chr.6: 131165155-131165155 on Build GRCh38
rs753599905 Chr.6: 131165225-131165225 on Build GRCh38
rs756679989 Chr.6: 131165376-131165376 on Build GRCh38
rs751971878 Chr.6: 131165267-131165267 on Build GRCh38
rs765653232 Chr.6: 131165084-131165084 on Build GRCh38
rs866840803 Chr.6: 131165451-131165451 on Build GRCh38
rs367723076 Chr.6: 131165039-131165039 on Build GRCh38
rs565826354 Chr.6: 131165093-131165093 on Build GRCh38
rs757022435 Chr.6: 131165227-131165227 on Build GRCh38
rs72991518 Chr.6: 131165326-131165326 on Build GRCh38
rs992348429 Chr.6: 131165327-131165327 on Build GRCh38
rs760118946 Chr.6: 131165209-131165209 on Build GRCh38
rs753395517 Chr.6: 131165355-131165355 on Build GRCh38
rs369455622 Chr.6: 131165266-131165266 on Build GRCh38
rs767806916 Chr.6: 131165264-131165268 on Build GRCh38
rs111657155 Chr.6: 131165197-131165197 on Build GRCh38
rs200670324 Chr.6: 131165194-131165194 on Build GRCh38
rs930359112 Chr.6: 131165392-131165392 on Build GRCh38
rs752803625 Chr.6: 131165217-131165217 on Build GRCh38
rs1049170160 Chr.6: 131165447-131165447 on Build GRCh38
rs746717383 Chr.6: 131165157-131165157 on Build GRCh38
rs753036812 Chr.6: 131165123-131165123 on Build GRCh38
rs762166997 Chr.6: 131165077-131165077 on Build GRCh38
rs767857459 Chr.6: 131165122-131165122 on Build GRCh38
rs145909307 Chr.6: 131165132-131165132 on Build GRCh38
rs953413239 Chr.6: 131165330-131165330 on Build GRCh38
rs547392548 Chr.6: 131165067-131165067 on Build GRCh38
rs767764609 Chr.6: 131165212-131165212 on Build GRCh38
rs1033941626 Chr.6: 131165296-131165296 on Build GRCh38
rs768331934 Chr.6: 131165169-131165169 on Build GRCh38
rs961828542 Chr.6: 131165367-131165367 on Build GRCh38
rs764340724 Chr.6: 131165224-131165224 on Build GRCh38
rs367987201 Chr.6: 131165242-131165242 on Build GRCh38
rs761021276 Chr.6: 131165218-131165218 on Build GRCh38
rs556592558 Chr.6: 131165253-131165253 on Build GRCh38
rs949761349 Chr.6: 131165033-131165033 on Build GRCh38
rs560107261 Chr.6: 131165450-131165450 on Build GRCh38
rs940426034 Chr.6: 131165036-131165036 on Build GRCh38
rs889022875 Chr.6: 131165179-131165179 on Build GRCh38
rs371809183 Chr.6: 131165189-131165189 on Build GRCh38
rs557843005 Chr.6: 131165133-131165133 on Build GRCh38
rs768224151 Chr.6: 131165255-131165255 on Build GRCh38
rs758313930 Chr.6: 131165249-131165249 on Build GRCh38
rs971861581 Chr.6: 131165370-131165370 on Build GRCh38
rs755856559 Chr.6: 131165318-131165318 on Build GRCh38
rs763334991 Chr.6: 131165188-131165188 on Build GRCh38
rs1001267887 Chr.6: 131165291-131165291 on Build GRCh38
rs771737400 Chr.6: 131165068-131165068 on Build GRCh38
rs1052224075 Chr.6: 131165245-131165245 on Build GRCh38
rs751620207 Chr.6: 131165115-131165115 on Build GRCh38
rs763418978 Chr.6: 131165099-131165099 on Build GRCh38
rs897516314 Chr.6: 131165059-131165059 on Build GRCh38
rs763532678 Chr.6: 131165145-131165145 on Build GRCh38
rs141586832 Chr.6: 131165129-131165129 on Build GRCh38
rs879102566 Chr.6: 131165301-131165301 on Build GRCh38
rs777478905 Chr.6: 131165131-131165131 on Build GRCh38
rs920720789 Chr.6: 131165064-131165064 on Build GRCh38
rs766878118 Chr.6: 131165114-131165114 on Build GRCh38
rs917007078 Chr.6: 131165019-131165019 on Build GRCh38
rs568311261 Chr.6: 131165017-131165017 on Build GRCh38
rs377503182 Chr.6: 131165460-131165460 on Build GRCh38
rs541869682 Chr.6: 131165389-131165389 on Build GRCh38
rs759638372 Chr.6: 131165119-131165119 on Build GRCh38
rs751674969 Chr.6: 131165436-131165436 on Build GRCh38
rs775062460 Chr.6: 131165069-131165069 on Build GRCh38
rs771500259 Chr.6: 131165147-131165147 on Build GRCh38
rs778716732 Chr.6: 131165228-131165228 on Build GRCh38
rs773488714 Chr.6: 131165187-131165187 on Build GRCh38
rs779584725 Chr.6: 131165250-131165250 on Build GRCh38

View Full Product Details