Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 131573122-131573635 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARG1
Entrez Gene ID
383
Gene Name
arginase 1
Gene Aliases
-
UniGene
Hs.440934
Gene Chromosome Location
Chr.6: 131573159-131584332 on Build GRCh38
Transcripts
NM_000045.3 NM_001244438.1 XM_011535801.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3702892 c.50A>C p.K17T Chr.6: 131573332-131573332 on Build GRCh38
4416324 c.57G>A p.Q19Q Chr.6: 131573339-131573339 on Build GRCh38
136571 c.26G>A p.G9E Chr.6: 131573308-131573308 on Build GRCh38
4423997 c.32T>C p.I11T Chr.6: 131573314-131573314 on Build GRCh38
145107 c.45C>A p.F15L Chr.6: 131573327-131573327 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1017005620 Chr.6: 131573191-131573191 on Build GRCh38
rs762277405 Chr.6: 131573214-131573214 on Build GRCh38
rs747646903 Chr.6: 131573386-131573386 on Build GRCh38
rs903578288 Chr.6: 131573440-131573440 on Build GRCh38
rs149935317 Chr.6: 131573521-131573521 on Build GRCh38
rs778014573 Chr.6: 131573354-131573354 on Build GRCh38
rs766705948 Chr.6: 131573246-131573246 on Build GRCh38
rs745624953 Chr.6: 131573285-131573285 on Build GRCh38
rs941313134 Chr.6: 131573492-131573492 on Build GRCh38
rs17788484 Chr.6: 131573218-131573218 on Build GRCh38
rs28941474 Chr.6: 131573314-131573314 on Build GRCh38
rs147419581 Chr.6: 131573317-131573317 on Build GRCh38
rs757187110 Chr.6: 131573346-131573346 on Build GRCh38
rs928272810 Chr.6: 131573412-131573412 on Build GRCh38
rs774603245 Chr.6: 131573306-131573306 on Build GRCh38
rs1008749393 Chr.6: 131573596-131573596 on Build GRCh38
rs374231313 Chr.6: 131573376-131573376 on Build GRCh38
rs753260969 Chr.6: 131573333-131573333 on Build GRCh38
rs768686705 Chr.6: 131573373-131573373 on Build GRCh38
rs892281308 Chr.6: 131573426-131573426 on Build GRCh38
rs887220299 Chr.6: 131573147-131573147 on Build GRCh38
rs532764881 Chr.6: 131573600-131573600 on Build GRCh38
rs767964945 Chr.6: 131573257-131573257 on Build GRCh38
rs779135201 Chr.6: 131573351-131573351 on Build GRCh38
rs750244206 Chr.6: 131573360-131573360 on Build GRCh38
rs376196920 Chr.6: 131573431-131573431 on Build GRCh38
rs919450094 Chr.6: 131573535-131573535 on Build GRCh38
rs1005622236 Chr.6: 131573181-131573181 on Build GRCh38
rs761516831 Chr.6: 131573240-131573240 on Build GRCh38
rs929474852 Chr.6: 131573562-131573562 on Build GRCh38
rs939256919 Chr.6: 131573324-131573324 on Build GRCh38
rs558377642 Chr.6: 131573289-131573289 on Build GRCh38
rs987130050 Chr.6: 131573275-131573275 on Build GRCh38
rs899876407 Chr.6: 131573520-131573520 on Build GRCh38
rs587776539 Chr.6: 131573340-131573340 on Build GRCh38
rs9493029 Chr.6: 131573534-131573534 on Build GRCh38
rs903547112 Chr.6: 131573203-131573203 on Build GRCh38
rs565522210 Chr.6: 131573569-131573569 on Build GRCh38
rs192988987 Chr.6: 131573533-131573533 on Build GRCh38
rs761148101 Chr.6: 131573338-131573338 on Build GRCh38
rs752788818 Chr.6: 131573259-131573259 on Build GRCh38
rs976317468 Chr.6: 131573230-131573230 on Build GRCh38
rs984931246 Chr.6: 131573575-131573575 on Build GRCh38
rs138584410 Chr.6: 131573288-131573288 on Build GRCh38
rs1009256260 Chr.6: 131573167-131573167 on Build GRCh38
rs879127897 Chr.6: 131573205-131573206 on Build GRCh38
rs758330818 Chr.6: 131573364-131573364 on Build GRCh38
rs1028910953 Chr.6: 131573219-131573219 on Build GRCh38
rs776217305 Chr.6: 131573236-131573236 on Build GRCh38
rs781171804 Chr.6: 131573377-131573381 on Build GRCh38
rs757468683 Chr.6: 131573279-131573279 on Build GRCh38
rs776185609 Chr.6: 131573304-131573304 on Build GRCh38
rs1047118722 Chr.6: 131573461-131573461 on Build GRCh38
rs890405423 Chr.6: 131573572-131573572 on Build GRCh38
rs757439680 Chr.6: 131573348-131573348 on Build GRCh38
rs573069792 Chr.6: 131573153-131573153 on Build GRCh38
rs149310631 Chr.6: 131573305-131573305 on Build GRCh38
rs192353153 Chr.6: 131573213-131573213 on Build GRCh38
rs183249041 Chr.6: 131573473-131573473 on Build GRCh38
rs779986367 Chr.6: 131573366-131573366 on Build GRCh38
rs778602133 Chr.6: 131573282-131573282 on Build GRCh38
rs755511669 Chr.6: 131573252-131573252 on Build GRCh38
rs370747314 Chr.6: 131573262-131573262 on Build GRCh38
rs188662464 Chr.6: 131573496-131573496 on Build GRCh38
rs777817531 Chr.6: 131573263-131573263 on Build GRCh38
rs989204710 Chr.6: 131573301-131573301 on Build GRCh38
rs768343568 Chr.6: 131573295-131573295 on Build GRCh38
rs945187879 Chr.6: 131573429-131573429 on Build GRCh38
rs1007229965 Chr.6: 131573450-131573450 on Build GRCh38
rs533598114 Chr.6: 131573504-131573504 on Build GRCh38
rs150766204 Chr.6: 131573339-131573339 on Build GRCh38
rs780600712 Chr.6: 131573379-131573379 on Build GRCh38
rs751924194 Chr.6: 131573250-131573250 on Build GRCh38
rs865842223 Chr.6: 131573228-131573228 on Build GRCh38
rs139165092 Chr.6: 131573476-131573476 on Build GRCh38
rs749521049 Chr.6: 131573277-131573277 on Build GRCh38

View Full Product Details