Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 131576306-131576826 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARG1
Entrez Gene ID
383
Gene Name
arginase 1
Gene Aliases
-
UniGene
Hs.440934
Gene Chromosome Location
Chr.6: 131573159-131584332 on Build GRCh38
Transcripts
NM_000045.3 NM_001244438.1 XM_011535801.2 NM_001270521.1 NM_015979.3

Gene Symbol
MED23
Entrez Gene ID
9439
Gene Name
mediator complex subunit 23
Gene Aliases
ARC130, CRSP130, CRSP133, CRSP3, DRIP130, MRT18, SUR-2, SUR2
UniGene
Hs.29679
Gene Chromosome Location
Chr.6: 131573966-131628239 on Build GRCh38
Transcripts
NM_000045.3 NM_001244438.1 XM_011535801.2 NM_001270521.1 NM_015979.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6602972 c.89T>C p.V30A Chr.6: 131576694-131576694 on Build GRCh38
1073242 c.58C>T p.P20S Chr.6: 131576663-131576663 on Build GRCh38
6602966 c.114G>T p.E38D Chr.6: 131576719-131576719 on Build GRCh38
69701 c.70G>T p.V24L Chr.6: 131576675-131576675 on Build GRCh38
6602965 c.114G>T p.E38D Chr.6: 131576719-131576719 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs972691825 Chr.6: 131576441-131576441 on Build GRCh38
rs913418921 Chr.6: 131576625-131576625 on Build GRCh38
rs376050897 Chr.6: 131576356-131576356 on Build GRCh38
rs920554932 Chr.6: 131576559-131576559 on Build GRCh38
rs930551837 Chr.6: 131576577-131576577 on Build GRCh38
rs952416837 Chr.6: 131576470-131576470 on Build GRCh38
rs756915518 Chr.6: 131576747-131576747 on Build GRCh38
rs73778020 Chr.6: 131576370-131576370 on Build GRCh38
rs763301564 Chr.6: 131576656-131576656 on Build GRCh38
rs770529240 Chr.6: 131576619-131576619 on Build GRCh38
rs1045461625 Chr.6: 131576679-131576679 on Build GRCh38
rs1055653258 Chr.6: 131576597-131576597 on Build GRCh38
rs908604424 Chr.6: 131576482-131576482 on Build GRCh38
rs745500077 Chr.6: 131576767-131576767 on Build GRCh38
rs755973004 Chr.6: 131576680-131576680 on Build GRCh38
rs368656171 Chr.6: 131576757-131576757 on Build GRCh38
rs755975244 Chr.6: 131576698-131576698 on Build GRCh38
rs1014367103 Chr.6: 131576788-131576788 on Build GRCh38
rs766662245 Chr.6: 131576664-131576664 on Build GRCh38
rs769118957 Chr.6: 131576774-131576774 on Build GRCh38
rs556545273 Chr.6: 131576375-131576375 on Build GRCh38
rs998513101 Chr.6: 131576681-131576681 on Build GRCh38
rs201959088 Chr.6: 131576631-131576631 on Build GRCh38
rs104893944 Chr.6: 131576666-131576666 on Build GRCh38
rs532250558 Chr.6: 131576475-131576475 on Build GRCh38
rs1056187104 Chr.6: 131576731-131576731 on Build GRCh38
rs748979820 Chr.6: 131576617-131576617 on Build GRCh38
rs551387878 Chr.6: 131576349-131576349 on Build GRCh38
rs113257850 Chr.6: 131576737-131576737 on Build GRCh38
rs975648163 Chr.6: 131576447-131576447 on Build GRCh38
rs1028892301 Chr.6: 131576455-131576455 on Build GRCh38
rs775853882 Chr.6: 131576621-131576621 on Build GRCh38
rs780487191 Chr.6: 131576647-131576647 on Build GRCh38
rs974034404 Chr.6: 131576538-131576538 on Build GRCh38
rs1057519231 Chr.6: 131576688-131576688 on Build GRCh38
rs144994895 Chr.6: 131576667-131576667 on Build GRCh38
rs183896871 Chr.6: 131576367-131576367 on Build GRCh38
rs769045782 Chr.6: 131576629-131576629 on Build GRCh38
rs779949397 Chr.6: 131576727-131576729 on Build GRCh38
rs767211950 Chr.6: 131576676-131576676 on Build GRCh38
rs748744950 Chr.6: 131576729-131576729 on Build GRCh38
rs773399859 Chr.6: 131576653-131576653 on Build GRCh38
rs762210089 Chr.6: 131576646-131576646 on Build GRCh38
rs976910251 Chr.6: 131576501-131576501 on Build GRCh38
rs575110162 Chr.6: 131576417-131576417 on Build GRCh38
rs113767658 Chr.6: 131576736-131576736 on Build GRCh38
rs747313511 Chr.6: 131576628-131576628 on Build GRCh38
rs943774985 Chr.6: 131576487-131576487 on Build GRCh38
rs138457449 Chr.6: 131576520-131576520 on Build GRCh38
rs1027285402 Chr.6: 131576443-131576443 on Build GRCh38
rs867672125 Chr.6: 131576477-131576477 on Build GRCh38

View Full Product Details