Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 131578850-131579355 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARG1
Entrez Gene ID
383
Gene Name
arginase 1
Gene Aliases
-
UniGene
Hs.440934
Gene Chromosome Location
Chr.6: 131573159-131584332 on Build GRCh38
Transcripts
NM_000045.3 NM_001244438.1 XM_011535801.2 NM_001270521.1 NM_015979.3

Gene Symbol
MED23
Entrez Gene ID
9439
Gene Name
mediator complex subunit 23
Gene Aliases
ARC130, CRSP130, CRSP133, CRSP3, DRIP130, MRT18, SUR-2, SUR2
UniGene
Hs.29679
Gene Chromosome Location
Chr.6: 131573966-131628239 on Build GRCh38
Transcripts
NM_000045.3 NM_001244438.1 XM_011535801.2 NM_001270521.1 NM_015979.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5869292 c.221G>A p.G74E Chr.6: 131579201-131579201 on Build GRCh38
1073243 c.215C>A p.S72Y Chr.6: 131579195-131579195 on Build GRCh38
6193362 c.294C>T p.G98G Chr.6: 131579274-131579274 on Build GRCh38
6297555 c.245G>T p.G82V Chr.6: 131579225-131579225 on Build GRCh38
360236 c.283C>A p.L95M Chr.6: 131579263-131579263 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs915361516 Chr.6: 131578939-131578939 on Build GRCh38
rs763960889 Chr.6: 131579156-131579156 on Build GRCh38
rs185623005 Chr.6: 131578940-131578940 on Build GRCh38
rs983224088 Chr.6: 131579313-131579313 on Build GRCh38
rs760319794 Chr.6: 131579269-131579269 on Build GRCh38
rs988680495 Chr.6: 131578936-131578936 on Build GRCh38
rs763868954 Chr.6: 131579274-131579274 on Build GRCh38
rs752395217 Chr.6: 131579140-131579140 on Build GRCh38
rs1022874044 Chr.6: 131579233-131579233 on Build GRCh38
rs773454478 Chr.6: 131578909-131578909 on Build GRCh38
rs749556084 Chr.6: 131579040-131579041 on Build GRCh38
rs377721661 Chr.6: 131579318-131579318 on Build GRCh38
rs144544360 Chr.6: 131578920-131578920 on Build GRCh38
rs999627155 Chr.6: 131579020-131579020 on Build GRCh38
rs753829097 Chr.6: 131579275-131579275 on Build GRCh38
rs756616090 Chr.6: 131579321-131579321 on Build GRCh38
rs982341943 Chr.6: 131578910-131578910 on Build GRCh38
rs745580041 Chr.6: 131579190-131579190 on Build GRCh38
rs928930827 Chr.6: 131579292-131579292 on Build GRCh38
rs112432420 Chr.6: 131579286-131579286 on Build GRCh38
rs756825623 Chr.6: 131579167-131579167 on Build GRCh38
rs766715122 Chr.6: 131579104-131579104 on Build GRCh38
rs748734055 Chr.6: 131579039-131579039 on Build GRCh38
rs753347547 Chr.6: 131579157-131579157 on Build GRCh38
rs956942732 Chr.6: 131579191-131579191 on Build GRCh38
rs1032803752 Chr.6: 131579059-131579059 on Build GRCh38
rs147146526 Chr.6: 131579150-131579150 on Build GRCh38
rs572633761 Chr.6: 131578914-131578914 on Build GRCh38
rs763070677 Chr.6: 131579092-131579092 on Build GRCh38
rs1047911283 Chr.6: 131578935-131578935 on Build GRCh38
rs148619575 Chr.6: 131579217-131579217 on Build GRCh38
rs929051884 Chr.6: 131578931-131578931 on Build GRCh38
rs796051923 Chr.6: 131579252-131579253 on Build GRCh38
rs768972828 Chr.6: 131579099-131579104 on Build GRCh38
rs761712444 Chr.6: 131579290-131579290 on Build GRCh38
rs1023355192 Chr.6: 131579060-131579060 on Build GRCh38
rs753111447 Chr.6: 131579320-131579320 on Build GRCh38
rs375655714 Chr.6: 131579124-131579124 on Build GRCh38
rs1003121552 Chr.6: 131579109-131579110 on Build GRCh38
rs188888028 Chr.6: 131579137-131579137 on Build GRCh38
rs370469825 Chr.6: 131578917-131578917 on Build GRCh38
rs910570845 Chr.6: 131578956-131578956 on Build GRCh38
rs34724840 Chr.6: 131579261-131579261 on Build GRCh38
rs56325116 Chr.6: 131579004-131579004 on Build GRCh38
rs764770439 Chr.6: 131579293-131579293 on Build GRCh38
rs147240575 Chr.6: 131578985-131578985 on Build GRCh38
rs544798219 Chr.6: 131578899-131578899 on Build GRCh38
rs142107090 Chr.6: 131579221-131579221 on Build GRCh38
rs571619062 Chr.6: 131579214-131579214 on Build GRCh38
rs531117440 Chr.6: 131579014-131579014 on Build GRCh38
rs535161157 Chr.6: 131579139-131579139 on Build GRCh38
rs886061060 Chr.6: 131579292-131579292 on Build GRCh38
rs111640755 Chr.6: 131579165-131579165 on Build GRCh38
rs373279537 Chr.6: 131579330-131579330 on Build GRCh38
rs55706510 Chr.6: 131579005-131579005 on Build GRCh38
rs957228613 Chr.6: 131579089-131579089 on Build GRCh38
rs1039301718 Chr.6: 131578951-131578951 on Build GRCh38
rs34504481 Chr.6: 131579250-131579250 on Build GRCh38
rs749865319 Chr.6: 131579309-131579309 on Build GRCh38
rs942154661 Chr.6: 131578948-131578948 on Build GRCh38
rs773451145 Chr.6: 131579073-131579073 on Build GRCh38
rs372415368 Chr.6: 131579115-131579115 on Build GRCh38
rs369393679 Chr.6: 131579211-131579211 on Build GRCh38
rs748414362 Chr.6: 131579196-131579196 on Build GRCh38
rs990089154 Chr.6: 131578894-131578894 on Build GRCh38
rs386705910 Chr.6: 131579004-131579005 on Build GRCh38
rs368978412 Chr.6: 131579303-131579303 on Build GRCh38
rs547122192 Chr.6: 131579212-131579212 on Build GRCh38
rs774273532 Chr.6: 131579225-131579225 on Build GRCh38
rs901057034 Chr.6: 131579001-131579001 on Build GRCh38
rs997503148 Chr.6: 131578988-131578988 on Build GRCh38
rs888226124 Chr.6: 131579006-131579006 on Build GRCh38
rs111885649 Chr.6: 131579329-131579330 on Build GRCh38
rs757959356 Chr.6: 131579192-131579192 on Build GRCh38
rs867223787 Chr.6: 131579198-131579198 on Build GRCh38
rs374676787 Chr.6: 131579251-131579251 on Build GRCh38
rs549339306 Chr.6: 131579062-131579062 on Build GRCh38
rs778376916 Chr.6: 131579176-131579176 on Build GRCh38
rs759832639 Chr.6: 131579246-131579246 on Build GRCh38
rs1063493 Chr.6: 131579236-131579236 on Build GRCh38
rs900757382 Chr.6: 131578968-131578968 on Build GRCh38
rs774347014 Chr.6: 131579107-131579109 on Build GRCh38
rs758094530 Chr.6: 131579310-131579310 on Build GRCh38
rs904884112 Chr.6: 131579070-131579070 on Build GRCh38
rs772521719 Chr.6: 131579271-131579272 on Build GRCh38

View Full Product Details