Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 131583879-131584426 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARG1
Entrez Gene ID
383
Gene Name
arginase 1
Gene Aliases
-
UniGene
Hs.440934
Gene Chromosome Location
Chr.6: 131573159-131584332 on Build GRCh38
Transcripts
NM_000045.3 NM_001244438.1 XM_011535801.2 NM_001270521.1 NM_015979.3

Gene Symbol
MED23
Entrez Gene ID
9439
Gene Name
mediator complex subunit 23
Gene Aliases
ARC130, CRSP130, CRSP133, CRSP3, DRIP130, MRT18, SUR-2, SUR2
UniGene
Hs.29679
Gene Chromosome Location
Chr.6: 131573966-131628239 on Build GRCh38
Transcripts
NM_000045.3 NM_001244438.1 XM_011535801.2 NM_001270521.1 NM_015979.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3173108 c.949T>G p.Y317D Chr.6: 131583888-131583888 on Build GRCh38
5562165 c.963T>G p.P321P Chr.6: 131583902-131583902 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs545874005 Chr.6: 131584365-131584365 on Build GRCh38
rs909797455 Chr.6: 131584202-131584202 on Build GRCh38
rs891605184 Chr.6: 131584374-131584374 on Build GRCh38
rs779017704 Chr.6: 131583946-131583946 on Build GRCh38
rs1021313033 Chr.6: 131584104-131584104 on Build GRCh38
rs777029258 Chr.6: 131583921-131583921 on Build GRCh38
rs1039354514 Chr.6: 131584234-131584234 on Build GRCh38
rs973308077 Chr.6: 131584212-131584212 on Build GRCh38
rs746107017 Chr.6: 131583947-131583947 on Build GRCh38
rs568284407 Chr.6: 131584180-131584180 on Build GRCh38
rs903391091 Chr.6: 131584338-131584338 on Build GRCh38
rs1021344046 Chr.6: 131584396-131584397 on Build GRCh38
rs1803151 Chr.6: 131584030-131584030 on Build GRCh38
rs1048340568 Chr.6: 131584196-131584196 on Build GRCh38
rs944253301 Chr.6: 131584209-131584209 on Build GRCh38
rs967511482 Chr.6: 131584015-131584015 on Build GRCh38
rs1003113919 Chr.6: 131583931-131583931 on Build GRCh38
rs769831041 Chr.6: 131584365-131584366 on Build GRCh38
rs749799579 Chr.6: 131583932-131583932 on Build GRCh38
rs371686924 Chr.6: 131584396-131584396 on Build GRCh38
rs1039895471 Chr.6: 131583927-131583927 on Build GRCh38
rs879734165 Chr.6: 131584371-131584371 on Build GRCh38
rs958923925 Chr.6: 131584373-131584373 on Build GRCh38
rs768028958 Chr.6: 131583922-131583922 on Build GRCh38
rs371339129 Chr.6: 131584363-131584364 on Build GRCh38
rs991492256 Chr.6: 131584097-131584097 on Build GRCh38
rs199864297 Chr.6: 131584364-131584367 on Build GRCh38
rs1035699012 Chr.6: 131584268-131584279 on Build GRCh38
rs763072469 Chr.6: 131584377-131584377 on Build GRCh38
rs553833088 Chr.6: 131584263-131584263 on Build GRCh38
rs912772679 Chr.6: 131584197-131584198 on Build GRCh38
rs778359686 Chr.6: 131583930-131583930 on Build GRCh38
rs972527136 Chr.6: 131584155-131584155 on Build GRCh38
rs867873385 Chr.6: 131584381-131584381 on Build GRCh38
rs1011346738 Chr.6: 131584394-131584395 on Build GRCh38
rs971523115 Chr.6: 131584110-131584110 on Build GRCh38
rs144214372 Chr.6: 131584364-131584365 on Build GRCh38
rs556580014 Chr.6: 131583925-131583925 on Build GRCh38
rs907113409 Chr.6: 131584337-131584338 on Build GRCh38
rs998785714 Chr.6: 131583957-131583957 on Build GRCh38
rs537322176 Chr.6: 131584325-131584325 on Build GRCh38
rs554233544 Chr.6: 131584261-131584261 on Build GRCh38
rs1031460462 Chr.6: 131583992-131583993 on Build GRCh38
rs1020469542 Chr.6: 131583963-131583963 on Build GRCh38
rs3138772 Chr.6: 131584365-131584366 on Build GRCh38
rs886061062 Chr.6: 131583981-131583981 on Build GRCh38
rs572234646 Chr.6: 131584273-131584273 on Build GRCh38
rs757327323 Chr.6: 131583936-131583936 on Build GRCh38
rs1033260259 Chr.6: 131584023-131584023 on Build GRCh38
rs1000451954 Chr.6: 131584017-131584017 on Build GRCh38
rs997958923 Chr.6: 131584294-131584294 on Build GRCh38
rs1031906792 Chr.6: 131584339-131584339 on Build GRCh38
rs1041819461 Chr.6: 131584328-131584328 on Build GRCh38
rs142812031 Chr.6: 131584218-131584218 on Build GRCh38
rs917342667 Chr.6: 131584103-131584103 on Build GRCh38
rs1044356580 Chr.6: 131584286-131584286 on Build GRCh38
rs953667116 Chr.6: 131584173-131584173 on Build GRCh38
rs779162184 Chr.6: 131584284-131584284 on Build GRCh38
rs918425492 Chr.6: 131584163-131584163 on Build GRCh38
rs958728303 Chr.6: 131584042-131584042 on Build GRCh38
rs985522029 Chr.6: 131584399-131584401 on Build GRCh38

View Full Product Details