Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 145800205-145800717 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FBXO30
Entrez Gene ID
84085
Gene Name
F-box protein 30
Gene Aliases
Fbx30
UniGene
Hs.421095
Gene Chromosome Location
Chr.6: 145798135-145864409 on Build GRCh38
Transcripts
NM_032145.4 XM_005267159.3 XM_011536177.2 XM_017011353.1 NR_038246.1

Gene Symbol
LOC100507557
Entrez Gene ID
100507557
Gene Name
uncharacterized LOC100507557
Gene Aliases
-
UniGene
Hs.124537
Gene Chromosome Location
Chr.6: 145734869-145886585 on Build GRCh38
Transcripts
NM_032145.4 XM_005267159.3 XM_011536177.2 XM_017011353.1 NR_038246.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3858823 c.2068A>G p.N690D Chr.6: 145800276-145800276 on Build GRCh38
4160265 c.2057T>G p.F686C Chr.6: 145800287-145800287 on Build GRCh38
4187448 c.2069A>G p.N690S Chr.6: 145800275-145800275 on Build GRCh38
7275875 c.2126A>G p.Y709C Chr.6: 145800218-145800218 on Build GRCh38
1440956 c.2068A>C p.N690H Chr.6: 145800276-145800276 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs973446356 Chr.6: 145800326-145800326 on Build GRCh38
rs562141490 Chr.6: 145800515-145800515 on Build GRCh38
rs9386131 Chr.6: 145800472-145800472 on Build GRCh38
rs781108999 Chr.6: 145800276-145800276 on Build GRCh38
rs138288045 Chr.6: 145800302-145800302 on Build GRCh38
rs760979203 Chr.6: 145800327-145800332 on Build GRCh38
rs770424255 Chr.6: 145800323-145800323 on Build GRCh38
rs1033007346 Chr.6: 145800663-145800664 on Build GRCh38
rs1041697738 Chr.6: 145800418-145800418 on Build GRCh38
rs547252903 Chr.6: 145800627-145800627 on Build GRCh38
rs377589494 Chr.6: 145800552-145800552 on Build GRCh38
rs769563792 Chr.6: 145800296-145800296 on Build GRCh38
rs1015159636 Chr.6: 145800683-145800683 on Build GRCh38
rs766742306 Chr.6: 145800360-145800360 on Build GRCh38
rs574425293 Chr.6: 145800238-145800238 on Build GRCh38
rs775893546 Chr.6: 145800347-145800347 on Build GRCh38
rs1037937384 Chr.6: 145800509-145800509 on Build GRCh38
rs917141505 Chr.6: 145800406-145800406 on Build GRCh38
rs1001162482 Chr.6: 145800657-145800657 on Build GRCh38
rs556702677 Chr.6: 145800303-145800303 on Build GRCh38
rs866615839 Chr.6: 145800451-145800451 on Build GRCh38
rs528427437 Chr.6: 145800448-145800448 on Build GRCh38
rs931823464 Chr.6: 145800539-145800540 on Build GRCh38
rs559396833 Chr.6: 145800404-145800404 on Build GRCh38
rs945696874 Chr.6: 145800576-145800576 on Build GRCh38
rs201104889 Chr.6: 145800351-145800351 on Build GRCh38
rs764774375 Chr.6: 145800639-145800639 on Build GRCh38
rs902835304 Chr.6: 145800432-145800432 on Build GRCh38
rs567046920 Chr.6: 145800646-145800646 on Build GRCh38
rs184413290 Chr.6: 145800390-145800390 on Build GRCh38
rs950002415 Chr.6: 145800415-145800415 on Build GRCh38
rs902753955 Chr.6: 145800613-145800613 on Build GRCh38
rs533572625 Chr.6: 145800617-145800617 on Build GRCh38
rs1009546017 Chr.6: 145800674-145800674 on Build GRCh38
rs747858191 Chr.6: 145800284-145800284 on Build GRCh38
rs768371521 Chr.6: 145800253-145800253 on Build GRCh38
rs551111638 Chr.6: 145800583-145800583 on Build GRCh38
rs761222558 Chr.6: 145800349-145800349 on Build GRCh38
rs762381831 Chr.6: 145800307-145800307 on Build GRCh38
rs746929294 Chr.6: 145800250-145800250 on Build GRCh38
rs895289480 Chr.6: 145800680-145800680 on Build GRCh38
rs996324812 Chr.6: 145800553-145800553 on Build GRCh38
rs189052357 Chr.6: 145800541-145800541 on Build GRCh38
rs753245980 Chr.6: 145800514-145800514 on Build GRCh38
rs772880851 Chr.6: 145800240-145800240 on Build GRCh38
rs920693369 Chr.6: 145800381-145800381 on Build GRCh38
rs549516669 Chr.6: 145800609-145800609 on Build GRCh38
rs962042613 Chr.6: 145800315-145800315 on Build GRCh38
rs1029567212 Chr.6: 145800575-145800575 on Build GRCh38

View Full Product Details