Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 150399597-150400139 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IYD
Entrez Gene ID
389434
Gene Name
iodotyrosine deiodinase
Gene Aliases
C6orf71, DEHAL1, IYD-1, TDH4
UniGene
Hs.310225
Gene Chromosome Location
Chr.6: 150368854-150404629 on Build GRCh38
Transcripts
NM_001164694.1 NM_001164695.1 NM_001318495.1 NM_203395.2 NR_134655.1

SNP Detail

SNP ID Chromosome Location
rs185342498 Chr.6: 150399974-150399974 on Build GRCh38
rs560878398 Chr.6: 150399811-150399811 on Build GRCh38
rs1055423688 Chr.6: 150399753-150399753 on Build GRCh38
rs1008073120 Chr.6: 150399796-150399796 on Build GRCh38
rs755090955 Chr.6: 150399704-150399704 on Build GRCh38
rs529938065 Chr.6: 150399849-150399849 on Build GRCh38
rs1016823011 Chr.6: 150400075-150400075 on Build GRCh38
rs796590097 Chr.6: 150399751-150399751 on Build GRCh38
rs1042206059 Chr.6: 150400005-150400005 on Build GRCh38
rs958565746 Chr.6: 150399647-150399647 on Build GRCh38
rs906100640 Chr.6: 150399794-150399794 on Build GRCh38
rs116064065 Chr.6: 150399938-150399938 on Build GRCh38
rs866995146 Chr.6: 150399992-150399992 on Build GRCh38
rs543815399 Chr.6: 150399934-150399934 on Build GRCh38
rs571428691 Chr.6: 150399642-150399642 on Build GRCh38
rs981126443 Chr.6: 150399692-150399692 on Build GRCh38
rs919563660 Chr.6: 150399959-150399959 on Build GRCh38
rs886061173 Chr.6: 150399991-150399991 on Build GRCh38
rs941097410 Chr.6: 150399699-150399699 on Build GRCh38
rs879258240 Chr.6: 150399852-150399853 on Build GRCh38
rs146165286 Chr.6: 150400070-150400070 on Build GRCh38
rs953927356 Chr.6: 150399891-150399891 on Build GRCh38
rs763425035 Chr.6: 150400009-150400009 on Build GRCh38
rs887034046 Chr.6: 150399990-150399990 on Build GRCh38
rs962701147 Chr.6: 150400092-150400092 on Build GRCh38
rs536906067 Chr.6: 150399997-150399997 on Build GRCh38
rs886061172 Chr.6: 150399625-150399626 on Build GRCh38
rs193155130 Chr.6: 150399654-150399654 on Build GRCh38
rs143064277 Chr.6: 150400067-150400067 on Build GRCh38
rs544793302 Chr.6: 150400041-150400041 on Build GRCh38
rs886914454 Chr.6: 150399795-150399795 on Build GRCh38
rs587713 Chr.6: 150400093-150400093 on Build GRCh38
rs374654397 Chr.6: 150400008-150400008 on Build GRCh38
rs899957925 Chr.6: 150399818-150399818 on Build GRCh38
rs659737 Chr.6: 150399853-150399853 on Build GRCh38
rs769705321 Chr.6: 150399793-150399793 on Build GRCh38
rs1029491137 Chr.6: 150400053-150400053 on Build GRCh38
rs72998948 Chr.6: 150399784-150399784 on Build GRCh38
rs949596831 Chr.6: 150399668-150399668 on Build GRCh38
rs35209272 Chr.6: 150399910-150399910 on Build GRCh38
rs988774945 Chr.6: 150399893-150399893 on Build GRCh38
rs660153 Chr.6: 150399772-150399772 on Build GRCh38
rs897312590 Chr.6: 150399760-150399760 on Build GRCh38
rs529950454 Chr.6: 150400115-150400115 on Build GRCh38
rs749689021 Chr.6: 150399881-150399882 on Build GRCh38
rs936967142 Chr.6: 150399722-150399722 on Build GRCh38
rs918354682 Chr.6: 150400097-150400097 on Build GRCh38
rs928394078 Chr.6: 150399720-150399720 on Build GRCh38
rs761836939 Chr.6: 150400083-150400083 on Build GRCh38
rs931249182 Chr.6: 150399783-150399783 on Build GRCh38
rs599339 Chr.6: 150399852-150399852 on Build GRCh38
rs866630028 Chr.6: 150399643-150399643 on Build GRCh38
rs1032444910 Chr.6: 150400060-150400060 on Build GRCh38
rs931009885 Chr.6: 150399966-150399966 on Build GRCh38
rs971268223 Chr.6: 150399918-150399918 on Build GRCh38
rs34257671 Chr.6: 150399951-150399951 on Build GRCh38
rs80030954 Chr.6: 150399999-150399999 on Build GRCh38
rs386707042 Chr.6: 150399852-150399853 on Build GRCh38
rs943849831 Chr.6: 150399991-150399991 on Build GRCh38
rs1019671109 Chr.6: 150399814-150399814 on Build GRCh38
rs534225787 Chr.6: 150399952-150399952 on Build GRCh38
rs901904891 Chr.6: 150400023-150400023 on Build GRCh38
rs146369688 Chr.6: 150399962-150399962 on Build GRCh38
rs1037236059 Chr.6: 150399709-150399709 on Build GRCh38
rs73619523 Chr.6: 150399648-150399648 on Build GRCh38
rs997998248 Chr.6: 150399863-150399863 on Build GRCh38
rs1047388971 Chr.6: 150399975-150399975 on Build GRCh38
rs1032593235 Chr.6: 150399644-150399644 on Build GRCh38
rs139736929 Chr.6: 150400043-150400043 on Build GRCh38

View Full Product Details