Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 161133824-161134336 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AGPAT4
Entrez Gene ID
56895
Gene Name
1-acylglycerol-3-phosphate O-acyltransferase 4
Gene Aliases
1-AGPAT4, LPAAT-delta, dJ473J16.2
UniGene
Hs.353175
Gene Chromosome Location
Chr.6: 161129980-161274108 on Build GRCh38
Transcripts
NM_020133.2 XM_005267053.3 XM_006715514.3 XM_017011059.1

SNP Detail

SNP ID Chromosome Location
rs781273183 Chr.6: 161134074-161134074 on Build GRCh38
rs919373249 Chr.6: 161134054-161134054 on Build GRCh38
rs371870807 Chr.6: 161133879-161133879 on Build GRCh38
rs558341660 Chr.6: 161134065-161134065 on Build GRCh38
rs912155858 Chr.6: 161134303-161134303 on Build GRCh38
rs757934819 Chr.6: 161133934-161133934 on Build GRCh38
rs1020619047 Chr.6: 161134197-161134197 on Build GRCh38
rs531007289 Chr.6: 161133999-161133999 on Build GRCh38
rs41268547 Chr.6: 161133901-161133901 on Build GRCh38
rs571538110 Chr.6: 161134010-161134010 on Build GRCh38
rs749484796 Chr.6: 161134199-161134200 on Build GRCh38
rs552977938 Chr.6: 161134152-161134152 on Build GRCh38
rs550803907 Chr.6: 161134101-161134101 on Build GRCh38
rs990540783 Chr.6: 161133998-161133998 on Build GRCh38
rs1036349413 Chr.6: 161134050-161134050 on Build GRCh38
rs1042293233 Chr.6: 161134011-161134011 on Build GRCh38
rs551988876 Chr.6: 161133898-161133898 on Build GRCh38
rs959078608 Chr.6: 161133975-161133975 on Build GRCh38
rs1000591172 Chr.6: 161134226-161134226 on Build GRCh38
rs542429188 Chr.6: 161134249-161134249 on Build GRCh38
rs960618721 Chr.6: 161133962-161133962 on Build GRCh38
rs978368565 Chr.6: 161134267-161134267 on Build GRCh38
rs967803880 Chr.6: 161134203-161134204 on Build GRCh38
rs73595013 Chr.6: 161134161-161134161 on Build GRCh38
rs374916985 Chr.6: 161134119-161134119 on Build GRCh38
rs1051727068 Chr.6: 161134034-161134034 on Build GRCh38
rs878913806 Chr.6: 161133997-161133997 on Build GRCh38
rs995083047 Chr.6: 161133912-161133912 on Build GRCh38
rs986746591 Chr.6: 161134026-161134026 on Build GRCh38
rs890474338 Chr.6: 161134049-161134049 on Build GRCh38
rs1007656236 Chr.6: 161133853-161133853 on Build GRCh38
rs1034616776 Chr.6: 161134153-161134153 on Build GRCh38
rs927637960 Chr.6: 161133969-161133969 on Build GRCh38
rs774825742 Chr.6: 161134014-161134014 on Build GRCh38
rs946564565 Chr.6: 161134006-161134006 on Build GRCh38
rs754578244 Chr.6: 161134042-161134042 on Build GRCh38
rs796784074 Chr.6: 161134012-161134012 on Build GRCh38
rs990509782 Chr.6: 161134167-161134167 on Build GRCh38
rs1027013597 Chr.6: 161133913-161133913 on Build GRCh38
rs934675257 Chr.6: 161134025-161134025 on Build GRCh38
rs1033178728 Chr.6: 161134239-161134239 on Build GRCh38
rs773093218 Chr.6: 161134306-161134306 on Build GRCh38
rs923799843 Chr.6: 161134311-161134311 on Build GRCh38
rs1026566437 Chr.6: 161134137-161134137 on Build GRCh38
rs1034695453 Chr.6: 161133944-161133946 on Build GRCh38
rs189622386 Chr.6: 161134138-161134138 on Build GRCh38
rs986307457 Chr.6: 161134291-161134291 on Build GRCh38
rs759489016 Chr.6: 161133931-161133933 on Build GRCh38
rs1039161430 Chr.6: 161134077-161134077 on Build GRCh38
rs994654885 Chr.6: 161133886-161133886 on Build GRCh38
rs1056819705 Chr.6: 161134159-161134159 on Build GRCh38
rs539581841 Chr.6: 161133915-161133915 on Build GRCh38
rs41268549 Chr.6: 161134194-161134194 on Build GRCh38
rs779765984 Chr.6: 161133947-161133947 on Build GRCh38
rs1007625303 Chr.6: 161134060-161134060 on Build GRCh38
rs1048899526 Chr.6: 161134075-161134075 on Build GRCh38
rs915001947 Chr.6: 161134168-161134168 on Build GRCh38

View Full Product Details