Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 161146436-161146932 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AGPAT4
Entrez Gene ID
56895
Gene Name
1-acylglycerol-3-phosphate O-acyltransferase 4
Gene Aliases
1-AGPAT4, LPAAT-delta, dJ473J16.2
UniGene
Hs.353175
Gene Chromosome Location
Chr.6: 161129980-161274108 on Build GRCh38
Transcripts
NM_020133.2 XM_005267053.3 XM_006715514.3 XM_017011059.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6596389 c.789C>T p.I263I Chr.6: 161146578-161146578 on Build GRCh38
6172953 c.791C>G p.P264R Chr.6: 161146576-161146576 on Build GRCh38
1176503 c.784G>T p.D262Y Chr.6: 161146583-161146583 on Build GRCh38
7278208 c.812C>T p.S271L Chr.6: 161146555-161146555 on Build GRCh38
6596394 c.768G>T p.R256S Chr.6: 161146599-161146599 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs2177588 Chr.6: 161146670-161146670 on Build GRCh38
rs781498209 Chr.6: 161146510-161146510 on Build GRCh38
rs751101624 Chr.6: 161146630-161146630 on Build GRCh38
rs749830897 Chr.6: 161146540-161146540 on Build GRCh38
rs867021511 Chr.6: 161146676-161146676 on Build GRCh38
rs199621690 Chr.6: 161146612-161146612 on Build GRCh38
rs972052402 Chr.6: 161146880-161146880 on Build GRCh38
rs757920893 Chr.6: 161146492-161146492 on Build GRCh38
rs776596724 Chr.6: 161146475-161146475 on Build GRCh38
rs757774501 Chr.6: 161146565-161146565 on Build GRCh38
rs530701071 Chr.6: 161146801-161146801 on Build GRCh38
rs761389671 Chr.6: 161146634-161146634 on Build GRCh38
rs142563808 Chr.6: 161146882-161146882 on Build GRCh38
rs772384211 Chr.6: 161146667-161146667 on Build GRCh38
rs909178967 Chr.6: 161146658-161146658 on Build GRCh38
rs779725287 Chr.6: 161146668-161146668 on Build GRCh38
rs905469082 Chr.6: 161146680-161146680 on Build GRCh38
rs756197557 Chr.6: 161146471-161146471 on Build GRCh38
rs774591636 Chr.6: 161146551-161146551 on Build GRCh38
rs776508549 Chr.6: 161146562-161146562 on Build GRCh38
rs1039738610 Chr.6: 161146899-161146899 on Build GRCh38
rs1030471751 Chr.6: 161146655-161146655 on Build GRCh38
rs200571825 Chr.6: 161146474-161146474 on Build GRCh38
rs777666817 Chr.6: 161146486-161146486 on Build GRCh38
rs996719458 Chr.6: 161146774-161146774 on Build GRCh38
rs112074287 Chr.6: 161146873-161146873 on Build GRCh38
rs963441288 Chr.6: 161146702-161146702 on Build GRCh38
rs185543139 Chr.6: 161146800-161146800 on Build GRCh38
rs746238707 Chr.6: 161146516-161146516 on Build GRCh38
rs762847778 Chr.6: 161146618-161146618 on Build GRCh38
rs769542610 Chr.6: 161146590-161146590 on Build GRCh38
rs374557581 Chr.6: 161146521-161146521 on Build GRCh38
rs765232628 Chr.6: 161146487-161146487 on Build GRCh38
rs900894569 Chr.6: 161146539-161146539 on Build GRCh38
rs545337522 Chr.6: 161146701-161146701 on Build GRCh38
rs373907543 Chr.6: 161146661-161146661 on Build GRCh38
rs754003591 Chr.6: 161146554-161146554 on Build GRCh38
rs752632755 Chr.6: 161146491-161146491 on Build GRCh38
rs41268553 Chr.6: 161146663-161146663 on Build GRCh38
rs574877874 Chr.6: 161146603-161146603 on Build GRCh38
rs1045361714 Chr.6: 161146458-161146458 on Build GRCh38
rs759477077 Chr.6: 161146485-161146485 on Build GRCh38
rs936895656 Chr.6: 161146693-161146693 on Build GRCh38
rs900922670 Chr.6: 161146751-161146751 on Build GRCh38
rs114493502 Chr.6: 161146493-161146493 on Build GRCh38
rs767173324 Chr.6: 161146643-161146643 on Build GRCh38
rs16892215 Chr.6: 161146569-161146569 on Build GRCh38
rs775961175 Chr.6: 161146761-161146761 on Build GRCh38
rs750137210 Chr.6: 161146644-161146644 on Build GRCh38
rs112206510 Chr.6: 161146817-161146817 on Build GRCh38
rs918037209 Chr.6: 161146901-161146901 on Build GRCh38
rs772424707 Chr.6: 161146559-161146559 on Build GRCh38
rs779574861 Chr.6: 161146595-161146595 on Build GRCh38
rs372706289 Chr.6: 161146478-161146478 on Build GRCh38
rs190647520 Chr.6: 161146499-161146499 on Build GRCh38
rs769708215 Chr.6: 161146863-161146863 on Build GRCh38
rs915113379 Chr.6: 161146858-161146858 on Build GRCh38
rs984414652 Chr.6: 161146852-161146852 on Build GRCh38
rs367746530 Chr.6: 161146461-161146461 on Build GRCh38
rs916670636 Chr.6: 161146738-161146738 on Build GRCh38
rs538078412 Chr.6: 161146544-161146544 on Build GRCh38
rs1009274213 Chr.6: 161146722-161146722 on Build GRCh38
rs745800526 Chr.6: 161146592-161146592 on Build GRCh38
rs182686983 Chr.6: 161146677-161146677 on Build GRCh38
rs935146823 Chr.6: 161146804-161146804 on Build GRCh38
rs768374661 Chr.6: 161146732-161146732 on Build GRCh38
rs763611006 Chr.6: 161146628-161146628 on Build GRCh38
rs892097991 Chr.6: 161146706-161146706 on Build GRCh38

View Full Product Details