Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 161149096-161149529 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AGPAT4
Entrez Gene ID
56895
Gene Name
1-acylglycerol-3-phosphate O-acyltransferase 4
Gene Aliases
1-AGPAT4, LPAAT-delta, dJ473J16.2
UniGene
Hs.353175
Gene Chromosome Location
Chr.6: 161129980-161274108 on Build GRCh38
Transcripts
NM_020133.2 XM_005267053.3 XM_006715514.3 XM_017011059.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1075788 c.764T>C p.V255A Chr.6: 161149190-161149190 on Build GRCh38
5659272 c.727G>A p.V243I Chr.6: 161149227-161149227 on Build GRCh38
6596385 c.720G>A p.L240L Chr.6: 161149234-161149234 on Build GRCh38
3024301 c.741G>T p.K247N Chr.6: 161149213-161149213 on Build GRCh38
6105666 c.761A>T p.Y254F Chr.6: 161149193-161149193 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs753322144 Chr.6: 161149276-161149276 on Build GRCh38
rs977270739 Chr.6: 161149158-161149158 on Build GRCh38
rs779254972 Chr.6: 161149309-161149318 on Build GRCh38
rs371787889 Chr.6: 161149216-161149216 on Build GRCh38
rs150925621 Chr.6: 161149278-161149278 on Build GRCh38
rs376393857 Chr.6: 161149321-161149321 on Build GRCh38
rs753981426 Chr.6: 161149154-161149154 on Build GRCh38
rs754317403 Chr.6: 161149156-161149156 on Build GRCh38
rs768483790 Chr.6: 161149211-161149211 on Build GRCh38
rs773794283 Chr.6: 161149212-161149212 on Build GRCh38
rs758519495 Chr.6: 161149305-161149305 on Build GRCh38
rs1021777488 Chr.6: 161149192-161149192 on Build GRCh38
rs967223381 Chr.6: 161149464-161149464 on Build GRCh38
rs374767306 Chr.6: 161149317-161149317 on Build GRCh38
rs114126692 Chr.6: 161149219-161149219 on Build GRCh38
rs905752655 Chr.6: 161149137-161149137 on Build GRCh38
rs780326756 Chr.6: 161149155-161149155 on Build GRCh38
rs1044584454 Chr.6: 161149418-161149418 on Build GRCh38
rs927222690 Chr.6: 161149455-161149456 on Build GRCh38
rs559784830 Chr.6: 161149380-161149380 on Build GRCh38
rs765583443 Chr.6: 161149132-161149132 on Build GRCh38
rs752150830 Chr.6: 161149180-161149180 on Build GRCh38
rs775169927 Chr.6: 161149204-161149204 on Build GRCh38
rs779546597 Chr.6: 161149243-161149243 on Build GRCh38
rs199680926 Chr.6: 161149179-161149179 on Build GRCh38
rs778047285 Chr.6: 161149312-161149312 on Build GRCh38
rs755052475 Chr.6: 161149301-161149305 on Build GRCh38
rs1021418698 Chr.6: 161149459-161149459 on Build GRCh38
rs749058027 Chr.6: 161149326-161149326 on Build GRCh38
rs748449288 Chr.6: 161149311-161149311 on Build GRCh38
rs954243165 Chr.6: 161149228-161149228 on Build GRCh38
rs919977499 Chr.6: 161149369-161149369 on Build GRCh38
rs766722668 Chr.6: 161149235-161149235 on Build GRCh38
rs762908731 Chr.6: 161149206-161149206 on Build GRCh38
rs1056977345 Chr.6: 161149496-161149496 on Build GRCh38
rs767825371 Chr.6: 161149348-161149348 on Build GRCh38
rs139732317 Chr.6: 161149236-161149236 on Build GRCh38
rs142714214 Chr.6: 161149262-161149262 on Build GRCh38
rs377012817 Chr.6: 161149133-161149133 on Build GRCh38
rs760297709 Chr.6: 161149224-161149224 on Build GRCh38
rs771063067 Chr.6: 161149318-161149318 on Build GRCh38
rs369950889 Chr.6: 161149327-161149327 on Build GRCh38
rs576502116 Chr.6: 161149447-161149447 on Build GRCh38
rs779065236 Chr.6: 161149165-161149165 on Build GRCh38
rs907439333 Chr.6: 161149319-161149319 on Build GRCh38
rs368251132 Chr.6: 161149149-161149149 on Build GRCh38
rs753091547 Chr.6: 161149470-161149470 on Build GRCh38
rs372076048 Chr.6: 161149355-161149355 on Build GRCh38
rs370825906 Chr.6: 161149163-161149163 on Build GRCh38
rs769609356 Chr.6: 161149197-161149197 on Build GRCh38
rs749013 Chr.6: 161149171-161149171 on Build GRCh38
rs755452224 Chr.6: 161149238-161149238 on Build GRCh38
rs767083784 Chr.6: 161149218-161149218 on Build GRCh38
rs1011249969 Chr.6: 161149403-161149403 on Build GRCh38

View Full Product Details