Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 169703117-169703620 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C6orf120
Entrez Gene ID
387263
Gene Name
chromosome 6 open reading frame 120
Gene Aliases
-
UniGene
Hs.591375
Gene Chromosome Location
Chr.6: 169702112-169706358 on Build GRCh38
Transcripts
NM_001029863.2 NM_001317342.1

SNP Detail

SNP ID Chromosome Location
rs1031376348 Chr.6: 169703534-169703534 on Build GRCh38
rs192009047 Chr.6: 169703576-169703576 on Build GRCh38
rs1038527696 Chr.6: 169703401-169703401 on Build GRCh38
rs1033390290 Chr.6: 169703402-169703402 on Build GRCh38
rs956707508 Chr.6: 169703275-169703275 on Build GRCh38
rs774099363 Chr.6: 169703422-169703422 on Build GRCh38
rs992235249 Chr.6: 169703194-169703194 on Build GRCh38
rs908711577 Chr.6: 169703358-169703358 on Build GRCh38
rs140885693 Chr.6: 169703306-169703306 on Build GRCh38
rs1038201602 Chr.6: 169703258-169703258 on Build GRCh38
rs772529928 Chr.6: 169703338-169703338 on Build GRCh38
rs889203953 Chr.6: 169703317-169703317 on Build GRCh38
rs974369828 Chr.6: 169703461-169703461 on Build GRCh38
rs541276865 Chr.6: 169703301-169703301 on Build GRCh38
rs975074468 Chr.6: 169703337-169703337 on Build GRCh38
rs143888444 Chr.6: 169703179-169703179 on Build GRCh38
rs941563740 Chr.6: 169703389-169703389 on Build GRCh38
rs187260913 Chr.6: 169703300-169703300 on Build GRCh38
rs892376687 Chr.6: 169703539-169703539 on Build GRCh38
rs111452450 Chr.6: 169703546-169703546 on Build GRCh38
rs892490917 Chr.6: 169703581-169703581 on Build GRCh38
rs530311501 Chr.6: 169703343-169703343 on Build GRCh38
rs1009582696 Chr.6: 169703595-169703595 on Build GRCh38
rs908418621 Chr.6: 169703231-169703231 on Build GRCh38
rs1012987499 Chr.6: 169703477-169703477 on Build GRCh38
rs200015159 Chr.6: 169703414-169703414 on Build GRCh38
rs892716994 Chr.6: 169703352-169703352 on Build GRCh38
rs572231838 Chr.6: 169703185-169703185 on Build GRCh38
rs963754375 Chr.6: 169703331-169703331 on Build GRCh38
rs980953596 Chr.6: 169703397-169703397 on Build GRCh38
rs5881838 Chr.6: 169703412-169703413 on Build GRCh38
rs397887115 Chr.6: 169703413-169703414 on Build GRCh38
rs539747397 Chr.6: 169703285-169703285 on Build GRCh38
rs545359793 Chr.6: 169703239-169703239 on Build GRCh38
rs564050334 Chr.6: 169703249-169703249 on Build GRCh38
rs1022447050 Chr.6: 169703387-169703387 on Build GRCh38
rs374173056 Chr.6: 169703456-169703456 on Build GRCh38
rs767732627 Chr.6: 169703379-169703379 on Build GRCh38
rs970651153 Chr.6: 169703202-169703202 on Build GRCh38
rs149834038 Chr.6: 169703540-169703540 on Build GRCh38
rs183783243 Chr.6: 169703180-169703180 on Build GRCh38
rs1030842593 Chr.6: 169703267-169703267 on Build GRCh38
rs1003432244 Chr.6: 169703257-169703257 on Build GRCh38
rs756235912 Chr.6: 169703498-169703498 on Build GRCh38
rs757578985 Chr.6: 169703373-169703373 on Build GRCh38
rs377582741 Chr.6: 169703525-169703525 on Build GRCh38
rs147277916 Chr.6: 169703201-169703201 on Build GRCh38
rs908242997 Chr.6: 169703441-169703441 on Build GRCh38
rs750710087 Chr.6: 169703440-169703440 on Build GRCh38
rs939932870 Chr.6: 169703455-169703455 on Build GRCh38
rs1038820093 Chr.6: 169703182-169703182 on Build GRCh38
rs919863486 Chr.6: 169703484-169703484 on Build GRCh38
rs1012956281 Chr.6: 169703190-169703190 on Build GRCh38
rs548925473 Chr.6: 169703368-169703368 on Build GRCh38
rs934255771 Chr.6: 169703340-169703340 on Build GRCh38
rs898365230 Chr.6: 169703265-169703265 on Build GRCh38
rs907168369 Chr.6: 169703511-169703511 on Build GRCh38
rs968178073 Chr.6: 169703394-169703394 on Build GRCh38

View Full Product Details