Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 169704576-169705093 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C6orf120
Entrez Gene ID
387263
Gene Name
chromosome 6 open reading frame 120
Gene Aliases
-
UniGene
Hs.591375
Gene Chromosome Location
Chr.6: 169702112-169706358 on Build GRCh38
Transcripts
NM_001029863.2 NM_001317342.1 NM_018288.3 NM_133325.2 XM_017010998.1

Gene Symbol
PHF10
Entrez Gene ID
55274
Gene Name
PHD finger protein 10
Gene Aliases
BAF45A, XAP135
UniGene
Hs.435933
Gene Chromosome Location
Chr.6: 169703902-169724049 on Build GRCh38
Transcripts
NM_001029863.2 NM_001317342.1 NM_018288.3 NM_133325.2 XM_017010998.1

SNP Detail

SNP ID Chromosome Location
rs1032395442 Chr.6: 169704759-169704759 on Build GRCh38
rs375523720 Chr.6: 169705025-169705026 on Build GRCh38
rs555014531 Chr.6: 169704967-169704967 on Build GRCh38
rs919723992 Chr.6: 169704951-169704951 on Build GRCh38
rs912508449 Chr.6: 169704910-169704910 on Build GRCh38
rs917949243 Chr.6: 169704775-169704775 on Build GRCh38
rs991228554 Chr.6: 169704773-169704773 on Build GRCh38
rs968849900 Chr.6: 169704863-169704863 on Build GRCh38
rs182665528 Chr.6: 169704907-169704907 on Build GRCh38
rs1012362379 Chr.6: 169704998-169704998 on Build GRCh38
rs41265395 Chr.6: 169705014-169705014 on Build GRCh38
rs762279643 Chr.6: 169705045-169705045 on Build GRCh38
rs1048551648 Chr.6: 169704826-169704826 on Build GRCh38
rs931056892 Chr.6: 169704958-169704958 on Build GRCh38
rs952998979 Chr.6: 169705046-169705046 on Build GRCh38
rs566612802 Chr.6: 169704942-169704942 on Build GRCh38
rs757052767 Chr.6: 169704786-169704786 on Build GRCh38
rs771842225 Chr.6: 169705061-169705061 on Build GRCh38
rs939913428 Chr.6: 169704928-169704928 on Build GRCh38
rs561851934 Chr.6: 169704702-169704702 on Build GRCh38
rs539231382 Chr.6: 169704881-169704881 on Build GRCh38
rs775133903 Chr.6: 169705003-169705003 on Build GRCh38
rs867743214 Chr.6: 169704831-169704831 on Build GRCh38
rs781018786 Chr.6: 169704800-169704800 on Build GRCh38
rs1010434466 Chr.6: 169704808-169704808 on Build GRCh38
rs931025791 Chr.6: 169704714-169704714 on Build GRCh38
rs1010882359 Chr.6: 169705058-169705058 on Build GRCh38
rs541225196 Chr.6: 169704809-169704809 on Build GRCh38
rs546650878 Chr.6: 169704660-169704660 on Build GRCh38
rs535931596 Chr.6: 169704775-169704778 on Build GRCh38
rs1021808089 Chr.6: 169705072-169705072 on Build GRCh38
rs564610768 Chr.6: 169704615-169704615 on Build GRCh38
rs889523693 Chr.6: 169704854-169704854 on Build GRCh38
rs949656057 Chr.6: 169704779-169704779 on Build GRCh38
rs749558975 Chr.6: 169704662-169704662 on Build GRCh38
rs918859348 Chr.6: 169704788-169704788 on Build GRCh38
rs954747717 Chr.6: 169704888-169704888 on Build GRCh38
rs901082381 Chr.6: 169704695-169704695 on Build GRCh38
rs113275655 Chr.6: 169705042-169705042 on Build GRCh38
rs768085007 Chr.6: 169705021-169705021 on Build GRCh38
rs978596237 Chr.6: 169704758-169704758 on Build GRCh38
rs568761769 Chr.6: 169704879-169704879 on Build GRCh38
rs879064523 Chr.6: 169704992-169704992 on Build GRCh38
rs146706178 Chr.6: 169704617-169704617 on Build GRCh38
rs529309613 Chr.6: 169704748-169704748 on Build GRCh38
rs974770444 Chr.6: 169704871-169704871 on Build GRCh38
rs959428203 Chr.6: 169704991-169704991 on Build GRCh38
rs140331004 Chr.6: 169704961-169704961 on Build GRCh38
rs371851951 Chr.6: 169704803-169704803 on Build GRCh38
rs375749587 Chr.6: 169704946-169704946 on Build GRCh38
rs1057190172 Chr.6: 169705035-169705035 on Build GRCh38
rs931440726 Chr.6: 169704802-169704802 on Build GRCh38
rs1010174959 Chr.6: 169704709-169704712 on Build GRCh38
rs921914497 Chr.6: 169704609-169704609 on Build GRCh38
rs1034225369 Chr.6: 169704982-169704982 on Build GRCh38
rs905491494 Chr.6: 169704746-169704746 on Build GRCh38
rs1044095377 Chr.6: 169704717-169704717 on Build GRCh38
rs1037257751 Chr.6: 169704672-169704672 on Build GRCh38
rs745511669 Chr.6: 169704804-169704804 on Build GRCh38
rs900863037 Chr.6: 169704933-169704933 on Build GRCh38
rs372449544 Chr.6: 169705028-169705028 on Build GRCh38
rs563688315 Chr.6: 169705024-169705024 on Build GRCh38
rs865942109 Chr.6: 169704754-169704754 on Build GRCh38
rs959480071 Chr.6: 169704772-169704772 on Build GRCh38
rs912467032 Chr.6: 169704641-169704641 on Build GRCh38
rs774895100 Chr.6: 169705027-169705027 on Build GRCh38
rs1019310330 Chr.6: 169704682-169704682 on Build GRCh38
rs1000023966 Chr.6: 169704969-169704969 on Build GRCh38

View Full Product Details