Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 169753773-169754270 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ERMARD
Entrez Gene ID
55780
Gene Name
ER membrane associated RNA degradation
Gene Aliases
C6orf70, PVNH6, dJ266L20.3
UniGene
Hs.47546
Gene Chromosome Location
Chr.6: 169751622-169781584 on Build GRCh38
Transcripts
NM_001278531.1 NM_001278532.1 NM_001278533.1 NM_018341.2 XM_011535938.2 XM_011535939.2 XM_011535940.2 XM_017011027.1 XM_017011028.1 XM_017011029.1 XM_017011030.1 XM_017011031.1 XM_017011032.1 XM_017011033.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7317276 c.127G>A p.E43K Chr.6: 169753984-169753984 on Build GRCh38
7193562 c.30C>A p.T10T Chr.6: 169753887-169753887 on Build GRCh38
4667935 c.90A>T p.E30D Chr.6: 169753947-169753947 on Build GRCh38
159651 c.84C>G p.L28L Chr.6: 169753941-169753941 on Build GRCh38
7193561 c.30C>A p.T10T Chr.6: 169753887-169753887 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs187725532 Chr.6: 169754028-169754028 on Build GRCh38
rs757396095 Chr.6: 169753916-169753916 on Build GRCh38
rs376057280 Chr.6: 169754072-169754072 on Build GRCh38
rs760228453 Chr.6: 169753971-169753971 on Build GRCh38
rs771879394 Chr.6: 169754047-169754047 on Build GRCh38
rs929964008 Chr.6: 169754075-169754075 on Build GRCh38
rs772495258 Chr.6: 169754132-169754132 on Build GRCh38
rs767434408 Chr.6: 169753838-169753838 on Build GRCh38
rs756298238 Chr.6: 169754031-169754031 on Build GRCh38
rs746340140 Chr.6: 169754017-169754017 on Build GRCh38
rs753468195 Chr.6: 169753884-169753884 on Build GRCh38
rs763273034 Chr.6: 169754071-169754071 on Build GRCh38
rs533667178 Chr.6: 169753946-169753946 on Build GRCh38
rs758842196 Chr.6: 169753910-169753910 on Build GRCh38
rs140054205 Chr.6: 169753890-169753890 on Build GRCh38
rs752224928 Chr.6: 169753837-169753844 on Build GRCh38
rs774814332 Chr.6: 169754074-169754074 on Build GRCh38
rs555474634 Chr.6: 169753973-169753973 on Build GRCh38
rs750475288 Chr.6: 169754030-169754030 on Build GRCh38
rs369223747 Chr.6: 169754066-169754066 on Build GRCh38
rs1029403308 Chr.6: 169754236-169754236 on Build GRCh38
rs564343371 Chr.6: 169754185-169754185 on Build GRCh38
rs747632937 Chr.6: 169754105-169754105 on Build GRCh38
rs778273730 Chr.6: 169753914-169753914 on Build GRCh38
rs921067108 Chr.6: 169753992-169753992 on Build GRCh38
rs913813619 Chr.6: 169753840-169753840 on Build GRCh38
rs753006522 Chr.6: 169753990-169753990 on Build GRCh38
rs770097620 Chr.6: 169753925-169753925 on Build GRCh38
rs61736820 Chr.6: 169753866-169753866 on Build GRCh38
rs763371570 Chr.6: 169753993-169753993 on Build GRCh38
rs751655186 Chr.6: 169753915-169753915 on Build GRCh38
rs575544012 Chr.6: 169754128-169754128 on Build GRCh38
rs746982395 Chr.6: 169754056-169754056 on Build GRCh38
rs768502290 Chr.6: 169753841-169753845 on Build GRCh38
rs192530286 Chr.6: 169754068-169754068 on Build GRCh38
rs777611820 Chr.6: 169754054-169754054 on Build GRCh38
rs375019990 Chr.6: 169754063-169754063 on Build GRCh38
rs572770346 Chr.6: 169753917-169753917 on Build GRCh38
rs751944841 Chr.6: 169753846-169753873 on Build GRCh38
rs1023212001 Chr.6: 169754153-169754153 on Build GRCh38
rs776080895 Chr.6: 169753977-169753977 on Build GRCh38
rs780323173 Chr.6: 169754032-169754032 on Build GRCh38
rs759210576 Chr.6: 169753986-169753986 on Build GRCh38
rs1050824807 Chr.6: 169754022-169754022 on Build GRCh38
rs770807460 Chr.6: 169754057-169754057 on Build GRCh38
rs888694495 Chr.6: 169754125-169754125 on Build GRCh38
rs770450852 Chr.6: 169754025-169754026 on Build GRCh38
rs998786016 Chr.6: 169754221-169754221 on Build GRCh38
rs183771901 Chr.6: 169754181-169754181 on Build GRCh38
rs757451102 Chr.6: 169754027-169754027 on Build GRCh38
rs150302870 Chr.6: 169753966-169753966 on Build GRCh38
rs369674761 Chr.6: 169754000-169754000 on Build GRCh38
rs371895613 Chr.6: 169754037-169754037 on Build GRCh38
rs759934226 Chr.6: 169753889-169753889 on Build GRCh38
rs778743297 Chr.6: 169753843-169753845 on Build GRCh38
rs780228998 Chr.6: 169754048-169754049 on Build GRCh38
rs925852536 Chr.6: 169754038-169754038 on Build GRCh38
rs760177283 Chr.6: 169753876-169753876 on Build GRCh38
rs781704200 Chr.6: 169753941-169753942 on Build GRCh38
rs752086371 Chr.6: 169754012-169754012 on Build GRCh38
rs764884752 Chr.6: 169753987-169753987 on Build GRCh38
rs1005718358 Chr.6: 169754162-169754162 on Build GRCh38
rs77846484 Chr.6: 169754123-169754123 on Build GRCh38
rs979250854 Chr.6: 169753942-169753942 on Build GRCh38
rs113015559 Chr.6: 169753885-169753885 on Build GRCh38
rs747912142 Chr.6: 169753943-169753943 on Build GRCh38
rs777553387 Chr.6: 169754003-169754003 on Build GRCh38
rs778589676 Chr.6: 169753941-169753941 on Build GRCh38
rs747627800 Chr.6: 169753890-169753891 on Build GRCh38

View Full Product Details