Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 169756236-169756747 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ERMARD
Entrez Gene ID
55780
Gene Name
ER membrane associated RNA degradation
Gene Aliases
C6orf70, PVNH6, dJ266L20.3
UniGene
Hs.47546
Gene Chromosome Location
Chr.6: 169751622-169781584 on Build GRCh38
Transcripts
NM_001278531.1 NM_001278532.1 NM_001278533.1 NM_018341.2 XM_011535938.2 XM_011535939.2 XM_011535940.2 XM_017011027.1 XM_017011028.1 XM_017011029.1 XM_017011030.1 XM_017011031.1 XM_017011032.1 XM_017011033.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3622987 c.322C>T p.P108S Chr.6: 169756344-169756344 on Build GRCh38
6213039 c.445C>T p.P149S Chr.6: 169756746-169756746 on Build GRCh38
5421825 c.418-8A>C p.? Chr.6: 169756711-169756711 on Build GRCh38
5896805 c.446C>T p.P149L Chr.6: 169756747-169756747 on Build GRCh38
1076176 c.393G>A p.S131S Chr.6: 169756415-169756415 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs905843345 Chr.6: 169756628-169756628 on Build GRCh38
rs556589978 Chr.6: 169756490-169756490 on Build GRCh38
rs764239580 Chr.6: 169756432-169756432 on Build GRCh38
rs1052307804 Chr.6: 169756561-169756561 on Build GRCh38
rs184359209 Chr.6: 169756428-169756428 on Build GRCh38
rs769832428 Chr.6: 169756414-169756414 on Build GRCh38
rs987381446 Chr.6: 169756529-169756529 on Build GRCh38
rs937217888 Chr.6: 169756519-169756519 on Build GRCh38
rs771802042 Chr.6: 169756378-169756378 on Build GRCh38
rs953176404 Chr.6: 169756435-169756435 on Build GRCh38
rs560874306 Chr.6: 169756691-169756691 on Build GRCh38
rs1041891817 Chr.6: 169756555-169756555 on Build GRCh38
rs377727194 Chr.6: 169756415-169756415 on Build GRCh38
rs777405432 Chr.6: 169756580-169756580 on Build GRCh38
rs764891278 Chr.6: 169756402-169756402 on Build GRCh38
rs556131065 Chr.6: 169756369-169756369 on Build GRCh38
rs966222161 Chr.6: 169756303-169756303 on Build GRCh38
rs754930413 Chr.6: 169756474-169756474 on Build GRCh38
rs189135542 Chr.6: 169756622-169756622 on Build GRCh38
rs1036373761 Chr.6: 169756400-169756400 on Build GRCh38
rs935169474 Chr.6: 169756262-169756262 on Build GRCh38
rs771533881 Chr.6: 169756401-169756401 on Build GRCh38
rs753448490 Chr.6: 169756370-169756370 on Build GRCh38
rs1049057980 Chr.6: 169756471-169756471 on Build GRCh38
rs558340050 Chr.6: 169756267-169756267 on Build GRCh38
rs779240200 Chr.6: 169756538-169756538 on Build GRCh38
rs972579678 Chr.6: 169756341-169756341 on Build GRCh38
rs374781539 Chr.6: 169756386-169756386 on Build GRCh38
rs771175836 Chr.6: 169756678-169756678 on Build GRCh38
rs369604833 Chr.6: 169756515-169756515 on Build GRCh38
rs760377140 Chr.6: 169756460-169756460 on Build GRCh38
rs1032613957 Chr.6: 169756647-169756647 on Build GRCh38
rs775557378 Chr.6: 169756709-169756709 on Build GRCh38
rs374284782 Chr.6: 169756300-169756300 on Build GRCh38
rs566903483 Chr.6: 169756292-169756292 on Build GRCh38
rs751016488 Chr.6: 169756585-169756585 on Build GRCh38
rs781751836 Chr.6: 169756607-169756607 on Build GRCh38
rs9478102 Chr.6: 169756587-169756587 on Build GRCh38
rs141826774 Chr.6: 169756407-169756407 on Build GRCh38
rs749295823 Chr.6: 169756715-169756715 on Build GRCh38
rs756087967 Chr.6: 169756368-169756368 on Build GRCh38
rs895662206 Chr.6: 169756542-169756542 on Build GRCh38
rs973561644 Chr.6: 169756425-169756425 on Build GRCh38
rs777287505 Chr.6: 169756673-169756673 on Build GRCh38
rs761635051 Chr.6: 169756312-169756312 on Build GRCh38
rs781742123 Chr.6: 169756681-169756681 on Build GRCh38
rs545128997 Chr.6: 169756568-169756568 on Build GRCh38
rs752893387 Chr.6: 169756304-169756304 on Build GRCh38
rs758130186 Chr.6: 169756669-169756669 on Build GRCh38
rs371095977 Chr.6: 169756459-169756459 on Build GRCh38
rs17860634 Chr.6: 169756375-169756375 on Build GRCh38
rs767328692 Chr.6: 169756319-169756319 on Build GRCh38
rs142350636 Chr.6: 169756675-169756675 on Build GRCh38
rs757846757 Chr.6: 169756491-169756491 on Build GRCh38
rs375610749 Chr.6: 169756463-169756463 on Build GRCh38
rs778214427 Chr.6: 169756384-169756384 on Build GRCh38
rs1001768121 Chr.6: 169756394-169756394 on Build GRCh38
rs750132665 Chr.6: 169756361-169756361 on Build GRCh38
rs754516803 Chr.6: 169756373-169756373 on Build GRCh38
rs1014085015 Chr.6: 169756549-169756549 on Build GRCh38
rs921578410 Chr.6: 169756426-169756426 on Build GRCh38
rs778752194 Chr.6: 169756374-169756374 on Build GRCh38
rs759419113 Chr.6: 169756441-169756441 on Build GRCh38
rs770232885 Chr.6: 169756698-169756698 on Build GRCh38
rs953641845 Chr.6: 169756652-169756652 on Build GRCh38

View Full Product Details