Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 169756723-169757230 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ERMARD
Entrez Gene ID
55780
Gene Name
ER membrane associated RNA degradation
Gene Aliases
C6orf70, PVNH6, dJ266L20.3
UniGene
Hs.47546
Gene Chromosome Location
Chr.6: 169751622-169781584 on Build GRCh38
Transcripts
NM_001278531.1 NM_001278532.1 NM_001278533.1 NM_018341.2 XM_011535938.2 XM_011535939.2 XM_011535940.2 XM_017011027.1 XM_017011028.1 XM_017011029.1 XM_017011030.1 XM_017011031.1 XM_017011032.1 XM_017011033.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4882426 c.470C>T p.S157L Chr.6: 169756771-169756771 on Build GRCh38
4882427 c.470C>T p.S157L Chr.6: 169756771-169756771 on Build GRCh38
6213039 c.445C>T p.P149S Chr.6: 169756746-169756746 on Build GRCh38
5896805 c.446C>T p.P149L Chr.6: 169756747-169756747 on Build GRCh38
3777323 c.466C>T p.L156F Chr.6: 169756767-169756767 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs766122955 Chr.6: 169756746-169756746 on Build GRCh38
rs895822417 Chr.6: 169757011-169757011 on Build GRCh38
rs150622904 Chr.6: 169756770-169756770 on Build GRCh38
rs186041744 Chr.6: 169757176-169757176 on Build GRCh38
rs774539824 Chr.6: 169757032-169757032 on Build GRCh38
rs780428551 Chr.6: 169756934-169756934 on Build GRCh38
rs565753902 Chr.6: 169756887-169756887 on Build GRCh38
rs757987474 Chr.6: 169756889-169756889 on Build GRCh38
rs558826289 Chr.6: 169757082-169757082 on Build GRCh38
rs994564718 Chr.6: 169756868-169756868 on Build GRCh38
rs749421766 Chr.6: 169756837-169756837 on Build GRCh38
rs55829984 Chr.6: 169757102-169757102 on Build GRCh38
rs534157072 Chr.6: 169756918-169756918 on Build GRCh38
rs1057098435 Chr.6: 169757004-169757004 on Build GRCh38
rs377591444 Chr.6: 169756847-169756847 on Build GRCh38
rs763855176 Chr.6: 169756785-169756785 on Build GRCh38
rs984897002 Chr.6: 169757116-169757116 on Build GRCh38
rs746169572 Chr.6: 169756807-169756807 on Build GRCh38
rs2011670 Chr.6: 169757192-169757192 on Build GRCh38
rs1036583887 Chr.6: 169757061-169757061 on Build GRCh38
rs759560711 Chr.6: 169756755-169756755 on Build GRCh38
rs756615645 Chr.6: 169756827-169756827 on Build GRCh38
rs1054122385 Chr.6: 169756976-169756976 on Build GRCh38
rs756893835 Chr.6: 169756792-169756792 on Build GRCh38
rs549596888 Chr.6: 169756947-169756947 on Build GRCh38
rs749591957 Chr.6: 169756935-169756935 on Build GRCh38
rs538501596 Chr.6: 169757056-169757056 on Build GRCh38
rs994075557 Chr.6: 169757081-169757081 on Build GRCh38
rs577933690 Chr.6: 169757096-169757096 on Build GRCh38
rs564497807 Chr.6: 169756846-169756846 on Build GRCh38
rs568001806 Chr.6: 169756969-169756969 on Build GRCh38
rs781429204 Chr.6: 169756806-169756806 on Build GRCh38
rs949972637 Chr.6: 169757201-169757201 on Build GRCh38
rs746262224 Chr.6: 169756761-169756761 on Build GRCh38
rs13209742 Chr.6: 169756883-169756883 on Build GRCh38
rs1029178149 Chr.6: 169756882-169756882 on Build GRCh38
rs369883629 Chr.6: 169757048-169757049 on Build GRCh38
rs776438180 Chr.6: 169756747-169756747 on Build GRCh38
rs377504977 Chr.6: 169756860-169756860 on Build GRCh38
rs952121726 Chr.6: 169757111-169757111 on Build GRCh38
rs780532189 Chr.6: 169756832-169756832 on Build GRCh38
rs182023999 Chr.6: 169757062-169757062 on Build GRCh38
rs768749493 Chr.6: 169756945-169756945 on Build GRCh38
rs11968997 Chr.6: 169757124-169757124 on Build GRCh38
rs764141360 Chr.6: 169756994-169756996 on Build GRCh38
rs991854301 Chr.6: 169756912-169756912 on Build GRCh38
rs902061681 Chr.6: 169757065-169757065 on Build GRCh38
rs531497550 Chr.6: 169756775-169756775 on Build GRCh38
rs139763023 Chr.6: 169756790-169756790 on Build GRCh38
rs937352199 Chr.6: 169756990-169756990 on Build GRCh38
rs888807425 Chr.6: 169756980-169756980 on Build GRCh38
rs752323554 Chr.6: 169756766-169756766 on Build GRCh38
rs374397711 Chr.6: 169756848-169756848 on Build GRCh38
rs988033970 Chr.6: 169757149-169757149 on Build GRCh38
rs917155533 Chr.6: 169757195-169757195 on Build GRCh38
rs796734719 Chr.6: 169757020-169757020 on Build GRCh38

View Full Product Details