Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 22942290-22942803 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM126A
Entrez Gene ID
84668
Gene Name
family with sequence similarity 126 member A
Gene Aliases
DRCTNNB1A, HCC, HLD5, HYCC1
UniGene
Hs.85603
Gene Chromosome Location
Chr.7: 22895848-23014195 on Build GRCh38
Transcripts
NM_032581.3 XM_005249894.3 XM_005249895.2 XM_011515589.2 XM_011515590.1

SNP Detail

SNP ID Chromosome Location
rs191973703 Chr.7: 22942483-22942483 on Build GRCh38
rs1009605953 Chr.7: 22942533-22942533 on Build GRCh38
rs924990000 Chr.7: 22942572-22942572 on Build GRCh38
rs1053725886 Chr.7: 22942506-22942506 on Build GRCh38
rs565649812 Chr.7: 22942685-22942685 on Build GRCh38
rs1053696540 Chr.7: 22942738-22942738 on Build GRCh38
rs755640127 Chr.7: 22942376-22942376 on Build GRCh38
rs745903632 Chr.7: 22942734-22942734 on Build GRCh38
rs1052076106 Chr.7: 22942589-22942589 on Build GRCh38
rs892460008 Chr.7: 22942527-22942527 on Build GRCh38
rs916063636 Chr.7: 22942514-22942514 on Build GRCh38
rs534758021 Chr.7: 22942715-22942715 on Build GRCh38
rs1039473512 Chr.7: 22942726-22942726 on Build GRCh38
rs546858657 Chr.7: 22942684-22942684 on Build GRCh38
rs960253170 Chr.7: 22942439-22942439 on Build GRCh38
rs886062198 Chr.7: 22942767-22942767 on Build GRCh38
rs544494239 Chr.7: 22942319-22942319 on Build GRCh38
rs371082438 Chr.7: 22942545-22942545 on Build GRCh38
rs1025056531 Chr.7: 22942543-22942543 on Build GRCh38
rs887170901 Chr.7: 22942705-22942705 on Build GRCh38
rs35754549 Chr.7: 22942654-22942655 on Build GRCh38
rs533116880 Chr.7: 22942660-22942660 on Build GRCh38
rs899680186 Chr.7: 22942468-22942468 on Build GRCh38
rs570624287 Chr.7: 22942655-22942655 on Build GRCh38
rs1046802239 Chr.7: 22942371-22942371 on Build GRCh38
rs943921712 Chr.7: 22942616-22942616 on Build GRCh38
rs75393023 Chr.7: 22942323-22942323 on Build GRCh38
rs75640317 Chr.7: 22942711-22942711 on Build GRCh38
rs919541852 Chr.7: 22942682-22942682 on Build GRCh38
rs995715482 Chr.7: 22942675-22942675 on Build GRCh38
rs947582671 Chr.7: 22942530-22942530 on Build GRCh38
rs2286495 Chr.7: 22942588-22942588 on Build GRCh38
rs1034237025 Chr.7: 22942627-22942627 on Build GRCh38
rs1009489363 Chr.7: 22942778-22942778 on Build GRCh38
rs770000212 Chr.7: 22942763-22942763 on Build GRCh38
rs921079211 Chr.7: 22942732-22942732 on Build GRCh38
rs912237747 Chr.7: 22942714-22942714 on Build GRCh38
rs535464620 Chr.7: 22942764-22942764 on Build GRCh38
rs779639667 Chr.7: 22942396-22942396 on Build GRCh38
rs560323467 Chr.7: 22942340-22942340 on Build GRCh38
rs529230114 Chr.7: 22942398-22942398 on Build GRCh38
rs951506134 Chr.7: 22942381-22942381 on Build GRCh38
rs982709922 Chr.7: 22942420-22942420 on Build GRCh38
rs899660131 Chr.7: 22942658-22942658 on Build GRCh38
rs757408164 Chr.7: 22942422-22942422 on Build GRCh38
rs151155558 Chr.7: 22942322-22942322 on Build GRCh38
rs17725943 Chr.7: 22942613-22942613 on Build GRCh38

View Full Product Details