Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 28570277-28570779 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CREB5
Entrez Gene ID
9586
Gene Name
cAMP responsive element binding protein 5
Gene Aliases
CRE-BPA, CREB-5
UniGene
Hs.437075
Gene Chromosome Location
Chr.7: 28299321-28825894 on Build GRCh38
Transcripts
NM_004904.3 NM_182898.3 NM_182899.4 XM_005249906.1 XM_017012806.1 XM_017012807.1 XM_017012808.1 XM_017012809.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
131375 c.430A>G p.I144V Chr.7: 28570503-28570503 on Build GRCh38
3027613 c.449C>A p.T150N Chr.7: 28570522-28570522 on Build GRCh38
3027611 c.411G>A p.S137S Chr.7: 28570484-28570484 on Build GRCh38
382457 c.455G>T p.R152L Chr.7: 28570528-28570528 on Build GRCh38
139697 c.371A>G p.N124S Chr.7: 28570444-28570444 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs148137879 Chr.7: 28570481-28570481 on Build GRCh38
rs148124349 Chr.7: 28570389-28570389 on Build GRCh38
rs778367926 Chr.7: 28570568-28570568 on Build GRCh38
rs919888979 Chr.7: 28570645-28570645 on Build GRCh38
rs1033613186 Chr.7: 28570346-28570346 on Build GRCh38
rs895022850 Chr.7: 28570368-28570368 on Build GRCh38
rs537303404 Chr.7: 28570403-28570403 on Build GRCh38
rs758944555 Chr.7: 28570464-28570464 on Build GRCh38
rs755453263 Chr.7: 28570397-28570397 on Build GRCh38
rs751687494 Chr.7: 28570347-28570347 on Build GRCh38
rs578021502 Chr.7: 28570592-28570592 on Build GRCh38
rs187764543 Chr.7: 28570548-28570548 on Build GRCh38
rs75755102 Chr.7: 28570718-28570718 on Build GRCh38
rs371699221 Chr.7: 28570499-28570499 on Build GRCh38
rs557766445 Chr.7: 28570430-28570430 on Build GRCh38
rs779844663 Chr.7: 28570582-28570582 on Build GRCh38
rs116248568 Chr.7: 28570435-28570435 on Build GRCh38
rs1003348535 Chr.7: 28570324-28570324 on Build GRCh38
rs374897586 Chr.7: 28570527-28570527 on Build GRCh38
rs780310291 Chr.7: 28570381-28570381 on Build GRCh38
rs375100511 Chr.7: 28570526-28570526 on Build GRCh38
rs939167899 Chr.7: 28570677-28570677 on Build GRCh38
rs762867307 Chr.7: 28570431-28570431 on Build GRCh38
rs555693522 Chr.7: 28570480-28570480 on Build GRCh38
rs370145745 Chr.7: 28570409-28570409 on Build GRCh38
rs752220683 Chr.7: 28570355-28570355 on Build GRCh38
rs776456460 Chr.7: 28570321-28570321 on Build GRCh38
rs994518591 Chr.7: 28570382-28570382 on Build GRCh38
rs781121756 Chr.7: 28570572-28570572 on Build GRCh38
rs778030866 Chr.7: 28570475-28570475 on Build GRCh38
rs1054928886 Chr.7: 28570692-28570692 on Build GRCh38
rs751259024 Chr.7: 28570372-28570372 on Build GRCh38
rs371367477 Chr.7: 28570484-28570484 on Build GRCh38
rs759322003 Chr.7: 28570422-28570422 on Build GRCh38
rs765095165 Chr.7: 28570423-28570423 on Build GRCh38
rs28699333 Chr.7: 28570561-28570561 on Build GRCh38
rs995158184 Chr.7: 28570700-28570700 on Build GRCh38
rs75512111 Chr.7: 28570518-28570518 on Build GRCh38
rs757839730 Chr.7: 28570358-28570358 on Build GRCh38
rs767135866 Chr.7: 28570437-28570437 on Build GRCh38
rs928717904 Chr.7: 28570716-28570716 on Build GRCh38
rs199608009 Chr.7: 28570457-28570457 on Build GRCh38
rs367885203 Chr.7: 28570494-28570494 on Build GRCh38
rs772760663 Chr.7: 28570339-28570339 on Build GRCh38
rs114450022 Chr.7: 28570528-28570528 on Build GRCh38
rs368333429 Chr.7: 28570706-28570706 on Build GRCh38
rs759422811 Chr.7: 28570323-28570323 on Build GRCh38
rs779409043 Chr.7: 28570398-28570398 on Build GRCh38
rs766137403 Chr.7: 28570318-28570318 on Build GRCh38
rs755954277 Chr.7: 28570458-28570460 on Build GRCh38
rs765177374 Chr.7: 28570338-28570338 on Build GRCh38
rs895096493 Chr.7: 28570699-28570699 on Build GRCh38
rs746154339 Chr.7: 28570578-28570578 on Build GRCh38
rs748715346 Chr.7: 28570458-28570458 on Build GRCh38
rs756899888 Chr.7: 28570373-28570373 on Build GRCh38
rs751599607 Chr.7: 28570571-28570571 on Build GRCh38
rs961310435 Chr.7: 28570377-28570377 on Build GRCh38
rs114327290 Chr.7: 28570402-28570402 on Build GRCh38
rs563653537 Chr.7: 28570715-28570715 on Build GRCh38
rs763592770 Chr.7: 28570361-28570361 on Build GRCh38
rs755436114 Chr.7: 28570455-28570455 on Build GRCh38
rs762906403 Chr.7: 28570496-28570496 on Build GRCh38
rs575224753 Chr.7: 28570729-28570729 on Build GRCh38
rs976850317 Chr.7: 28570459-28570459 on Build GRCh38
rs750146422 Chr.7: 28570535-28570535 on Build GRCh38
rs112201672 Chr.7: 28570628-28570628 on Build GRCh38
rs753073429 Chr.7: 28570558-28570558 on Build GRCh38
rs368299466 Chr.7: 28570317-28570317 on Build GRCh38
rs536658106 Chr.7: 28570734-28570734 on Build GRCh38
rs1027724917 Chr.7: 28570406-28570406 on Build GRCh38
rs373416267 Chr.7: 28570379-28570379 on Build GRCh38
rs1044493052 Chr.7: 28570446-28570446 on Build GRCh38
rs906591857 Chr.7: 28570629-28570629 on Build GRCh38
rs143789664 Chr.7: 28570410-28570410 on Build GRCh38
rs776313239 Chr.7: 28570414-28570414 on Build GRCh38
rs573930655 Chr.7: 28570536-28570536 on Build GRCh38
rs754158273 Chr.7: 28570447-28570447 on Build GRCh38
rs746075945 Chr.7: 28570483-28570483 on Build GRCh38
rs1027354280 Chr.7: 28570732-28570732 on Build GRCh38
rs761100378 Chr.7: 28570434-28570434 on Build GRCh38

View Full Product Details