Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 30160107-30160644 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MTURN
Entrez Gene ID
222166
Gene Name
maturin, neural progenitor differentiation regulator homolog (Xenopus)
Gene Aliases
C7orf41, Ells1
UniGene
Hs.200100
Gene Chromosome Location
Chr.7: 30134936-30162765 on Build GRCh38
Transcripts
NM_152793.2 XM_005249652.3

SNP Detail

SNP ID Chromosome Location
rs905951040 Chr.7: 30160613-30160613 on Build GRCh38
rs773958583 Chr.7: 30160600-30160600 on Build GRCh38
rs572475988 Chr.7: 30160350-30160350 on Build GRCh38
rs900935335 Chr.7: 30160237-30160237 on Build GRCh38
rs193251343 Chr.7: 30160496-30160496 on Build GRCh38
rs189304848 Chr.7: 30160446-30160446 on Build GRCh38
rs867107808 Chr.7: 30160309-30160309 on Build GRCh38
rs76612741 Chr.7: 30160571-30160571 on Build GRCh38
rs772403390 Chr.7: 30160377-30160377 on Build GRCh38
rs891852242 Chr.7: 30160200-30160200 on Build GRCh38
rs758759023 Chr.7: 30160571-30160572 on Build GRCh38
rs917980463 Chr.7: 30160451-30160451 on Build GRCh38
rs747088476 Chr.7: 30160441-30160441 on Build GRCh38
rs776859742 Chr.7: 30160524-30160524 on Build GRCh38
rs17547923 Chr.7: 30160181-30160181 on Build GRCh38
rs929024255 Chr.7: 30160440-30160440 on Build GRCh38
rs1021704206 Chr.7: 30160598-30160598 on Build GRCh38
rs952564395 Chr.7: 30160336-30160336 on Build GRCh38
rs184362894 Chr.7: 30160421-30160421 on Build GRCh38
rs3750090 Chr.7: 30160344-30160344 on Build GRCh38
rs902057470 Chr.7: 30160525-30160525 on Build GRCh38
rs549396038 Chr.7: 30160356-30160356 on Build GRCh38
rs1005466762 Chr.7: 30160337-30160337 on Build GRCh38
rs192748203 Chr.7: 30160297-30160297 on Build GRCh38
rs997886005 Chr.7: 30160162-30160162 on Build GRCh38
rs775825865 Chr.7: 30160599-30160602 on Build GRCh38
rs943571220 Chr.7: 30160481-30160481 on Build GRCh38
rs1039725561 Chr.7: 30160142-30160142 on Build GRCh38
rs1054002168 Chr.7: 30160177-30160177 on Build GRCh38
rs746393822 Chr.7: 30160365-30160365 on Build GRCh38
rs139256709 Chr.7: 30160562-30160562 on Build GRCh38
rs570495763 Chr.7: 30160147-30160147 on Build GRCh38
rs576071397 Chr.7: 30160425-30160425 on Build GRCh38
rs574657425 Chr.7: 30160503-30160503 on Build GRCh38
rs900862907 Chr.7: 30160617-30160617 on Build GRCh38
rs967889537 Chr.7: 30160242-30160242 on Build GRCh38
rs1022072161 Chr.7: 30160220-30160220 on Build GRCh38
rs550541843 Chr.7: 30160286-30160286 on Build GRCh38
rs755893602 Chr.7: 30160143-30160143 on Build GRCh38
rs1006518382 Chr.7: 30160192-30160192 on Build GRCh38
rs933600435 Chr.7: 30160530-30160530 on Build GRCh38
rs527364553 Chr.7: 30160588-30160588 on Build GRCh38
rs993962246 Chr.7: 30160265-30160265 on Build GRCh38
rs1053025015 Chr.7: 30160537-30160537 on Build GRCh38
rs918946814 Chr.7: 30160409-30160409 on Build GRCh38
rs775967520 Chr.7: 30160216-30160216 on Build GRCh38
rs763066978 Chr.7: 30160607-30160610 on Build GRCh38
rs943618360 Chr.7: 30160141-30160141 on Build GRCh38
rs779109166 Chr.7: 30160301-30160301 on Build GRCh38
rs536004871 Chr.7: 30160303-30160303 on Build GRCh38
rs28701848 Chr.7: 30160381-30160381 on Build GRCh38
rs909369912 Chr.7: 30160454-30160454 on Build GRCh38
rs942344977 Chr.7: 30160586-30160586 on Build GRCh38

View Full Product Details