Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 34929933-34930473 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DPY19L1
Entrez Gene ID
23333
Gene Name
dpy-19 like 1 (C. elegans)
Gene Aliases
-
UniGene
Hs.408623
Gene Chromosome Location
Chr.7: 34928881-35038041 on Build GRCh38
Transcripts
NM_015283.1 XM_011515245.2 XM_011515246.2

SNP Detail

SNP ID Chromosome Location
rs936525843 Chr.7: 34930212-34930212 on Build GRCh38
rs151228391 Chr.7: 34930447-34930451 on Build GRCh38
rs933150359 Chr.7: 34929963-34929963 on Build GRCh38
rs1050726183 Chr.7: 34929969-34929969 on Build GRCh38
rs538561525 Chr.7: 34930011-34930011 on Build GRCh38
rs933099297 Chr.7: 34930202-34930202 on Build GRCh38
rs560494941 Chr.7: 34930311-34930311 on Build GRCh38
rs922895109 Chr.7: 34930419-34930419 on Build GRCh38
rs566400574 Chr.7: 34930001-34930001 on Build GRCh38
rs940326590 Chr.7: 34930152-34930152 on Build GRCh38
rs546481662 Chr.7: 34930393-34930393 on Build GRCh38
rs892441492 Chr.7: 34930025-34930025 on Build GRCh38
rs906543815 Chr.7: 34930307-34930307 on Build GRCh38
rs555080819 Chr.7: 34930210-34930210 on Build GRCh38
rs984350164 Chr.7: 34930089-34930089 on Build GRCh38
rs565564908 Chr.7: 34930396-34930396 on Build GRCh38
rs535875039 Chr.7: 34929954-34929954 on Build GRCh38
rs73693344 Chr.7: 34930361-34930361 on Build GRCh38
rs537944998 Chr.7: 34930076-34930076 on Build GRCh38
rs1032221297 Chr.7: 34930024-34930024 on Build GRCh38
rs1009503139 Chr.7: 34930264-34930264 on Build GRCh38
rs558426175 Chr.7: 34930027-34930027 on Build GRCh38
rs762346508 Chr.7: 34929991-34929991 on Build GRCh38
rs952828671 Chr.7: 34930326-34930326 on Build GRCh38
rs866229213 Chr.7: 34929953-34929953 on Build GRCh38
rs908808675 Chr.7: 34930145-34930145 on Build GRCh38
rs574865029 Chr.7: 34930213-34930213 on Build GRCh38
rs952914775 Chr.7: 34930077-34930077 on Build GRCh38
rs961662413 Chr.7: 34930373-34930373 on Build GRCh38
rs892410030 Chr.7: 34930233-34930233 on Build GRCh38
rs1002662259 Chr.7: 34930311-34930312 on Build GRCh38
rs1033788583 Chr.7: 34930323-34930323 on Build GRCh38
rs1034232113 Chr.7: 34930061-34930061 on Build GRCh38
rs1015836555 Chr.7: 34930363-34930363 on Build GRCh38
rs971241045 Chr.7: 34930031-34930031 on Build GRCh38
rs116517688 Chr.7: 34929960-34929960 on Build GRCh38
rs182844321 Chr.7: 34930028-34930028 on Build GRCh38
rs910336849 Chr.7: 34930208-34930208 on Build GRCh38
rs889030141 Chr.7: 34929976-34929976 on Build GRCh38
rs1050190664 Chr.7: 34930205-34930205 on Build GRCh38
rs540606802 Chr.7: 34930229-34930229 on Build GRCh38
rs1046868965 Chr.7: 34930274-34930274 on Build GRCh38
rs971715405 Chr.7: 34930375-34930375 on Build GRCh38
rs980958630 Chr.7: 34930040-34930040 on Build GRCh38
rs923043407 Chr.7: 34930188-34930188 on Build GRCh38
rs971747966 Chr.7: 34930153-34930153 on Build GRCh38

View Full Product Details