Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 34949513-34950011 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DPY19L1
Entrez Gene ID
23333
Gene Name
dpy-19 like 1 (C. elegans)
Gene Aliases
-
UniGene
Hs.408623
Gene Chromosome Location
Chr.7: 34928881-35038041 on Build GRCh38
Transcripts
NM_015283.1 XM_011515245.2 XM_011515246.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6383428 c.1102G>T p.A368S Chr.7: 34949898-34949898 on Build GRCh38
6848928 c.1180G>A p.E394K Chr.7: 34949820-34949820 on Build GRCh38
6177553 c.1182G>A p.E394E Chr.7: 34949818-34949818 on Build GRCh38
260574 c.1134C>A p.F378L Chr.7: 34949866-34949866 on Build GRCh38
7282697 c.1168A>G p.T390A Chr.7: 34949832-34949832 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs923972975 Chr.7: 34949770-34949770 on Build GRCh38
rs770147327 Chr.7: 34949893-34949893 on Build GRCh38
rs539139473 Chr.7: 34949913-34949913 on Build GRCh38
rs749831714 Chr.7: 34949902-34949902 on Build GRCh38
rs760833085 Chr.7: 34949949-34949949 on Build GRCh38
rs898720536 Chr.7: 34949966-34949966 on Build GRCh38
rs1025973775 Chr.7: 34949978-34949978 on Build GRCh38
rs773126149 Chr.7: 34949924-34949924 on Build GRCh38
rs530822573 Chr.7: 34949693-34949693 on Build GRCh38
rs987249232 Chr.7: 34949669-34949669 on Build GRCh38
rs529825976 Chr.7: 34949760-34949760 on Build GRCh38
rs759465714 Chr.7: 34949766-34949790 on Build GRCh38
rs758131531 Chr.7: 34949878-34949878 on Build GRCh38
rs974609201 Chr.7: 34949707-34949707 on Build GRCh38
rs769002140 Chr.7: 34949912-34949912 on Build GRCh38
rs767785697 Chr.7: 34949918-34949918 on Build GRCh38
rs2021391 Chr.7: 34949752-34949752 on Build GRCh38
rs778895208 Chr.7: 34949863-34949863 on Build GRCh38
rs200207679 Chr.7: 34949822-34949822 on Build GRCh38
rs540843412 Chr.7: 34949603-34949603 on Build GRCh38
rs750529586 Chr.7: 34949818-34949818 on Build GRCh38
rs994444037 Chr.7: 34949969-34949969 on Build GRCh38
rs573411766 Chr.7: 34949703-34949703 on Build GRCh38
rs2021390 Chr.7: 34949725-34949725 on Build GRCh38
rs1038538359 Chr.7: 34949941-34949941 on Build GRCh38
rs115629967 Chr.7: 34949665-34949665 on Build GRCh38
rs777093123 Chr.7: 34949880-34949880 on Build GRCh38
rs746557160 Chr.7: 34949892-34949892 on Build GRCh38
rs753639394 Chr.7: 34949833-34949833 on Build GRCh38
rs948187493 Chr.7: 34949935-34949935 on Build GRCh38
rs754798845 Chr.7: 34949836-34949836 on Build GRCh38
rs939418698 Chr.7: 34949773-34949773 on Build GRCh38
rs747800979 Chr.7: 34949873-34949873 on Build GRCh38
rs558989791 Chr.7: 34949608-34949608 on Build GRCh38
rs564583395 Chr.7: 34949690-34949690 on Build GRCh38
rs915101824 Chr.7: 34949716-34949716 on Build GRCh38
rs552603930 Chr.7: 34949963-34949963 on Build GRCh38
rs1018773486 Chr.7: 34949700-34949700 on Build GRCh38
rs761964134 Chr.7: 34949914-34949914 on Build GRCh38
rs370647575 Chr.7: 34949901-34949901 on Build GRCh38
rs895311404 Chr.7: 34949911-34949911 on Build GRCh38
rs767481613 Chr.7: 34949771-34949771 on Build GRCh38
rs1026006382 Chr.7: 34949622-34949622 on Build GRCh38
rs762087250 Chr.7: 34949764-34949764 on Build GRCh38
rs755938303 Chr.7: 34949821-34949821 on Build GRCh38
rs572611513 Chr.7: 34949614-34949614 on Build GRCh38
rs776801356 Chr.7: 34949751-34949753 on Build GRCh38
rs183013768 Chr.7: 34949586-34949586 on Build GRCh38

View Full Product Details