Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 36325998-36326497 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KIAA0895
Entrez Gene ID
23366
Gene Name
KIAA0895
Gene Aliases
-
UniGene
Hs.6224
Gene Chromosome Location
Chr.7: 36324150-36390125 on Build GRCh38
Transcripts
NM_001100425.1 NM_001199706.1 NM_001199707.1 NM_001199708.1 NM_001300956.1 NM_015314.2 XM_005249689.3 XM_005249690.4

SNP Detail

SNP ID Chromosome Location
rs1036406106 Chr.7: 36326039-36326039 on Build GRCh38
rs940336101 Chr.7: 36326185-36326185 on Build GRCh38
rs966109095 Chr.7: 36326184-36326184 on Build GRCh38
rs867309170 Chr.7: 36326408-36326408 on Build GRCh38
rs981544213 Chr.7: 36326087-36326087 on Build GRCh38
rs958855202 Chr.7: 36326443-36326443 on Build GRCh38
rs1036396111 Chr.7: 36326417-36326417 on Build GRCh38
rs781467927 Chr.7: 36326059-36326059 on Build GRCh38
rs748667038 Chr.7: 36326168-36326168 on Build GRCh38
rs1010632165 Chr.7: 36326293-36326293 on Build GRCh38
rs546753537 Chr.7: 36326299-36326299 on Build GRCh38
rs150689669 Chr.7: 36326208-36326208 on Build GRCh38
rs376961191 Chr.7: 36326343-36326343 on Build GRCh38
rs552879406 Chr.7: 36326079-36326079 on Build GRCh38
rs927471446 Chr.7: 36326105-36326105 on Build GRCh38
rs997275045 Chr.7: 36326217-36326217 on Build GRCh38
rs149663178 Chr.7: 36326288-36326288 on Build GRCh38
rs868006038 Chr.7: 36326164-36326164 on Build GRCh38
rs971418126 Chr.7: 36326315-36326315 on Build GRCh38
rs530816315 Chr.7: 36326204-36326205 on Build GRCh38
rs981491677 Chr.7: 36326320-36326320 on Build GRCh38
rs1045730432 Chr.7: 36326361-36326361 on Build GRCh38
rs1049296533 Chr.7: 36326161-36326161 on Build GRCh38
rs1018584505 Chr.7: 36326144-36326144 on Build GRCh38
rs1007907404 Chr.7: 36326135-36326135 on Build GRCh38
rs888960287 Chr.7: 36326459-36326459 on Build GRCh38
rs897941701 Chr.7: 36326430-36326430 on Build GRCh38
rs1036051949 Chr.7: 36326205-36326208 on Build GRCh38
rs34466907 Chr.7: 36326221-36326221 on Build GRCh38
rs182078934 Chr.7: 36326451-36326451 on Build GRCh38
rs927944734 Chr.7: 36326414-36326414 on Build GRCh38
rs1027514113 Chr.7: 36326130-36326130 on Build GRCh38
rs1894877 Chr.7: 36326450-36326450 on Build GRCh38
rs1032662754 Chr.7: 36326286-36326286 on Build GRCh38
rs888950801 Chr.7: 36326267-36326267 on Build GRCh38
rs994662985 Chr.7: 36326091-36326091 on Build GRCh38
rs548058257 Chr.7: 36326341-36326341 on Build GRCh38
rs939386178 Chr.7: 36326416-36326416 on Build GRCh38
rs1003593391 Chr.7: 36326026-36326026 on Build GRCh38
rs927420500 Chr.7: 36326402-36326402 on Build GRCh38
rs924396889 Chr.7: 36326218-36326218 on Build GRCh38
rs139486352 Chr.7: 36326283-36326283 on Build GRCh38
rs191203190 Chr.7: 36326131-36326131 on Build GRCh38
rs866093202 Chr.7: 36326028-36326028 on Build GRCh38

View Full Product Details