Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 50444633-50445140 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FIGNL1
Entrez Gene ID
63979
Gene Name
fidgetin like 1
Gene Aliases
-
UniGene
Hs.137516
Gene Chromosome Location
Chr.7: 50437327-50450390 on Build GRCh38
Transcripts
NM_001042762.2 NM_001287492.1 NM_001287493.1 NM_001287494.1 NM_001287495.1 NM_001287496.1 NM_022116.4 XM_011515470.2 XM_017012501.1 XM_017012502.1

SNP Detail

SNP ID Chromosome Location
rs540418239 Chr.7: 50445060-50445060 on Build GRCh38
rs765361948 Chr.7: 50444698-50444698 on Build GRCh38
rs117834634 Chr.7: 50444756-50444756 on Build GRCh38
rs554918085 Chr.7: 50444897-50444897 on Build GRCh38
rs570925423 Chr.7: 50444784-50444784 on Build GRCh38
rs1053462449 Chr.7: 50445102-50445102 on Build GRCh38
rs920558535 Chr.7: 50444872-50444872 on Build GRCh38
rs752709815 Chr.7: 50444742-50444742 on Build GRCh38
rs529188313 Chr.7: 50445069-50445069 on Build GRCh38
rs1017549289 Chr.7: 50444801-50444801 on Build GRCh38
rs182615943 Chr.7: 50444786-50444786 on Build GRCh38
rs187628199 Chr.7: 50445026-50445026 on Build GRCh38
rs952602989 Chr.7: 50445005-50445005 on Build GRCh38
rs1001902365 Chr.7: 50444694-50444694 on Build GRCh38
rs1032119042 Chr.7: 50444776-50444776 on Build GRCh38
rs151218426 Chr.7: 50444989-50444989 on Build GRCh38
rs1050339814 Chr.7: 50444689-50444689 on Build GRCh38
rs535778643 Chr.7: 50444837-50444837 on Build GRCh38
rs906097864 Chr.7: 50444690-50444690 on Build GRCh38
rs983542251 Chr.7: 50444984-50444984 on Build GRCh38
rs957785863 Chr.7: 50444723-50444723 on Build GRCh38
rs932786136 Chr.7: 50444685-50444685 on Build GRCh38
rs144556627 Chr.7: 50444959-50444959 on Build GRCh38
rs976727633 Chr.7: 50445081-50445081 on Build GRCh38
rs566813317 Chr.7: 50444830-50444830 on Build GRCh38
rs970641267 Chr.7: 50444915-50444915 on Build GRCh38
rs1044941730 Chr.7: 50445059-50445059 on Build GRCh38
rs574913173 Chr.7: 50444949-50444949 on Build GRCh38
rs1024859621 Chr.7: 50444819-50444819 on Build GRCh38
rs193220404 Chr.7: 50445068-50445068 on Build GRCh38
rs1020800643 Chr.7: 50444762-50444762 on Build GRCh38
rs922636591 Chr.7: 50445112-50445112 on Build GRCh38
rs1470746 Chr.7: 50444816-50444816 on Build GRCh38
rs551079147 Chr.7: 50444783-50444783 on Build GRCh38
rs367723994 Chr.7: 50444657-50444657 on Build GRCh38
rs769216413 Chr.7: 50445015-50445016 on Build GRCh38
rs950250105 Chr.7: 50444996-50444996 on Build GRCh38
rs531005387 Chr.7: 50444770-50444770 on Build GRCh38

View Full Product Details