Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 66921095-66921587 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TMEM248
Entrez Gene ID
55069
Gene Name
transmembrane protein 248
Gene Aliases
C7orf42
UniGene
Hs.488478
Gene Chromosome Location
Chr.7: 66921216-66958551 on Build GRCh38
Transcripts
NM_017994.4

SNP Detail

SNP ID Chromosome Location
rs986679907 Chr.7: 66921502-66921502 on Build GRCh38
rs1043385247 Chr.7: 66921166-66921166 on Build GRCh38
rs962348787 Chr.7: 66921449-66921449 on Build GRCh38
rs768134332 Chr.7: 66921208-66921208 on Build GRCh38
rs1049940215 Chr.7: 66921119-66921119 on Build GRCh38
rs1021751632 Chr.7: 66921543-66921543 on Build GRCh38
rs892956157 Chr.7: 66921135-66921135 on Build GRCh38
rs952137812 Chr.7: 66921223-66921223 on Build GRCh38
rs1043335095 Chr.7: 66921317-66921317 on Build GRCh38
rs966766683 Chr.7: 66921463-66921463 on Build GRCh38
rs946371972 Chr.7: 66921151-66921151 on Build GRCh38
rs959323365 Chr.7: 66921182-66921182 on Build GRCh38
rs1024123432 Chr.7: 66921212-66921212 on Build GRCh38
rs1006199619 Chr.7: 66921224-66921224 on Build GRCh38
rs935046352 Chr.7: 66921332-66921332 on Build GRCh38
rs976730436 Chr.7: 66921268-66921268 on Build GRCh38
rs997550158 Chr.7: 66921459-66921459 on Build GRCh38
rs1015048085 Chr.7: 66921438-66921438 on Build GRCh38
rs946486592 Chr.7: 66921305-66921305 on Build GRCh38
rs1014934983 Chr.7: 66921238-66921238 on Build GRCh38
rs986345132 Chr.7: 66921286-66921287 on Build GRCh38
rs368089373 Chr.7: 66921471-66921471 on Build GRCh38
rs999852571 Chr.7: 66921186-66921186 on Build GRCh38
rs1030987354 Chr.7: 66921468-66921468 on Build GRCh38
rs962252881 Chr.7: 66921264-66921264 on Build GRCh38
rs910941227 Chr.7: 66921301-66921301 on Build GRCh38
rs532249935 Chr.7: 66921170-66921170 on Build GRCh38
rs1056266355 Chr.7: 66921193-66921193 on Build GRCh38
rs887840357 Chr.7: 66921425-66921425 on Build GRCh38
rs1012483139 Chr.7: 66921389-66921389 on Build GRCh38
rs902416307 Chr.7: 66921173-66921173 on Build GRCh38
rs553678308 Chr.7: 66921334-66921334 on Build GRCh38
rs1044934450 Chr.7: 66921423-66921423 on Build GRCh38
rs992463141 Chr.7: 66921225-66921225 on Build GRCh38
rs2293350 Chr.7: 66921348-66921348 on Build GRCh38
rs932200885 Chr.7: 66921118-66921118 on Build GRCh38
rs370252488 Chr.7: 66921198-66921198 on Build GRCh38
rs947274156 Chr.7: 66921148-66921148 on Build GRCh38
rs896301815 Chr.7: 66921197-66921197 on Build GRCh38
rs953666257 Chr.7: 66921473-66921473 on Build GRCh38
rs923617848 Chr.7: 66921322-66921322 on Build GRCh38
rs542945150 Chr.7: 66921185-66921185 on Build GRCh38
rs923583919 Chr.7: 66921273-66921273 on Build GRCh38
rs76993623 Chr.7: 66921163-66921163 on Build GRCh38
rs760468722 Chr.7: 66921275-66921275 on Build GRCh38
rs1044688905 Chr.7: 66921150-66921150 on Build GRCh38
rs1006316067 Chr.7: 66921430-66921430 on Build GRCh38
rs544565640 Chr.7: 66921407-66921407 on Build GRCh38
rs1012123654 Chr.7: 66921200-66921200 on Build GRCh38

View Full Product Details