Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 100089158-100089684 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COPS6
Entrez Gene ID
10980
Gene Name
COP9 signalosome subunit 6
Gene Aliases
CSN6, MOV34-34KD
UniGene
Hs.15591
Gene Chromosome Location
Chr.7: 100088960-100092200 on Build GRCh38
Transcripts
NM_006833.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6634105 c.173T>C p.M58T Chr.7: 100089386-100089386 on Build GRCh38
1453346 c.179C>T p.S60F Chr.7: 100089392-100089392 on Build GRCh38
6012799 c.182A>C p.Q61P Chr.7: 100089395-100089395 on Build GRCh38
3784105 c.80T>G p.V27G Chr.7: 100089293-100089293 on Build GRCh38
6634101 c.94G>T p.A32S Chr.7: 100089307-100089307 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs769679183 Chr.7: 100089652-100089652 on Build GRCh38
rs759665460 Chr.7: 100089272-100089272 on Build GRCh38
rs764743282 Chr.7: 100089460-100089460 on Build GRCh38
rs905104838 Chr.7: 100089567-100089567 on Build GRCh38
rs746980015 Chr.7: 100089363-100089363 on Build GRCh38
rs377698165 Chr.7: 100089610-100089610 on Build GRCh38
rs936614442 Chr.7: 100089597-100089597 on Build GRCh38
rs201177602 Chr.7: 100089426-100089426 on Build GRCh38
rs940060737 Chr.7: 100089510-100089510 on Build GRCh38
rs112097083 Chr.7: 100089287-100089287 on Build GRCh38
rs981614771 Chr.7: 100089213-100089213 on Build GRCh38
rs776623662 Chr.7: 100089398-100089398 on Build GRCh38
rs908728266 Chr.7: 100089506-100089506 on Build GRCh38
rs560041581 Chr.7: 100089200-100089200 on Build GRCh38
rs928394693 Chr.7: 100089582-100089582 on Build GRCh38
rs74881871 Chr.7: 100089568-100089568 on Build GRCh38
rs530584388 Chr.7: 100089505-100089505 on Build GRCh38
rs767625256 Chr.7: 100089427-100089427 on Build GRCh38
rs774664390 Chr.7: 100089419-100089419 on Build GRCh38
rs780113727 Chr.7: 100089351-100089351 on Build GRCh38
rs189921210 Chr.7: 100089209-100089209 on Build GRCh38
rs981199781 Chr.7: 100089538-100089538 on Build GRCh38
rs757812170 Chr.7: 100089577-100089577 on Build GRCh38
rs781279064 Chr.7: 100089630-100089630 on Build GRCh38
rs779774012 Chr.7: 100089588-100089588 on Build GRCh38
rs564221516 Chr.7: 100089481-100089481 on Build GRCh38
rs928255610 Chr.7: 100089220-100089220 on Build GRCh38
rs747987937 Chr.7: 100089405-100089405 on Build GRCh38
rs895821090 Chr.7: 100089233-100089233 on Build GRCh38
rs371808452 Chr.7: 100089253-100089253 on Build GRCh38
rs754138783 Chr.7: 100089449-100089449 on Build GRCh38
rs778635465 Chr.7: 100089315-100089315 on Build GRCh38
rs767862494 Chr.7: 100089277-100089277 on Build GRCh38
rs999520098 Chr.7: 100089443-100089443 on Build GRCh38
rs766467408 Chr.7: 100089271-100089271 on Build GRCh38
rs948719077 Chr.7: 100089532-100089532 on Build GRCh38
rs764511194 Chr.7: 100089295-100089295 on Build GRCh38
rs532223057 Chr.7: 100089483-100089483 on Build GRCh38
rs369283004 Chr.7: 100089381-100089381 on Build GRCh38
rs745334784 Chr.7: 100089333-100089333 on Build GRCh38
rs761230269 Chr.7: 100089432-100089432 on Build GRCh38
rs753478190 Chr.7: 100089301-100089301 on Build GRCh38
rs965048279 Chr.7: 100089452-100089452 on Build GRCh38
rs997586354 Chr.7: 100089375-100089375 on Build GRCh38
rs373332701 Chr.7: 100089564-100089564 on Build GRCh38
rs915736840 Chr.7: 100089183-100089183 on Build GRCh38
rs771318921 Chr.7: 100089520-100089520 on Build GRCh38
rs374887563 Chr.7: 100089431-100089431 on Build GRCh38
rs992935192 Chr.7: 100089516-100089516 on Build GRCh38
rs957277064 Chr.7: 100089476-100089476 on Build GRCh38
rs952972028 Chr.7: 100089491-100089491 on Build GRCh38
rs746928513 Chr.7: 100089609-100089609 on Build GRCh38
rs775974257 Chr.7: 100089429-100089429 on Build GRCh38
rs4729575 Chr.7: 100089250-100089250 on Build GRCh38
rs747857640 Chr.7: 100089641-100089641 on Build GRCh38
rs375171440 Chr.7: 100089259-100089259 on Build GRCh38
rs762179736 Chr.7: 100089594-100089594 on Build GRCh38
rs553341823 Chr.7: 100089521-100089521 on Build GRCh38
rs756440179 Chr.7: 100089294-100089294 on Build GRCh38
rs776811618 Chr.7: 100089542-100089542 on Build GRCh38
rs764601110 Chr.7: 100089438-100089438 on Build GRCh38
rs942187605 Chr.7: 100089251-100089251 on Build GRCh38
rs753252878 Chr.7: 100089288-100089288 on Build GRCh38
rs369230943 Chr.7: 100089457-100089457 on Build GRCh38
rs1032204134 Chr.7: 100089347-100089347 on Build GRCh38
rs371283537 Chr.7: 100089628-100089628 on Build GRCh38
rs1019658111 Chr.7: 100089430-100089430 on Build GRCh38
rs773946699 Chr.7: 100089446-100089448 on Build GRCh38
rs949271193 Chr.7: 100089237-100089237 on Build GRCh38
rs756804877 Chr.7: 100089312-100089312 on Build GRCh38
rs757945167 Chr.7: 100089342-100089342 on Build GRCh38
rs750136119 Chr.7: 100089464-100089464 on Build GRCh38

View Full Product Details