Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 158184848-158185395 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CD1D
Entrez Gene ID
912
Gene Name
CD1d molecule
Gene Aliases
CD1A, R3, R3G1
UniGene
Hs.1799
Gene Chromosome Location
Chr.1: 158178038-158186427 on Build GRCh38
Transcripts
NM_001319145.1 NM_001766.3 XM_005245583.1 XM_006711621.1 XM_011510125.1 XM_011510127.1

SNP Detail

SNP ID Chromosome Location
rs961489891 Chr.1: 158185206-158185206 on Build GRCh38
rs936478980 Chr.1: 158184921-158184921 on Build GRCh38
rs1040724209 Chr.1: 158185019-158185019 on Build GRCh38
rs981296822 Chr.1: 158185302-158185302 on Build GRCh38
rs929438650 Chr.1: 158185013-158185013 on Build GRCh38
rs1049271727 Chr.1: 158185020-158185020 on Build GRCh38
rs531897145 Chr.1: 158185265-158185265 on Build GRCh38
rs888162852 Chr.1: 158185055-158185055 on Build GRCh38
rs1005693645 Chr.1: 158185085-158185085 on Build GRCh38
rs984918183 Chr.1: 158185360-158185360 on Build GRCh38
rs926468074 Chr.1: 158184916-158184918 on Build GRCh38
rs926654876 Chr.1: 158184951-158184951 on Build GRCh38
rs781293143 Chr.1: 158185111-158185111 on Build GRCh38
rs961015927 Chr.1: 158185308-158185308 on Build GRCh38
rs112871117 Chr.1: 158184882-158184882 on Build GRCh38
rs993311116 Chr.1: 158185248-158185248 on Build GRCh38
rs949313652 Chr.1: 158184883-158184883 on Build GRCh38
rs188278184 Chr.1: 158185257-158185257 on Build GRCh38
rs892395546 Chr.1: 158185177-158185177 on Build GRCh38
rs569925501 Chr.1: 158185014-158185014 on Build GRCh38
rs968100452 Chr.1: 158185288-158185288 on Build GRCh38
rs989333479 Chr.1: 158185366-158185366 on Build GRCh38
rs968110262 Chr.1: 158184872-158184872 on Build GRCh38
rs762331197 Chr.1: 158184953-158184953 on Build GRCh38
rs998093108 Chr.1: 158185150-158185150 on Build GRCh38
rs370288659 Chr.1: 158184976-158184976 on Build GRCh38
rs1024347035 Chr.1: 158185253-158185253 on Build GRCh38
rs1039369398 Chr.1: 158185075-158185075 on Build GRCh38
rs970027663 Chr.1: 158185296-158185296 on Build GRCh38
rs572580113 Chr.1: 158185167-158185167 on Build GRCh38
rs1009318539 Chr.1: 158185280-158185280 on Build GRCh38
rs746834667 Chr.1: 158185007-158185007 on Build GRCh38
rs992193490 Chr.1: 158185318-158185318 on Build GRCh38
rs422236 Chr.1: 158185303-158185303 on Build GRCh38
rs192753052 Chr.1: 158184895-158184895 on Build GRCh38
rs573806789 Chr.1: 158185000-158185000 on Build GRCh38
rs184594994 Chr.1: 158185162-158185162 on Build GRCh38
rs899525113 Chr.1: 158185139-158185139 on Build GRCh38
rs756527425 Chr.1: 158185352-158185352 on Build GRCh38
rs1022085169 Chr.1: 158185287-158185287 on Build GRCh38
rs542337996 Chr.1: 158185065-158185065 on Build GRCh38
rs546172188 Chr.1: 158185226-158185226 on Build GRCh38
rs892236070 Chr.1: 158185003-158185003 on Build GRCh38
rs997095744 Chr.1: 158185031-158185031 on Build GRCh38
rs1057149523 Chr.1: 158184993-158184993 on Build GRCh38
rs769766041 Chr.1: 158185346-158185346 on Build GRCh38
rs1045000256 Chr.1: 158184913-158184913 on Build GRCh38
rs941093398 Chr.1: 158185057-158185057 on Build GRCh38
rs900827488 Chr.1: 158185030-158185031 on Build GRCh38
rs770804531 Chr.1: 158185011-158185011 on Build GRCh38

View Full Product Details