Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 158292510-158293022 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CD1B
Entrez Gene ID
910
Gene Name
CD1b molecule
Gene Aliases
CD1, CD1A, R1
UniGene
Hs.1310
Gene Chromosome Location
Chr.1: 158286841-158332038 on Build GRCh38
Transcripts
XM_017002785.1 NM_001765.2 XM_005245579.4

Gene Symbol
CD1C
Entrez Gene ID
911
Gene Name
CD1c molecule
Gene Aliases
BDCA1, CD1, CD1A, R7
UniGene
Hs.132448
Gene Chromosome Location
Chr.1: 158289773-158294774 on Build GRCh38
Transcripts
XM_017002785.1 NM_001765.2 XM_005245579.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4559466 c.805G>A p.E269K Chr.1: 158292790-158292790 on Build GRCh38
6459456 c.859C>T p.L287L Chr.1: 158292844-158292844 on Build GRCh38
6339784 c.858T>C p.S286S Chr.1: 158292843-158292843 on Build GRCh38
3976285 c.889+5G>T p.? Chr.1: 158292879-158292879 on Build GRCh38
5796903 c.763C>G p.L255V Chr.1: 158292748-158292748 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs758902960 Chr.1: 158292751-158292751 on Build GRCh38
rs150425079 Chr.1: 158292706-158292706 on Build GRCh38
rs762791288 Chr.1: 158292718-158292718 on Build GRCh38
rs989408136 Chr.1: 158292839-158292839 on Build GRCh38
rs185843886 Chr.1: 158292577-158292577 on Build GRCh38
rs776701345 Chr.1: 158292606-158292606 on Build GRCh38
rs774351498 Chr.1: 158292908-158292908 on Build GRCh38
rs149354390 Chr.1: 158292728-158292728 on Build GRCh38
rs180809448 Chr.1: 158292536-158292536 on Build GRCh38
rs144561206 Chr.1: 158292623-158292623 on Build GRCh38
rs766792421 Chr.1: 158292845-158292845 on Build GRCh38
rs146296717 Chr.1: 158292754-158292754 on Build GRCh38
rs145198081 Chr.1: 158292648-158292648 on Build GRCh38
rs760282222 Chr.1: 158292601-158292601 on Build GRCh38
rs766156680 Chr.1: 158292720-158292720 on Build GRCh38
rs369535551 Chr.1: 158292670-158292670 on Build GRCh38
rs761591081 Chr.1: 158292612-158292612 on Build GRCh38
rs780501974 Chr.1: 158292753-158292753 on Build GRCh38
rs947848706 Chr.1: 158292671-158292671 on Build GRCh38
rs867045352 Chr.1: 158292740-158292740 on Build GRCh38
rs35696985 Chr.1: 158292629-158292629 on Build GRCh38
rs3138103 Chr.1: 158292669-158292669 on Build GRCh38
rs775252151 Chr.1: 158292597-158292597 on Build GRCh38
rs988714073 Chr.1: 158292934-158292934 on Build GRCh38
rs201219470 Chr.1: 158292686-158292686 on Build GRCh38
rs376541578 Chr.1: 158292721-158292721 on Build GRCh38
rs756090125 Chr.1: 158292679-158292695 on Build GRCh38
rs777205106 Chr.1: 158292885-158292885 on Build GRCh38
rs780763972 Chr.1: 158292865-158292865 on Build GRCh38
rs992600148 Chr.1: 158292662-158292662 on Build GRCh38
rs945386090 Chr.1: 158292943-158292943 on Build GRCh38
rs767772289 Chr.1: 158292642-158292642 on Build GRCh38
rs768221526 Chr.1: 158292694-158292694 on Build GRCh38
rs3176970 Chr.1: 158292787-158292787 on Build GRCh38
rs957968420 Chr.1: 158292626-158292626 on Build GRCh38
rs768042167 Chr.1: 158292602-158292602 on Build GRCh38
rs752096853 Chr.1: 158292655-158292663 on Build GRCh38
rs774315129 Chr.1: 158292889-158292889 on Build GRCh38
rs373456243 Chr.1: 158292805-158292805 on Build GRCh38
rs150377514 Chr.1: 158292613-158292613 on Build GRCh38
rs866382982 Chr.1: 158292742-158292742 on Build GRCh38
rs3138104 Chr.1: 158292904-158292904 on Build GRCh38
rs41273485 Chr.1: 158292832-158292832 on Build GRCh38
rs762063601 Chr.1: 158292910-158292910 on Build GRCh38
rs374378621 Chr.1: 158292917-158292917 on Build GRCh38
rs147191023 Chr.1: 158292862-158292862 on Build GRCh38
rs73023948 Chr.1: 158292969-158292969 on Build GRCh38
rs767163637 Chr.1: 158292611-158292612 on Build GRCh38
rs111303778 Chr.1: 158292807-158292807 on Build GRCh38
rs548070401 Chr.1: 158292891-158292891 on Build GRCh38
rs529981470 Chr.1: 158292886-158292886 on Build GRCh38
rs770048157 Chr.1: 158292815-158292815 on Build GRCh38
rs3176969 Chr.1: 158292786-158292786 on Build GRCh38
rs980012146 Chr.1: 158292676-158292676 on Build GRCh38
rs763589486 Chr.1: 158292922-158292922 on Build GRCh38
rs200926914 Chr.1: 158292596-158292596 on Build GRCh38
rs770813634 Chr.1: 158292887-158292887 on Build GRCh38
rs139020586 Chr.1: 158292779-158292779 on Build GRCh38
rs751906751 Chr.1: 158292860-158292860 on Build GRCh38
rs961101351 Chr.1: 158292652-158292652 on Build GRCh38
rs901306034 Chr.1: 158292785-158292785 on Build GRCh38
rs768148459 Chr.1: 158292771-158292771 on Build GRCh38
rs773968611 Chr.1: 158292814-158292814 on Build GRCh38
rs746276859 Chr.1: 158292675-158292675 on Build GRCh38
rs536842285 Chr.1: 158292654-158292654 on Build GRCh38
rs765406937 Chr.1: 158292831-158292831 on Build GRCh38
rs757329481 Chr.1: 158292729-158292729 on Build GRCh38
rs756122506 Chr.1: 158292558-158292558 on Build GRCh38
rs775950742 Chr.1: 158292699-158292699 on Build GRCh38
rs1050646856 Chr.1: 158292929-158292929 on Build GRCh38
rs142848250 Chr.1: 158292942-158292942 on Build GRCh38
rs773196926 Chr.1: 158292788-158292788 on Build GRCh38
rs781278698 Chr.1: 158292550-158292550 on Build GRCh38
rs770723646 Chr.1: 158292811-158292811 on Build GRCh38
rs762703287 Chr.1: 158292625-158292625 on Build GRCh38
rs3176971 Chr.1: 158292799-158292799 on Build GRCh38
rs746301692 Chr.1: 158292800-158292800 on Build GRCh38
rs140255811 Chr.1: 158292588-158292588 on Build GRCh38
rs771329467 Chr.1: 158292586-158292586 on Build GRCh38
rs748262671 Chr.1: 158292551-158292551 on Build GRCh38
rs778428678 Chr.1: 158292923-158292923 on Build GRCh38
rs750954740 Chr.1: 158292743-158292743 on Build GRCh38
rs139069182 Chr.1: 158292833-158292833 on Build GRCh38
rs374551806 Chr.1: 158292959-158292959 on Build GRCh38
rs1029000417 Chr.1: 158292587-158292587 on Build GRCh38
rs778402029 Chr.1: 158292560-158292560 on Build GRCh38
rs779452323 Chr.1: 158292673-158292673 on Build GRCh38
rs144494205 Chr.1: 158292872-158292872 on Build GRCh38
rs768742456 Chr.1: 158292903-158292903 on Build GRCh38
rs969251812 Chr.1: 158292866-158292866 on Build GRCh38
rs760379040 Chr.1: 158292591-158292591 on Build GRCh38
rs772544606 Chr.1: 158292685-158292685 on Build GRCh38
rs772400542 Chr.1: 158292982-158292982 on Build GRCh38
rs375607782 Chr.1: 158292826-158292826 on Build GRCh38
rs760750400 Chr.1: 158292722-158292722 on Build GRCh38
rs754584129 Chr.1: 158292693-158292693 on Build GRCh38
rs147876610 Chr.1: 158292678-158292678 on Build GRCh38
rs982391642 Chr.1: 158292874-158292874 on Build GRCh38
rs913895196 Chr.1: 158292936-158292936 on Build GRCh38
rs115248892 Chr.1: 158292829-158292829 on Build GRCh38
rs370381988 Chr.1: 158292915-158292915 on Build GRCh38
rs776891906 Chr.1: 158292827-158292827 on Build GRCh38
rs1024484957 Chr.1: 158292868-158292868 on Build GRCh38
rs761837821 Chr.1: 158292700-158292700 on Build GRCh38
rs779613937 Chr.1: 158292734-158292734 on Build GRCh38
rs940800437 Chr.1: 158292894-158292894 on Build GRCh38
rs899159182 Chr.1: 158292758-158292758 on Build GRCh38
rs571642419 Chr.1: 158292541-158292541 on Build GRCh38
rs777648298 Chr.1: 158292656-158292656 on Build GRCh38
rs899463372 Chr.1: 158292538-158292538 on Build GRCh38

View Full Product Details