Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 101240730-101241233 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FIS1
Entrez Gene ID
51024
Gene Name
fission, mitochondrial 1
Gene Aliases
CGI-135, TTC11
UniGene
Hs.423968
Gene Chromosome Location
Chr.7: 101239612-101245090 on Build GRCh38
Transcripts
NM_016068.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4981723 c.247C>T p.R83W Chr.7: 101240838-101240838 on Build GRCh38
1739381 c.211C>T p.R71W Chr.7: 101240874-101240874 on Build GRCh38
6175367 c.206A>G p.E69G Chr.7: 101240879-101240879 on Build GRCh38
3631371 c.231G>T p.L77L Chr.7: 101240854-101240854 on Build GRCh38
7411435 c.245A>G p.Y82C Chr.7: 101240840-101240840 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs757592961 Chr.7: 101241107-101241107 on Build GRCh38
rs968227562 Chr.7: 101241042-101241042 on Build GRCh38
rs965550742 Chr.7: 101240810-101240810 on Build GRCh38
rs149215290 Chr.7: 101240791-101240791 on Build GRCh38
rs776112558 Chr.7: 101240787-101240787 on Build GRCh38
rs911142793 Chr.7: 101240780-101240780 on Build GRCh38
rs769627113 Chr.7: 101240958-101240958 on Build GRCh38
rs747831582 Chr.7: 101240858-101240858 on Build GRCh38
rs746492655 Chr.7: 101240783-101240783 on Build GRCh38
rs756116741 Chr.7: 101240821-101240821 on Build GRCh38
rs544751334 Chr.7: 101240851-101240851 on Build GRCh38
rs910854159 Chr.7: 101241177-101241177 on Build GRCh38
rs1049275559 Chr.7: 101240768-101240768 on Build GRCh38
rs775838689 Chr.7: 101240875-101240875 on Build GRCh38
rs930570100 Chr.7: 101241025-101241025 on Build GRCh38
rs542410129 Chr.7: 101240913-101240913 on Build GRCh38
rs769503978 Chr.7: 101240863-101240863 on Build GRCh38
rs986874168 Chr.7: 101241141-101241141 on Build GRCh38
rs572567105 Chr.7: 101241114-101241114 on Build GRCh38
rs774867698 Chr.7: 101240864-101240864 on Build GRCh38
rs754031660 Chr.7: 101240833-101240833 on Build GRCh38
rs142461818 Chr.7: 101240905-101240905 on Build GRCh38
rs184772721 Chr.7: 101241029-101241029 on Build GRCh38
rs774812654 Chr.7: 101240798-101240798 on Build GRCh38
rs1020515671 Chr.7: 101240814-101240814 on Build GRCh38
rs780569706 Chr.7: 101240950-101240950 on Build GRCh38
rs1040136154 Chr.7: 101241100-101241100 on Build GRCh38
rs747603109 Chr.7: 101240956-101240956 on Build GRCh38
rs967337681 Chr.7: 101240832-101240832 on Build GRCh38
rs768135871 Chr.7: 101240874-101240874 on Build GRCh38
rs146832792 Chr.7: 101240929-101240929 on Build GRCh38
rs917080940 Chr.7: 101240866-101240866 on Build GRCh38
rs764426375 Chr.7: 101240827-101240827 on Build GRCh38
rs752249823 Chr.7: 101240759-101240759 on Build GRCh38
rs368637918 Chr.7: 101240796-101240796 on Build GRCh38
rs757603851 Chr.7: 101240845-101240845 on Build GRCh38
rs577596491 Chr.7: 101240849-101240849 on Build GRCh38
rs768025810 Chr.7: 101240850-101240850 on Build GRCh38
rs903411118 Chr.7: 101241184-101241184 on Build GRCh38
rs550074612 Chr.7: 101241052-101241052 on Build GRCh38
rs1035001848 Chr.7: 101240887-101240887 on Build GRCh38
rs942352370 Chr.7: 101241203-101241203 on Build GRCh38
rs757432356 Chr.7: 101240836-101240836 on Build GRCh38
rs1012468824 Chr.7: 101240988-101240988 on Build GRCh38
rs751470497 Chr.7: 101240939-101240939 on Build GRCh38
rs752027618 Chr.7: 101240839-101240839 on Build GRCh38
rs371058423 Chr.7: 101240786-101240786 on Build GRCh38
rs888785117 Chr.7: 101241065-101241065 on Build GRCh38
rs758955458 Chr.7: 101240790-101240792 on Build GRCh38
rs761195778 Chr.7: 101240890-101240890 on Build GRCh38
rs754901533 Chr.7: 101240947-101240947 on Build GRCh38
rs559824763 Chr.7: 101240901-101240901 on Build GRCh38
rs998336345 Chr.7: 101241145-101241145 on Build GRCh38
rs778687437 Chr.7: 101240837-101240837 on Build GRCh38
rs943122649 Chr.7: 101241071-101241071 on Build GRCh38
rs374953576 Chr.7: 101240813-101240813 on Build GRCh38
rs200145052 Chr.7: 101240806-101240806 on Build GRCh38
rs781415072 Chr.7: 101240873-101240873 on Build GRCh38
rs867735279 Chr.7: 101240824-101240824 on Build GRCh38
rs775081392 Chr.7: 101240782-101240782 on Build GRCh38
rs752127401 Chr.7: 101240942-101240952 on Build GRCh38
rs746033784 Chr.7: 101241064-101241064 on Build GRCh38
rs760123325 Chr.7: 101240805-101240805 on Build GRCh38
rs745607674 Chr.7: 101240838-101240838 on Build GRCh38
rs764112759 Chr.7: 101240896-101240896 on Build GRCh38
rs868828787 Chr.7: 101241015-101241015 on Build GRCh38

View Full Product Details