Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 116499922-116500424 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CAV2
Entrez Gene ID
858
Gene Name
caveolin 2
Gene Aliases
CAV
UniGene
Hs.212332
Gene Chromosome Location
Chr.7: 116499601-116508541 on Build GRCh38
Transcripts
NM_001206747.1 NM_001206748.1 NM_001233.4 NM_198212.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6228449 c.173C>T p.A58V Chr.7: 116500282-116500282 on Build GRCh38
4006603 c.258G>A p.K86K Chr.7: 116500367-116500367 on Build GRCh38
3256883 c.189G>A p.T63T Chr.7: 116500298-116500298 on Build GRCh38
5958996 c.259delT p.F87fs*2 Chr.7: 116500368-116500368 on Build GRCh38
1496452 c.160G>A p.E54K Chr.7: 116500269-116500269 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs747078112 Chr.7: 116500303-116500303 on Build GRCh38
rs754623381 Chr.7: 116500392-116500392 on Build GRCh38
rs2535220 Chr.7: 116500022-116500022 on Build GRCh38
rs952452062 Chr.7: 116500230-116500230 on Build GRCh38
rs191930578 Chr.7: 116500049-116500049 on Build GRCh38
rs937773029 Chr.7: 116500085-116500085 on Build GRCh38
rs754748732 Chr.7: 116500288-116500288 on Build GRCh38
rs904634730 Chr.7: 116500063-116500063 on Build GRCh38
rs892257916 Chr.7: 116500100-116500100 on Build GRCh38
rs886367779 Chr.7: 116500187-116500187 on Build GRCh38
rs930299183 Chr.7: 116500171-116500171 on Build GRCh38
rs900369985 Chr.7: 116500256-116500256 on Build GRCh38
rs763238511 Chr.7: 116500348-116500348 on Build GRCh38
rs200069822 Chr.7: 116500298-116500298 on Build GRCh38
rs201434174 Chr.7: 116500388-116500388 on Build GRCh38
rs768506237 Chr.7: 116500214-116500214 on Build GRCh38
rs777056674 Chr.7: 116500237-116500237 on Build GRCh38
rs137971888 Chr.7: 116500394-116500394 on Build GRCh38
rs771929848 Chr.7: 116500224-116500224 on Build GRCh38
rs982234489 Chr.7: 116500064-116500064 on Build GRCh38
rs778273717 Chr.7: 116500287-116500287 on Build GRCh38
rs745398541 Chr.7: 116499947-116499947 on Build GRCh38
rs755681173 Chr.7: 116500275-116500275 on Build GRCh38
rs149320804 Chr.7: 116500365-116500365 on Build GRCh38
rs901243845 Chr.7: 116499996-116499996 on Build GRCh38
rs774126064 Chr.7: 116500219-116500219 on Build GRCh38
rs775854571 Chr.7: 116500335-116500335 on Build GRCh38
rs377393850 Chr.7: 116500034-116500034 on Build GRCh38
rs944310825 Chr.7: 116500016-116500016 on Build GRCh38
rs371624407 Chr.7: 116499964-116499964 on Build GRCh38
rs1001851074 Chr.7: 116500200-116500200 on Build GRCh38
rs13235183 Chr.7: 116500095-116500095 on Build GRCh38
rs201428661 Chr.7: 116500361-116500361 on Build GRCh38
rs144669871 Chr.7: 116499982-116499982 on Build GRCh38
rs752639459 Chr.7: 116500236-116500236 on Build GRCh38
rs199577791 Chr.7: 116500391-116500391 on Build GRCh38
rs200400870 Chr.7: 116500331-116500331 on Build GRCh38
rs921302855 Chr.7: 116500025-116500025 on Build GRCh38
rs765408285 Chr.7: 116500334-116500334 on Build GRCh38
rs866968027 Chr.7: 116499980-116499980 on Build GRCh38
rs760146851 Chr.7: 116500375-116500375 on Build GRCh38
rs771981557 Chr.7: 116500290-116500290 on Build GRCh38
rs908674864 Chr.7: 116499992-116499992 on Build GRCh38
rs760848661 Chr.7: 116500234-116500234 on Build GRCh38
rs758897137 Chr.7: 116500265-116500265 on Build GRCh38
rs375431787 Chr.7: 116500281-116500281 on Build GRCh38
rs757915346 Chr.7: 116500283-116500284 on Build GRCh38
rs773078932 Chr.7: 116500296-116500296 on Build GRCh38
rs764651052 Chr.7: 116500360-116500360 on Build GRCh38
rs764512649 Chr.7: 116500270-116500270 on Build GRCh38
rs535289894 Chr.7: 116500395-116500395 on Build GRCh38
rs200257358 Chr.7: 116500358-116500358 on Build GRCh38
rs765682808 Chr.7: 116500239-116500239 on Build GRCh38
rs143606406 Chr.7: 116500376-116500376 on Build GRCh38
rs11767363 Chr.7: 116500221-116500221 on Build GRCh38
rs201038703 Chr.7: 116500307-116500307 on Build GRCh38
rs202168833 Chr.7: 116500316-116500316 on Build GRCh38
rs934717558 Chr.7: 116500075-116500075 on Build GRCh38
rs201746184 Chr.7: 116500260-116500260 on Build GRCh38
rs533493704 Chr.7: 116500050-116500050 on Build GRCh38
rs753372066 Chr.7: 116500381-116500381 on Build GRCh38
rs755695447 Chr.7: 116499949-116499949 on Build GRCh38
rs368012050 Chr.7: 116500319-116500319 on Build GRCh38
rs867203582 Chr.7: 116500164-116500164 on Build GRCh38
rs752070410 Chr.7: 116500381-116500382 on Build GRCh38
rs201790924 Chr.7: 116500355-116500355 on Build GRCh38
rs202132850 Chr.7: 116500293-116500293 on Build GRCh38
rs771170610 Chr.7: 116500304-116500304 on Build GRCh38
rs749892853 Chr.7: 116500271-116500271 on Build GRCh38
rs1028320561 Chr.7: 116500031-116500031 on Build GRCh38
rs567885211 Chr.7: 116500276-116500276 on Build GRCh38
rs570074462 Chr.7: 116500192-116500192 on Build GRCh38
rs2535219 Chr.7: 116499945-116499945 on Build GRCh38
rs753025158 Chr.7: 116500254-116500254 on Build GRCh38
rs904886075 Chr.7: 116500186-116500186 on Build GRCh38
rs924186959 Chr.7: 116499979-116499979 on Build GRCh38
rs1013544572 Chr.7: 116500139-116500139 on Build GRCh38
rs989656405 Chr.7: 116500040-116500040 on Build GRCh38
rs201363499 Chr.7: 116500291-116500291 on Build GRCh38
rs199880412 Chr.7: 116500289-116500289 on Build GRCh38
rs549311062 Chr.7: 116500228-116500228 on Build GRCh38
rs200858684 Chr.7: 116500377-116500377 on Build GRCh38
rs1015765548 Chr.7: 116500363-116500363 on Build GRCh38
rs932642317 Chr.7: 116500028-116500028 on Build GRCh38
rs201185444 Chr.7: 116500344-116500344 on Build GRCh38
rs2535221 Chr.7: 116500030-116500030 on Build GRCh38
rs966822888 Chr.7: 116500057-116500057 on Build GRCh38

View Full Product Details