Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 122319442-122319940 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CADPS2
Entrez Gene ID
93664
Gene Name
calcium dependent secretion activator 2
Gene Aliases
CAPS2
UniGene
Hs.649459
Gene Chromosome Location
Chr.7: 122318424-122886759 on Build GRCh38
Transcripts
NM_001009571.3 NM_001167940.1 NM_017954.10 XM_005250695.4 XM_005250696.4 XM_005250697.4 XM_005250698.4 XM_005250699.4 XM_005250700.4 XM_005250701.4 XM_005250702.4 XM_005250703.4 XM_005250704.4 XM_005250705.4 XM_005250706.4 XM_005250707.4 XM_005250708.3 XM_011516695.2 XM_011516696.2 XM_011516697.2 XM_017012794.1 XM_017012795.1 XM_017012796.1 XM_017012797.1 XM_017012798.1 XM_017012799.1 XM_017012800.1

SNP Detail

SNP ID Chromosome Location
rs973442919 Chr.7: 122319897-122319897 on Build GRCh38
rs556568061 Chr.7: 122319760-122319760 on Build GRCh38
rs754816703 Chr.7: 122319797-122319797 on Build GRCh38
rs912442614 Chr.7: 122319752-122319752 on Build GRCh38
rs906738885 Chr.7: 122319862-122319863 on Build GRCh38
rs1039473684 Chr.7: 122319799-122319799 on Build GRCh38
rs1041646140 Chr.7: 122319488-122319488 on Build GRCh38
rs984998891 Chr.7: 122319747-122319747 on Build GRCh38
rs1012496774 Chr.7: 122319602-122319602 on Build GRCh38
rs970906872 Chr.7: 122319651-122319651 on Build GRCh38
rs1031976066 Chr.7: 122319548-122319548 on Build GRCh38
rs1000093748 Chr.7: 122319508-122319508 on Build GRCh38
rs1005142812 Chr.7: 122319863-122319863 on Build GRCh38
rs1050148224 Chr.7: 122319465-122319465 on Build GRCh38
rs777370441 Chr.7: 122319848-122319848 on Build GRCh38
rs754082634 Chr.7: 122319503-122319503 on Build GRCh38
rs79784430 Chr.7: 122319684-122319684 on Build GRCh38
rs564913110 Chr.7: 122319820-122319820 on Build GRCh38
rs983996090 Chr.7: 122319659-122319661 on Build GRCh38
rs985382665 Chr.7: 122319713-122319713 on Build GRCh38
rs760006410 Chr.7: 122319738-122319738 on Build GRCh38
rs933740810 Chr.7: 122319802-122319802 on Build GRCh38
rs775862433 Chr.7: 122319804-122319804 on Build GRCh38
rs902239604 Chr.7: 122319524-122319524 on Build GRCh38
rs757619413 Chr.7: 122319592-122319592 on Build GRCh38
rs765542344 Chr.7: 122319637-122319637 on Build GRCh38
rs1013311868 Chr.7: 122319915-122319915 on Build GRCh38
rs1024773781 Chr.7: 122319678-122319678 on Build GRCh38
rs867528565 Chr.7: 122319787-122319787 on Build GRCh38
rs12112854 Chr.7: 122319780-122319780 on Build GRCh38
rs936395848 Chr.7: 122319789-122319789 on Build GRCh38
rs1000149532 Chr.7: 122319859-122319859 on Build GRCh38
rs530485172 Chr.7: 122319863-122319863 on Build GRCh38
rs1033047923 Chr.7: 122319519-122319519 on Build GRCh38
rs117156132 Chr.7: 122319811-122319811 on Build GRCh38
rs758521372 Chr.7: 122319683-122319683 on Build GRCh38
rs958689340 Chr.7: 122319614-122319614 on Build GRCh38
rs1024656207 Chr.7: 122319841-122319841 on Build GRCh38
rs750581192 Chr.7: 122319642-122319642 on Build GRCh38
rs62476012 Chr.7: 122319804-122319804 on Build GRCh38
rs922313195 Chr.7: 122319800-122319800 on Build GRCh38
rs769478180 Chr.7: 122319694-122319694 on Build GRCh38
rs143616363 Chr.7: 122319624-122319624 on Build GRCh38
rs75643376 Chr.7: 122319877-122319877 on Build GRCh38
rs369879996 Chr.7: 122319782-122319782 on Build GRCh38
rs772829930 Chr.7: 122319698-122319703 on Build GRCh38
rs373661891 Chr.7: 122319890-122319890 on Build GRCh38
rs890945575 Chr.7: 122319472-122319472 on Build GRCh38
rs1000617361 Chr.7: 122319533-122319533 on Build GRCh38
rs1013012516 Chr.7: 122319618-122319618 on Build GRCh38
rs959059909 Chr.7: 122319598-122319598 on Build GRCh38
rs12706405 Chr.7: 122319692-122319692 on Build GRCh38

View Full Product Details