Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 151148869-151149368 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GBX1
Entrez Gene ID
2636
Gene Name
gastrulation brain homeobox 1
Gene Aliases
Huh-17
UniGene
Gene Chromosome Location
Chr.7: 151148009-151167548 on Build GRCh38
Transcripts
NM_001098834.2 XM_017011964.1 XM_017011965.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6480520 c.671G>A p.G224E Chr.7: 151149010-151149010 on Build GRCh38
4994067 c.743C>T p.A248V Chr.7: 151148938-151148938 on Build GRCh38
4686573 c.568_570delGAG p.E190delE Chr.7: 151149111-151149113 on Build GRCh38
5595814 c.612G>A p.Q204Q Chr.7: 151149069-151149069 on Build GRCh38
3261695 c.790C>T p.R264C Chr.7: 151148891-151148891 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1038526784 Chr.7: 151149006-151149006 on Build GRCh38
rs1006001851 Chr.7: 151149098-151149098 on Build GRCh38
rs779955416 Chr.7: 151149013-151149013 on Build GRCh38
rs201395169 Chr.7: 151148935-151148935 on Build GRCh38
rs747998051 Chr.7: 151149026-151149026 on Build GRCh38
rs778941621 Chr.7: 151149114-151149114 on Build GRCh38
rs746157537 Chr.7: 151149143-151149143 on Build GRCh38
rs1032541478 Chr.7: 151149219-151149219 on Build GRCh38
rs6970768 Chr.7: 151148971-151148971 on Build GRCh38
rs900131308 Chr.7: 151149108-151149108 on Build GRCh38
rs762209524 Chr.7: 151149056-151149056 on Build GRCh38
rs141040962 Chr.7: 151149235-151149235 on Build GRCh38
rs749252419 Chr.7: 151149014-151149014 on Build GRCh38
rs370970805 Chr.7: 151148976-151148976 on Build GRCh38
rs769215052 Chr.7: 151149050-151149050 on Build GRCh38
rs908566973 Chr.7: 151149263-151149263 on Build GRCh38
rs776802887 Chr.7: 151148925-151148925 on Build GRCh38
rs374261532 Chr.7: 151148923-151148923 on Build GRCh38
rs527931311 Chr.7: 151148909-151148909 on Build GRCh38
rs773480725 Chr.7: 151148913-151148913 on Build GRCh38
rs756008862 Chr.7: 151148894-151148894 on Build GRCh38
rs929121443 Chr.7: 151149066-151149066 on Build GRCh38
rs1024389823 Chr.7: 151149205-151149205 on Build GRCh38
rs776383074 Chr.7: 151149111-151149113 on Build GRCh38
rs375529208 Chr.7: 151148980-151148980 on Build GRCh38
rs982820185 Chr.7: 151148975-151148975 on Build GRCh38
rs577883610 Chr.7: 151149340-151149340 on Build GRCh38
rs11975799 Chr.7: 151149101-151149101 on Build GRCh38
rs777032330 Chr.7: 151149033-151149033 on Build GRCh38
rs749404097 Chr.7: 151149191-151149191 on Build GRCh38
rs775848180 Chr.7: 151149023-151149023 on Build GRCh38
rs373834136 Chr.7: 151149164-151149164 on Build GRCh38
rs745408260 Chr.7: 151148893-151148893 on Build GRCh38
rs755673022 Chr.7: 151149009-151149009 on Build GRCh38
rs747134618 Chr.7: 151148921-151148921 on Build GRCh38
rs536614779 Chr.7: 151149342-151149342 on Build GRCh38
rs767136931 Chr.7: 151149074-151149074 on Build GRCh38
rs974333084 Chr.7: 151149329-151149329 on Build GRCh38
rs971922640 Chr.7: 151149232-151149232 on Build GRCh38
rs771237910 Chr.7: 151149032-151149032 on Build GRCh38
rs570026897 Chr.7: 151149042-151149042 on Build GRCh38
rs941621919 Chr.7: 151148999-151148999 on Build GRCh38
rs887504545 Chr.7: 151149094-151149094 on Build GRCh38
rs770054570 Chr.7: 151149036-151149036 on Build GRCh38
rs1047593908 Chr.7: 151149086-151149086 on Build GRCh38
rs1000084655 Chr.7: 151149211-151149211 on Build GRCh38
rs749090563 Chr.7: 151148908-151148908 on Build GRCh38
rs770708910 Chr.7: 151148896-151148898 on Build GRCh38
rs990839729 Chr.7: 151149326-151149326 on Build GRCh38
rs759719860 Chr.7: 151148934-151148934 on Build GRCh38
rs200119934 Chr.7: 151148996-151148996 on Build GRCh38
rs535536993 Chr.7: 151149051-151149051 on Build GRCh38
rs749907803 Chr.7: 151149004-151149004 on Build GRCh38
rs765740892 Chr.7: 151149089-151149089 on Build GRCh38
rs757031253 Chr.7: 151149131-151149131 on Build GRCh38
rs900015489 Chr.7: 151149007-151149007 on Build GRCh38
rs367624740 Chr.7: 151149085-151149085 on Build GRCh38
rs753403517 Chr.7: 151149090-151149090 on Build GRCh38
rs770142451 Chr.7: 151149163-151149163 on Build GRCh38
rs950082266 Chr.7: 151148958-151148958 on Build GRCh38
rs1009182591 Chr.7: 151149071-151149071 on Build GRCh38
rs200333232 Chr.7: 151148890-151148890 on Build GRCh38
rs184506455 Chr.7: 151149328-151149328 on Build GRCh38
rs759084993 Chr.7: 151149190-151149191 on Build GRCh38
rs756840352 Chr.7: 151148991-151148991 on Build GRCh38
rs958212188 Chr.7: 151149320-151149321 on Build GRCh38
rs780010334 Chr.7: 151148896-151148896 on Build GRCh38
rs991944293 Chr.7: 151149195-151149195 on Build GRCh38
rs553461932 Chr.7: 151149215-151149215 on Build GRCh38
rs1000756906 Chr.7: 151149127-151149127 on Build GRCh38
rs564806375 Chr.7: 151148983-151148983 on Build GRCh38
rs751954730 Chr.7: 151148989-151148989 on Build GRCh38
rs192479027 Chr.7: 151149018-151149018 on Build GRCh38
rs548106872 Chr.7: 151148964-151148964 on Build GRCh38
rs745918218 Chr.7: 151149035-151149035 on Build GRCh38
rs557725971 Chr.7: 151149330-151149330 on Build GRCh38
rs959243329 Chr.7: 151149169-151149169 on Build GRCh38
rs865819643 Chr.7: 151148905-151148905 on Build GRCh38
rs982766280 Chr.7: 151149248-151149248 on Build GRCh38
rs781550382 Chr.7: 151149133-151149133 on Build GRCh38

View Full Product Details