Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 151167281-151167817 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GBX1
Entrez Gene ID
2636
Gene Name
gastrulation brain homeobox 1
Gene Aliases
Huh-17
UniGene
Gene Chromosome Location
Chr.7: 151148009-151167548 on Build GRCh38
Transcripts
NM_001098834.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6039532 c.47delG p.G16fs*15 Chr.7: 151167502-151167502 on Build GRCh38
7369297 c.244C>T p.R82C Chr.7: 151167305-151167305 on Build GRCh38
5002662 c.58G>C p.G20R Chr.7: 151167491-151167491 on Build GRCh38
1207987 c.253A>G p.N85D Chr.7: 151167296-151167296 on Build GRCh38
7254583 c.173C>A p.P58Q Chr.7: 151167376-151167376 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs558744648 Chr.7: 151167417-151167417 on Build GRCh38
rs942482235 Chr.7: 151167514-151167514 on Build GRCh38
rs941779280 Chr.7: 151167376-151167376 on Build GRCh38
rs903386203 Chr.7: 151167302-151167302 on Build GRCh38
rs942409810 Chr.7: 151167708-151167708 on Build GRCh38
rs1033193344 Chr.7: 151167605-151167605 on Build GRCh38
rs958722252 Chr.7: 151167326-151167326 on Build GRCh38
rs554826470 Chr.7: 151167575-151167575 on Build GRCh38
rs760449195 Chr.7: 151167569-151167569 on Build GRCh38
rs575510107 Chr.7: 151167426-151167426 on Build GRCh38
rs1025544744 Chr.7: 151167671-151167671 on Build GRCh38
rs951312041 Chr.7: 151167672-151167672 on Build GRCh38
rs981974576 Chr.7: 151167633-151167633 on Build GRCh38
rs776413830 Chr.7: 151167596-151167596 on Build GRCh38
rs779851693 Chr.7: 151167386-151167386 on Build GRCh38
rs867027174 Chr.7: 151167304-151167304 on Build GRCh38
rs1041981047 Chr.7: 151167793-151167793 on Build GRCh38
rs983822156 Chr.7: 151167468-151167468 on Build GRCh38
rs1013009208 Chr.7: 151167617-151167617 on Build GRCh38
rs887502195 Chr.7: 151167347-151167347 on Build GRCh38
rs778439051 Chr.7: 151167420-151167420 on Build GRCh38
rs770758251 Chr.7: 151167440-151167442 on Build GRCh38
rs13242341 Chr.7: 151167456-151167456 on Build GRCh38
rs909719688 Chr.7: 151167697-151167697 on Build GRCh38
rs971459264 Chr.7: 151167627-151167627 on Build GRCh38
rs929834883 Chr.7: 151167451-151167451 on Build GRCh38
rs748884945 Chr.7: 151167396-151167396 on Build GRCh38
rs574334722 Chr.7: 151167778-151167778 on Build GRCh38
rs1033121301 Chr.7: 151167325-151167331 on Build GRCh38
rs975225294 Chr.7: 151167713-151167713 on Build GRCh38
rs763102842 Chr.7: 151167597-151167597 on Build GRCh38
rs772014500 Chr.7: 151167479-151167479 on Build GRCh38
rs866752241 Chr.7: 151167726-151167726 on Build GRCh38
rs903335488 Chr.7: 151167557-151167557 on Build GRCh38
rs1042031820 Chr.7: 151167549-151167549 on Build GRCh38
rs894530357 Chr.7: 151167606-151167606 on Build GRCh38
rs983895743 Chr.7: 151167684-151167684 on Build GRCh38
rs1039738605 Chr.7: 151167521-151167521 on Build GRCh38
rs756608160 Chr.7: 151167382-151167382 on Build GRCh38
rs765397008 Chr.7: 151167314-151167314 on Build GRCh38
rs1024718059 Chr.7: 151167626-151167626 on Build GRCh38
rs991872346 Chr.7: 151167366-151167366 on Build GRCh38
rs922259243 Chr.7: 151167722-151167722 on Build GRCh38
rs544173525 Chr.7: 151167530-151167530 on Build GRCh38
rs971015580 Chr.7: 151167407-151167407 on Build GRCh38
rs1010495212 Chr.7: 151167501-151167501 on Build GRCh38

View Full Product Details