Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 158730808-158731308 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ESYT2
Entrez Gene ID
57488
Gene Name
extended synaptotagmin 2
Gene Aliases
CHR2SYT, E-Syt2, FAM62B
UniGene
Hs.490795
Gene Chromosome Location
Chr.7: 158730998-158829628 on Build GRCh38
Transcripts
NM_020728.2

SNP Detail

SNP ID Chromosome Location
rs770961581 Chr.7: 158730891-158730891 on Build GRCh38
rs979842080 Chr.7: 158730860-158730860 on Build GRCh38
rs964770298 Chr.7: 158731075-158731076 on Build GRCh38
rs535051624 Chr.7: 158731185-158731185 on Build GRCh38
rs1030840531 Chr.7: 158731094-158731094 on Build GRCh38
rs954923132 Chr.7: 158730975-158730975 on Build GRCh38
rs964802879 Chr.7: 158730879-158730879 on Build GRCh38
rs1053070266 Chr.7: 158730910-158730910 on Build GRCh38
rs902413740 Chr.7: 158731202-158731202 on Build GRCh38
rs555126588 Chr.7: 158730864-158730864 on Build GRCh38
rs557386842 Chr.7: 158731277-158731277 on Build GRCh38
rs1030416253 Chr.7: 158730964-158730964 on Build GRCh38
rs114181147 Chr.7: 158731097-158731097 on Build GRCh38
rs140114146 Chr.7: 158730945-158730945 on Build GRCh38
rs112439287 Chr.7: 158731147-158731147 on Build GRCh38
rs928361714 Chr.7: 158731108-158731108 on Build GRCh38
rs147502240 Chr.7: 158730922-158730922 on Build GRCh38
rs386720372 Chr.7: 158731197-158731198 on Build GRCh38
rs774662806 Chr.7: 158731012-158731012 on Build GRCh38
rs577321414 Chr.7: 158730885-158730885 on Build GRCh38
rs974862947 Chr.7: 158731075-158731075 on Build GRCh38
rs948480953 Chr.7: 158730880-158730880 on Build GRCh38
rs996294422 Chr.7: 158730962-158730962 on Build GRCh38
rs529757183 Chr.7: 158731050-158731050 on Build GRCh38
rs1030394123 Chr.7: 158731257-158731257 on Build GRCh38
rs73729952 Chr.7: 158730893-158730893 on Build GRCh38
rs900537077 Chr.7: 158731253-158731253 on Build GRCh38
rs537601969 Chr.7: 158731179-158731179 on Build GRCh38
rs773548930 Chr.7: 158731103-158731103 on Build GRCh38
rs868226373 Chr.7: 158731083-158731083 on Build GRCh38
rs938414581 Chr.7: 158731118-158731118 on Build GRCh38
rs1062119 Chr.7: 158731198-158731198 on Build GRCh38
rs1003102474 Chr.7: 158730897-158730897 on Build GRCh38
rs1044149015 Chr.7: 158731105-158731105 on Build GRCh38
rs149498337 Chr.7: 158731116-158731117 on Build GRCh38
rs984043593 Chr.7: 158731098-158731098 on Build GRCh38
rs1015517335 Chr.7: 158731254-158731254 on Build GRCh38
rs10242425 Chr.7: 158730875-158730875 on Build GRCh38
rs1038013870 Chr.7: 158730943-158730943 on Build GRCh38
rs879345551 Chr.7: 158731197-158731198 on Build GRCh38
rs745339276 Chr.7: 158730926-158730926 on Build GRCh38
rs1032339877 Chr.7: 158731221-158731221 on Build GRCh38
rs963570560 Chr.7: 158731273-158731273 on Build GRCh38
rs111421302 Chr.7: 158730923-158730923 on Build GRCh38
rs560685579 Chr.7: 158731018-158731018 on Build GRCh38
rs17837836 Chr.7: 158731146-158731146 on Build GRCh38
rs1054882807 Chr.7: 158731002-158731002 on Build GRCh38
rs758354319 Chr.7: 158731241-158731241 on Build GRCh38
rs908014371 Chr.7: 158731041-158731041 on Build GRCh38
rs752512084 Chr.7: 158731218-158731218 on Build GRCh38
rs142359623 Chr.7: 158730874-158730874 on Build GRCh38
rs548617528 Chr.7: 158731186-158731186 on Build GRCh38
rs17837835 Chr.7: 158731066-158731066 on Build GRCh38
rs923456464 Chr.7: 158730906-158730906 on Build GRCh38
rs769368234 Chr.7: 158730966-158730966 on Build GRCh38
rs2013 Chr.7: 158731197-158731197 on Build GRCh38
rs919628244 Chr.7: 158731088-158731088 on Build GRCh38
rs538500115 Chr.7: 158731272-158731272 on Build GRCh38
rs1032372508 Chr.7: 158731071-158731071 on Build GRCh38
rs896355169 Chr.7: 158731033-158731033 on Build GRCh38
rs532322482 Chr.7: 158731136-158731136 on Build GRCh38
rs954972267 Chr.7: 158731267-158731267 on Build GRCh38
rs73167232 Chr.7: 158730859-158730859 on Build GRCh38
rs1047942793 Chr.7: 158730916-158730916 on Build GRCh38
rs566052589 Chr.7: 158731054-158731054 on Build GRCh38

View Full Product Details