Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 158731971-158732470 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ESYT2
Entrez Gene ID
57488
Gene Name
extended synaptotagmin 2
Gene Aliases
CHR2SYT, E-Syt2, FAM62B
UniGene
Hs.490795
Gene Chromosome Location
Chr.7: 158730998-158829628 on Build GRCh38
Transcripts
NM_020728.2

SNP Detail

SNP ID Chromosome Location
rs971992242 Chr.7: 158732351-158732351 on Build GRCh38
rs953365786 Chr.7: 158732003-158732003 on Build GRCh38
rs3763408 Chr.7: 158732081-158732081 on Build GRCh38
rs1014432136 Chr.7: 158732389-158732389 on Build GRCh38
rs961988910 Chr.7: 158732034-158732034 on Build GRCh38
rs1028637028 Chr.7: 158732282-158732282 on Build GRCh38
rs3763412 Chr.7: 158732395-158732395 on Build GRCh38
rs3763411 Chr.7: 158732374-158732374 on Build GRCh38
rs188540295 Chr.7: 158732190-158732190 on Build GRCh38
rs113770251 Chr.7: 158732340-158732340 on Build GRCh38
rs777051393 Chr.7: 158732410-158732410 on Build GRCh38
rs987337052 Chr.7: 158732016-158732016 on Build GRCh38
rs1009026249 Chr.7: 158732311-158732311 on Build GRCh38
rs904120091 Chr.7: 158732448-158732448 on Build GRCh38
rs747530930 Chr.7: 158732391-158732391 on Build GRCh38
rs193238351 Chr.7: 158732439-158732439 on Build GRCh38
rs957803967 Chr.7: 158732135-158732135 on Build GRCh38
rs566015427 Chr.7: 158732027-158732027 on Build GRCh38
rs945519194 Chr.7: 158732426-158732426 on Build GRCh38
rs888891251 Chr.7: 158732288-158732288 on Build GRCh38
rs911692172 Chr.7: 158732018-158732018 on Build GRCh38
rs573074944 Chr.7: 158731992-158731992 on Build GRCh38
rs530956763 Chr.7: 158732393-158732393 on Build GRCh38
rs3763407 Chr.7: 158732080-158732080 on Build GRCh38
rs570724952 Chr.7: 158732400-158732400 on Build GRCh38
rs1056651508 Chr.7: 158732166-158732166 on Build GRCh38
rs779618908 Chr.7: 158732199-158732199 on Build GRCh38
rs1043998869 Chr.7: 158732248-158732248 on Build GRCh38
rs1028730544 Chr.7: 158731996-158731996 on Build GRCh38
rs574139573 Chr.7: 158732113-158732113 on Build GRCh38
rs569613867 Chr.7: 158732328-158732328 on Build GRCh38
rs543381983 Chr.7: 158732136-158732136 on Build GRCh38
rs558512643 Chr.7: 158732070-158732073 on Build GRCh38
rs904213207 Chr.7: 158732254-158732254 on Build GRCh38
rs3763406 Chr.7: 158732072-158732072 on Build GRCh38
rs545551186 Chr.7: 158732206-158732206 on Build GRCh38
rs3763410 Chr.7: 158732370-158732370 on Build GRCh38
rs931911226 Chr.7: 158732220-158732220 on Build GRCh38
rs3763409 Chr.7: 158732139-158732139 on Build GRCh38
rs999852760 Chr.7: 158732255-158732255 on Build GRCh38
rs914114864 Chr.7: 158732416-158732416 on Build GRCh38
rs930751018 Chr.7: 158732365-158732365 on Build GRCh38

View Full Product Details