Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 158732312-158732840 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ESYT2
Entrez Gene ID
57488
Gene Name
extended synaptotagmin 2
Gene Aliases
CHR2SYT, E-Syt2, FAM62B
UniGene
Hs.490795
Gene Chromosome Location
Chr.7: 158730998-158829628 on Build GRCh38
Transcripts
NM_020728.2

SNP Detail

SNP ID Chromosome Location
rs971992242 Chr.7: 158732351-158732351 on Build GRCh38
rs1008678845 Chr.7: 158732533-158732533 on Build GRCh38
rs1014432136 Chr.7: 158732389-158732389 on Build GRCh38
rs950234426 Chr.7: 158732688-158732688 on Build GRCh38
rs3763411 Chr.7: 158732374-158732374 on Build GRCh38
rs777051393 Chr.7: 158732410-158732410 on Build GRCh38
rs538119492 Chr.7: 158732676-158732676 on Build GRCh38
rs1002036768 Chr.7: 158732649-158732649 on Build GRCh38
rs904120091 Chr.7: 158732448-158732448 on Build GRCh38
rs992280264 Chr.7: 158732682-158732682 on Build GRCh38
rs555665923 Chr.7: 158732508-158732508 on Build GRCh38
rs546733302 Chr.7: 158732456-158732456 on Build GRCh38
rs193238351 Chr.7: 158732439-158732439 on Build GRCh38
rs945519194 Chr.7: 158732426-158732426 on Build GRCh38
rs893443834 Chr.7: 158732732-158732732 on Build GRCh38
rs554254779 Chr.7: 158732485-158732485 on Build GRCh38
rs980571267 Chr.7: 158732679-158732679 on Build GRCh38
rs867766632 Chr.7: 158732466-158732468 on Build GRCh38
rs780349738 Chr.7: 158732722-158732722 on Build GRCh38
rs558362830 Chr.7: 158732675-158732675 on Build GRCh38
rs920634305 Chr.7: 158732656-158732656 on Build GRCh38
rs745968033 Chr.7: 158732637-158732637 on Build GRCh38
rs925404303 Chr.7: 158732730-158732730 on Build GRCh38
rs1050092740 Chr.7: 158732488-158732488 on Build GRCh38
rs951981838 Chr.7: 158732639-158732639 on Build GRCh38
rs111433171 Chr.7: 158732469-158732471 on Build GRCh38
rs3763410 Chr.7: 158732370-158732370 on Build GRCh38
rs888744273 Chr.7: 158732524-158732524 on Build GRCh38
rs566735346 Chr.7: 158732474-158732474 on Build GRCh38
rs914114864 Chr.7: 158732416-158732416 on Build GRCh38
rs1030666603 Chr.7: 158732731-158732731 on Build GRCh38
rs3763412 Chr.7: 158732395-158732395 on Build GRCh38
rs115737267 Chr.7: 158732687-158732687 on Build GRCh38
rs113770251 Chr.7: 158732340-158732340 on Build GRCh38
rs758355711 Chr.7: 158732683-158732683 on Build GRCh38
rs747530930 Chr.7: 158732391-158732391 on Build GRCh38
rs1050421405 Chr.7: 158732813-158732813 on Build GRCh38
rs1010582031 Chr.7: 158732738-158732738 on Build GRCh38
rs953167546 Chr.7: 158732798-158732798 on Build GRCh38
rs897679199 Chr.7: 158732568-158732568 on Build GRCh38
rs966758246 Chr.7: 158732708-158732708 on Build GRCh38
rs530956763 Chr.7: 158732393-158732393 on Build GRCh38
rs1061620 Chr.7: 158732475-158732475 on Build GRCh38
rs1027415812 Chr.7: 158732607-158732607 on Build GRCh38
rs570724952 Chr.7: 158732400-158732400 on Build GRCh38
rs545892074 Chr.7: 158732715-158732715 on Build GRCh38
rs999156568 Chr.7: 158732725-158732725 on Build GRCh38
rs113067377 Chr.7: 158732600-158732600 on Build GRCh38
rs1019105094 Chr.7: 158732556-158732556 on Build GRCh38
rs1045928660 Chr.7: 158732703-158732706 on Build GRCh38
rs987342098 Chr.7: 158732564-158732564 on Build GRCh38
rs576716507 Chr.7: 158732712-158732712 on Build GRCh38
rs148842569 Chr.7: 158732750-158732750 on Build GRCh38
rs930751018 Chr.7: 158732365-158732365 on Build GRCh38

View Full Product Details