Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 160996882-160997386 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
F11R
Entrez Gene ID
50848
Gene Name
F11 receptor
Gene Aliases
CD321, JAM, JAM1, JAMA, JCAM, KAT, PAM-1
UniGene
Hs.517293
Gene Chromosome Location
Chr.1: 160995211-161021343 on Build GRCh38
Transcripts
NM_016946.4

SNP Detail

SNP ID Chromosome Location
rs537407070 Chr.1: 160997084-160997084 on Build GRCh38
rs1010386877 Chr.1: 160997120-160997120 on Build GRCh38
rs748153041 Chr.1: 160997198-160997198 on Build GRCh38
rs962267730 Chr.1: 160997353-160997353 on Build GRCh38
rs41267953 Chr.1: 160996980-160996980 on Build GRCh38
rs190154202 Chr.1: 160997022-160997022 on Build GRCh38
rs557244536 Chr.1: 160996968-160996968 on Build GRCh38
rs543247963 Chr.1: 160997185-160997185 on Build GRCh38
rs1062827 Chr.1: 160997054-160997054 on Build GRCh38
rs917899528 Chr.1: 160997002-160997002 on Build GRCh38
rs905742078 Chr.1: 160997042-160997042 on Build GRCh38
rs746754627 Chr.1: 160997058-160997058 on Build GRCh38
rs11542268 Chr.1: 160997325-160997325 on Build GRCh38
rs12759428 Chr.1: 160997013-160997013 on Build GRCh38
rs193135565 Chr.1: 160997174-160997174 on Build GRCh38
rs980904250 Chr.1: 160996994-160996994 on Build GRCh38
rs992634993 Chr.1: 160997365-160997365 on Build GRCh38
rs952258531 Chr.1: 160997230-160997230 on Build GRCh38
rs1045101055 Chr.1: 160997028-160997028 on Build GRCh38
rs940770881 Chr.1: 160996990-160996990 on Build GRCh38
rs928169758 Chr.1: 160996996-160996996 on Build GRCh38
rs578076976 Chr.1: 160997078-160997078 on Build GRCh38
rs749952837 Chr.1: 160996906-160996906 on Build GRCh38
rs915368999 Chr.1: 160996962-160996962 on Build GRCh38
rs140186836 Chr.1: 160997244-160997244 on Build GRCh38
rs935903027 Chr.1: 160997044-160997044 on Build GRCh38
rs998491494 Chr.1: 160997334-160997334 on Build GRCh38
rs566003128 Chr.1: 160996978-160996978 on Build GRCh38
rs989557071 Chr.1: 160996942-160996942 on Build GRCh38
rs1044478 Chr.1: 160997173-160997173 on Build GRCh38
rs1048127755 Chr.1: 160997143-160997143 on Build GRCh38
rs1036642354 Chr.1: 160997027-160997027 on Build GRCh38
rs185746191 Chr.1: 160997344-160997344 on Build GRCh38
rs888155103 Chr.1: 160997153-160997153 on Build GRCh38
rs901024521 Chr.1: 160997214-160997214 on Build GRCh38

View Full Product Details