Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 161218551-161219048 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FCER1G
Entrez Gene ID
2207
Gene Name
Fc fragment of IgE receptor Ig
Gene Aliases
FCRG
UniGene
Hs.433300
Gene Chromosome Location
Chr.1: 161215297-161219248 on Build GRCh38
Transcripts
NM_004106.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5442055 c.216C>G p.N72K Chr.1: 161218898-161218898 on Build GRCh38
6997443 c.235_236delCT p.L79fs*3 Chr.1: 161218917-161218918 on Build GRCh38
7235169 c.199-4C>G p.? Chr.1: 161218877-161218877 on Build GRCh38
5620804 c.188G>A p.G63D Chr.1: 161218713-161218713 on Build GRCh38
7275560 c.224C>T p.T75I Chr.1: 161218906-161218906 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs764590075 Chr.1: 161218992-161218992 on Build GRCh38
rs1018104023 Chr.1: 161218858-161218858 on Build GRCh38
rs944396497 Chr.1: 161218812-161218812 on Build GRCh38
rs956815460 Chr.1: 161219020-161219020 on Build GRCh38
rs985168949 Chr.1: 161218833-161218833 on Build GRCh38
rs765791571 Chr.1: 161218801-161218801 on Build GRCh38
rs375133827 Chr.1: 161218765-161218765 on Build GRCh38
rs753109167 Chr.1: 161218713-161218713 on Build GRCh38
rs111662633 Chr.1: 161218692-161218692 on Build GRCh38
rs911043061 Chr.1: 161218618-161218618 on Build GRCh38
rs943992874 Chr.1: 161218645-161218645 on Build GRCh38
rs1039291063 Chr.1: 161218623-161218623 on Build GRCh38
rs41270849 Chr.1: 161218859-161218859 on Build GRCh38
rs764345067 Chr.1: 161218870-161218870 on Build GRCh38
rs1027051297 Chr.1: 161218817-161218817 on Build GRCh38
rs773074207 Chr.1: 161218967-161218967 on Build GRCh38
rs756543032 Chr.1: 161218868-161218868 on Build GRCh38
rs565212327 Chr.1: 161218678-161218678 on Build GRCh38
rs776667723 Chr.1: 161218766-161218766 on Build GRCh38
rs569178245 Chr.1: 161218811-161218811 on Build GRCh38
rs972777606 Chr.1: 161218579-161218579 on Build GRCh38
rs778377623 Chr.1: 161218723-161218723 on Build GRCh38
rs746161856 Chr.1: 161218753-161218753 on Build GRCh38
rs923824989 Chr.1: 161218687-161218687 on Build GRCh38
rs749782068 Chr.1: 161218727-161218727 on Build GRCh38
rs772430805 Chr.1: 161218754-161218754 on Build GRCh38
rs761748391 Chr.1: 161218964-161218966 on Build GRCh38
rs544235134 Chr.1: 161218807-161218807 on Build GRCh38
rs993868847 Chr.1: 161218809-161218809 on Build GRCh38
rs760539502 Chr.1: 161218764-161218764 on Build GRCh38
rs377661089 Chr.1: 161218616-161218616 on Build GRCh38
rs372658412 Chr.1: 161218866-161218866 on Build GRCh38
rs764102612 Chr.1: 161218685-161218685 on Build GRCh38
rs764784767 Chr.1: 161218831-161218831 on Build GRCh38
rs113730786 Chr.1: 161218877-161218877 on Build GRCh38
rs557760626 Chr.1: 161218800-161218800 on Build GRCh38
rs952547635 Chr.1: 161218589-161218589 on Build GRCh38
rs372128651 Chr.1: 161218758-161218758 on Build GRCh38
rs1035037466 Chr.1: 161218749-161218749 on Build GRCh38
rs779763603 Chr.1: 161218802-161218802 on Build GRCh38
rs750123263 Chr.1: 161218700-161218700 on Build GRCh38
rs367852713 Chr.1: 161218697-161218697 on Build GRCh38
rs867064007 Chr.1: 161218782-161218782 on Build GRCh38
rs777095733 Chr.1: 161218832-161218832 on Build GRCh38
rs758357837 Chr.1: 161218922-161218922 on Build GRCh38
rs544886416 Chr.1: 161218615-161218615 on Build GRCh38
rs757838295 Chr.1: 161218890-161218890 on Build GRCh38
rs985336717 Chr.1: 161218603-161218603 on Build GRCh38
rs769725948 Chr.1: 161218959-161218959 on Build GRCh38
rs759099014 Chr.1: 161218982-161218982 on Build GRCh38
rs768542265 Chr.1: 161218940-161218940 on Build GRCh38
rs923898539 Chr.1: 161218997-161218997 on Build GRCh38
rs774103027 Chr.1: 161218976-161218976 on Build GRCh38
rs761716383 Chr.1: 161218767-161218767 on Build GRCh38
rs758031936 Chr.1: 161218704-161218704 on Build GRCh38
rs780647823 Chr.1: 161218969-161218969 on Build GRCh38
rs970379825 Chr.1: 161218796-161218796 on Build GRCh38
rs780351889 Chr.1: 161218928-161218928 on Build GRCh38
rs748150116 Chr.1: 161218958-161218958 on Build GRCh38
rs776397418 Chr.1: 161218948-161218948 on Build GRCh38
rs765777454 Chr.1: 161218974-161218974 on Build GRCh38
rs756386336 Chr.1: 161218721-161218721 on Build GRCh38
rs781474375 Chr.1: 161218711-161218711 on Build GRCh38
rs779497508 Chr.1: 161218898-161218898 on Build GRCh38
rs930870959 Chr.1: 161218882-161218882 on Build GRCh38
rs775353592 Chr.1: 161218665-161218665 on Build GRCh38
rs532181835 Chr.1: 161218783-161218783 on Build GRCh38
rs776819597 Chr.1: 161218925-161218926 on Build GRCh38
rs745961564 Chr.1: 161218910-161218910 on Build GRCh38
rs1056475247 Chr.1: 161218981-161218981 on Build GRCh38
rs754457378 Chr.1: 161218881-161218881 on Build GRCh38
rs771188761 Chr.1: 161218732-161218732 on Build GRCh38
rs988578847 Chr.1: 161218806-161218807 on Build GRCh38
rs762381466 Chr.1: 161218857-161218857 on Build GRCh38
rs779126504 Chr.1: 161218744-161218744 on Build GRCh38
rs373951783 Chr.1: 161218781-161218781 on Build GRCh38
rs911574042 Chr.1: 161218590-161218590 on Build GRCh38
rs976636202 Chr.1: 161218660-161218660 on Build GRCh38
rs761340322 Chr.1: 161218694-161218694 on Build GRCh38
rs1026749607 Chr.1: 161218585-161218585 on Build GRCh38
rs11590967 Chr.1: 161218797-161218797 on Build GRCh38
rs878909701 Chr.1: 161218925-161218925 on Build GRCh38
rs747165089 Chr.1: 161218937-161218937 on Build GRCh38

View Full Product Details