Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 162855251-162855766 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC190
Entrez Gene ID
339512
Gene Name
coiled-coil domain containing 190
Gene Aliases
C1orf110
UniGene
Hs.407631
Gene Chromosome Location
Chr.1: 162851291-162868918 on Build GRCh38
Transcripts
NM_178550.4 XM_005245125.3 XM_006711289.3 XM_011509477.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4224598 c.299G>T p.R100I Chr.1: 162855632-162855632 on Build GRCh38
6122458 c.333G>T p.M111I Chr.1: 162855341-162855341 on Build GRCh38
3750540 c.264G>T p.R88S Chr.1: 162855667-162855667 on Build GRCh38
397074 c.215C>G p.S72C Chr.1: 162855731-162855731 on Build GRCh38
5963702 c.312G>C p.M104I Chr.1: 162855634-162855634 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs756472091 Chr.1: 162855373-162855373 on Build GRCh38
rs764690427 Chr.1: 162855352-162855352 on Build GRCh38
rs953355511 Chr.1: 162855542-162855542 on Build GRCh38
rs747998813 Chr.1: 162855688-162855688 on Build GRCh38
rs749944437 Chr.1: 162855355-162855355 on Build GRCh38
rs906272776 Chr.1: 162855471-162855471 on Build GRCh38
rs758458325 Chr.1: 162855663-162855663 on Build GRCh38
rs536438255 Chr.1: 162855504-162855504 on Build GRCh38
rs572837238 Chr.1: 162855608-162855608 on Build GRCh38
rs759039651 Chr.1: 162855614-162855615 on Build GRCh38
rs772131375 Chr.1: 162855590-162855590 on Build GRCh38
rs528220353 Chr.1: 162855372-162855372 on Build GRCh38
rs766885894 Chr.1: 162855714-162855714 on Build GRCh38
rs956552962 Chr.1: 162855442-162855442 on Build GRCh38
rs149806280 Chr.1: 162855390-162855390 on Build GRCh38
rs768289439 Chr.1: 162855602-162855602 on Build GRCh38
rs754951245 Chr.1: 162855678-162855678 on Build GRCh38
rs775343219 Chr.1: 162855292-162855292 on Build GRCh38
rs975015642 Chr.1: 162855546-162855546 on Build GRCh38
rs915474849 Chr.1: 162855706-162855706 on Build GRCh38
rs766995543 Chr.1: 162855695-162855697 on Build GRCh38
rs377737928 Chr.1: 162855726-162855726 on Build GRCh38
rs773940880 Chr.1: 162855704-162855704 on Build GRCh38
rs1012214022 Chr.1: 162855375-162855375 on Build GRCh38
rs757582779 Chr.1: 162855397-162855397 on Build GRCh38
rs762512371 Chr.1: 162855619-162855619 on Build GRCh38
rs776523013 Chr.1: 162855604-162855604 on Build GRCh38
rs536907515 Chr.1: 162855722-162855722 on Build GRCh38
rs531085956 Chr.1: 162855410-162855410 on Build GRCh38
rs139941480 Chr.1: 162855700-162855700 on Build GRCh38
rs780807127 Chr.1: 162855488-162855488 on Build GRCh38
rs77227460 Chr.1: 162855667-162855667 on Build GRCh38
rs1030889671 Chr.1: 162855718-162855718 on Build GRCh38
rs767962605 Chr.1: 162855371-162855371 on Build GRCh38
rs372327417 Chr.1: 162855313-162855313 on Build GRCh38
rs965441996 Chr.1: 162855612-162855612 on Build GRCh38
rs756039250 Chr.1: 162855730-162855731 on Build GRCh38
rs772887259 Chr.1: 162855693-162855693 on Build GRCh38
rs369304295 Chr.1: 162855639-162855639 on Build GRCh38
rs764643155 Chr.1: 162855731-162855733 on Build GRCh38
rs553198862 Chr.1: 162855541-162855541 on Build GRCh38
rs759983081 Chr.1: 162855361-162855361 on Build GRCh38
rs375996919 Chr.1: 162855618-162855618 on Build GRCh38
rs752982716 Chr.1: 162855514-162855514 on Build GRCh38
rs746883277 Chr.1: 162855673-162855673 on Build GRCh38
rs766486154 Chr.1: 162855297-162855297 on Build GRCh38
rs147909245 Chr.1: 162855611-162855611 on Build GRCh38
rs896536006 Chr.1: 162855649-162855650 on Build GRCh38
rs1014639412 Chr.1: 162855495-162855495 on Build GRCh38
rs1045127574 Chr.1: 162855658-162855658 on Build GRCh38
rs745760397 Chr.1: 162855286-162855286 on Build GRCh38
rs199928833 Chr.1: 162855382-162855382 on Build GRCh38
rs753190588 Chr.1: 162855621-162855621 on Build GRCh38
rs765622936 Chr.1: 162855656-162855656 on Build GRCh38
rs769453631 Chr.1: 162855616-162855616 on Build GRCh38
rs768033654 Chr.1: 162855620-162855620 on Build GRCh38
rs368605278 Chr.1: 162855301-162855301 on Build GRCh38
rs765534003 Chr.1: 162855728-162855728 on Build GRCh38
rs373634463 Chr.1: 162855646-162855646 on Build GRCh38
rs894063644 Chr.1: 162855439-162855439 on Build GRCh38
rs566605845 Chr.1: 162855560-162855560 on Build GRCh38
rs772139595 Chr.1: 162855288-162855288 on Build GRCh38
rs754258885 Chr.1: 162855648-162855648 on Build GRCh38
rs368861148 Chr.1: 162855586-162855586 on Build GRCh38
rs1003274337 Chr.1: 162855481-162855481 on Build GRCh38
rs746798567 Chr.1: 162855601-162855601 on Build GRCh38
rs779154974 Chr.1: 162855277-162855277 on Build GRCh38
rs780910438 Chr.1: 162855685-162855685 on Build GRCh38
rs769427289 Chr.1: 162855692-162855692 on Build GRCh38
rs748771375 Chr.1: 162855694-162855694 on Build GRCh38
rs750622469 Chr.1: 162855737-162855737 on Build GRCh38
rs1049729902 Chr.1: 162855275-162855275 on Build GRCh38
rs377556745 Chr.1: 162855650-162855650 on Build GRCh38
rs779068988 Chr.1: 162855406-162855406 on Build GRCh38
rs761478107 Chr.1: 162855345-162855345 on Build GRCh38
rs12036026 Chr.1: 162855468-162855468 on Build GRCh38
rs1035099655 Chr.1: 162855364-162855364 on Build GRCh38
rs371455711 Chr.1: 162855597-162855597 on Build GRCh38
rs979338024 Chr.1: 162855419-162855419 on Build GRCh38
rs551262154 Chr.1: 162855454-162855454 on Build GRCh38
rs761018736 Chr.1: 162855709-162855709 on Build GRCh38
rs776197002 Chr.1: 162855315-162855315 on Build GRCh38
rs976901171 Chr.1: 162855346-162855346 on Build GRCh38
rs752668355 Chr.1: 162855729-162855731 on Build GRCh38
rs868774458 Chr.1: 162855574-162855574 on Build GRCh38

View Full Product Details