Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 86467430-86467967 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RMDN1
Entrez Gene ID
51115
Gene Name
regulator of microtubule dynamics 1
Gene Aliases
CGI-90, FAM82B, RMD-1, RMD1
UniGene
Hs.145386
Gene Chromosome Location
Chr.8: 86467116-86514348 on Build GRCh38
Transcripts
XM_011517090.2 XM_017013518.1 XM_017013525.1 NM_007013.3 XM_005250760.3 XM_005250761.3 XM_011516802.2 XM_017012993.1 XM_017012994.1 XM_017012995.1 XM_017012996.1 XM_017012997.1

Gene Symbol
WWP1
Entrez Gene ID
11059
Gene Name
WW domain containing E3 ubiquitin protein ligase 1
Gene Aliases
AIP5, Tiul1, hSDRP1
UniGene
Hs.655189
Gene Chromosome Location
Chr.8: 86342711-86468357 on Build GRCh38
Transcripts
XM_011517090.2 XM_017013518.1 XM_017013525.1 NM_007013.3 XM_005250760.3 XM_005250761.3 XM_011516802.2 XM_017012993.1 XM_017012994.1 XM_017012995.1 XM_017012996.1 XM_017012997.1

SNP Detail

SNP ID Chromosome Location
rs970869304 Chr.8: 86467615-86467615 on Build GRCh38
rs765804394 Chr.8: 86467475-86467475 on Build GRCh38
rs8635 Chr.8: 86467581-86467581 on Build GRCh38
rs577277867 Chr.8: 86467857-86467857 on Build GRCh38
rs116580489 Chr.8: 86467618-86467618 on Build GRCh38
rs564549750 Chr.8: 86467637-86467637 on Build GRCh38
rs538968619 Chr.8: 86467524-86467524 on Build GRCh38
rs570067572 Chr.8: 86467479-86467479 on Build GRCh38
rs903050812 Chr.8: 86467753-86467753 on Build GRCh38
rs996833145 Chr.8: 86467554-86467554 on Build GRCh38
rs143113633 Chr.8: 86467776-86467776 on Build GRCh38
rs1008187569 Chr.8: 86467476-86467476 on Build GRCh38
rs56328568 Chr.8: 86467856-86467856 on Build GRCh38
rs772429373 Chr.8: 86467808-86467808 on Build GRCh38
rs947117320 Chr.8: 86467741-86467741 on Build GRCh38
rs1033905262 Chr.8: 86467892-86467892 on Build GRCh38
rs943840041 Chr.8: 86467774-86467774 on Build GRCh38
rs558951181 Chr.8: 86467556-86467556 on Build GRCh38
rs991141089 Chr.8: 86467720-86467720 on Build GRCh38
rs1017249099 Chr.8: 86467801-86467801 on Build GRCh38
rs3087641 Chr.8: 86467868-86467868 on Build GRCh38
rs1041854935 Chr.8: 86467746-86467746 on Build GRCh38
rs1004342424 Chr.8: 86467830-86467830 on Build GRCh38
rs991563887 Chr.8: 86467911-86467911 on Build GRCh38
rs528527309 Chr.8: 86467650-86467650 on Build GRCh38
rs888533741 Chr.8: 86467822-86467822 on Build GRCh38
rs371121779 Chr.8: 86467736-86467736 on Build GRCh38
rs937024498 Chr.8: 86467671-86467671 on Build GRCh38
rs540688009 Chr.8: 86467777-86467777 on Build GRCh38
rs191645316 Chr.8: 86467792-86467792 on Build GRCh38
rs959581364 Chr.8: 86467910-86467910 on Build GRCh38
rs184184991 Chr.8: 86467468-86467468 on Build GRCh38
rs372493959 Chr.8: 86467603-86467603 on Build GRCh38
rs983365216 Chr.8: 86467582-86467582 on Build GRCh38
rs188738859 Chr.8: 86467562-86467562 on Build GRCh38
rs748508218 Chr.8: 86467740-86467740 on Build GRCh38
rs914282354 Chr.8: 86467912-86467912 on Build GRCh38
rs1019535195 Chr.8: 86467485-86467486 on Build GRCh38
rs1016079140 Chr.8: 86467846-86467846 on Build GRCh38
rs562621499 Chr.8: 86467888-86467888 on Build GRCh38
rs1026954543 Chr.8: 86467802-86467802 on Build GRCh38
rs926856784 Chr.8: 86467641-86467644 on Build GRCh38

View Full Product Details