Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 171210116-171210667 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FMO2
Entrez Gene ID
2327
Gene Name
flavin containing monooxygenase 2
Gene Aliases
FMO1B1
UniGene
Hs.144912
Gene Chromosome Location
Chr.1: 171185208-171212683 on Build GRCh38
Transcripts
NM_001301347.1 NM_001460.4

SNP Detail

SNP ID Chromosome Location
rs888842140 Chr.1: 171210525-171210525 on Build GRCh38
rs181926124 Chr.1: 171210641-171210641 on Build GRCh38
rs1019949912 Chr.1: 171210433-171210433 on Build GRCh38
rs561882861 Chr.1: 171210169-171210174 on Build GRCh38
rs1036390783 Chr.1: 171210292-171210292 on Build GRCh38
rs28369918 Chr.1: 171210531-171210531 on Build GRCh38
rs888929090 Chr.1: 171210283-171210283 on Build GRCh38
rs189882105 Chr.1: 171210443-171210443 on Build GRCh38
rs981917111 Chr.1: 171210364-171210364 on Build GRCh38
rs1035417157 Chr.1: 171210398-171210398 on Build GRCh38
rs544659974 Chr.1: 171210172-171210172 on Build GRCh38
rs568994433 Chr.1: 171210504-171210504 on Build GRCh38
rs911870207 Chr.1: 171210248-171210248 on Build GRCh38
rs186886674 Chr.1: 171210441-171210441 on Build GRCh38
rs141674151 Chr.1: 171210332-171210332 on Build GRCh38
rs781565523 Chr.1: 171210319-171210319 on Build GRCh38
rs532398948 Chr.1: 171210377-171210377 on Build GRCh38
rs7515157 Chr.1: 171210338-171210338 on Build GRCh38
rs1028788469 Chr.1: 171210456-171210456 on Build GRCh38
rs997668328 Chr.1: 171210252-171210252 on Build GRCh38
rs968601012 Chr.1: 171210446-171210446 on Build GRCh38
rs7512785 Chr.1: 171210148-171210148 on Build GRCh38
rs959142339 Chr.1: 171210402-171210402 on Build GRCh38
rs920627666 Chr.1: 171210611-171210611 on Build GRCh38
rs895599121 Chr.1: 171210409-171210409 on Build GRCh38
rs748531480 Chr.1: 171210331-171210333 on Build GRCh38
rs569488694 Chr.1: 171210543-171210543 on Build GRCh38
rs12023185 Chr.1: 171210527-171210527 on Build GRCh38
rs905365372 Chr.1: 171210349-171210349 on Build GRCh38
rs1009839421 Chr.1: 171210420-171210420 on Build GRCh38
rs1009478108 Chr.1: 171210560-171210560 on Build GRCh38
rs1000680585 Chr.1: 171210286-171210286 on Build GRCh38
rs984890846 Chr.1: 171210405-171210405 on Build GRCh38
rs1020322919 Chr.1: 171210573-171210573 on Build GRCh38
rs76011618 Chr.1: 171210506-171210506 on Build GRCh38
rs181865915 Chr.1: 171210256-171210256 on Build GRCh38
rs899396767 Chr.1: 171210295-171210295 on Build GRCh38
rs73032526 Chr.1: 171210640-171210640 on Build GRCh38
rs971824645 Chr.1: 171210619-171210619 on Build GRCh38
rs979096239 Chr.1: 171210447-171210447 on Build GRCh38
rs778731871 Chr.1: 171210509-171210509 on Build GRCh38
rs1001268975 Chr.1: 171210363-171210363 on Build GRCh38
rs940944872 Chr.1: 171210431-171210431 on Build GRCh38
rs16864194 Chr.1: 171210212-171210212 on Build GRCh38
rs530141793 Chr.1: 171210210-171210210 on Build GRCh38

View Full Product Details