Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 171267429-171267926 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FMO1
Entrez Gene ID
2326
Gene Name
flavin containing monooxygenase 1
Gene Aliases
-
UniGene
Hs.1424
Gene Chromosome Location
Chr.1: 171248471-171285978 on Build GRCh38
Transcripts
NM_001282692.1 NM_001282693.1 NM_001282694.1 NM_002021.2 XM_005245038.3 XM_006711241.3 XM_006711242.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4933631 c.152G>A p.R51K Chr.1: 171267562-171267562 on Build GRCh38
5745752 c.194A>G p.E65G Chr.1: 171267604-171267604 on Build GRCh38
3478274 c.160C>T p.L54F Chr.1: 171267570-171267570 on Build GRCh38
7336125 c.148G>A p.G50S Chr.1: 171267558-171267558 on Build GRCh38
5930545 c.254C>T p.S85F Chr.1: 171267664-171267664 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs769317725 Chr.1: 171267689-171267689 on Build GRCh38
rs1010838527 Chr.1: 171267455-171267455 on Build GRCh38
rs150563721 Chr.1: 171267650-171267650 on Build GRCh38
rs778711459 Chr.1: 171267629-171267629 on Build GRCh38
rs745316661 Chr.1: 171267786-171267786 on Build GRCh38
rs139810905 Chr.1: 171267566-171267566 on Build GRCh38
rs763939315 Chr.1: 171267723-171267723 on Build GRCh38
rs780220392 Chr.1: 171267537-171267537 on Build GRCh38
rs528339440 Chr.1: 171267885-171267885 on Build GRCh38
rs759054191 Chr.1: 171267578-171267578 on Build GRCh38
rs764099049 Chr.1: 171267618-171267618 on Build GRCh38
rs150820926 Chr.1: 171267768-171267768 on Build GRCh38
rs779282675 Chr.1: 171267674-171267674 on Build GRCh38
rs28360382 Chr.1: 171267788-171267788 on Build GRCh38
rs56841822 Chr.1: 171267701-171267701 on Build GRCh38
rs574601327 Chr.1: 171267637-171267637 on Build GRCh38
rs902432848 Chr.1: 171267886-171267886 on Build GRCh38
rs893670639 Chr.1: 171267803-171267803 on Build GRCh38
rs752430187 Chr.1: 171267607-171267607 on Build GRCh38
rs200210209 Chr.1: 171267550-171267550 on Build GRCh38
rs774305675 Chr.1: 171267726-171267726 on Build GRCh38
rs542596940 Chr.1: 171267456-171267456 on Build GRCh38
rs754020506 Chr.1: 171267761-171267761 on Build GRCh38
rs781079038 Chr.1: 171267638-171267638 on Build GRCh38
rs775938545 Chr.1: 171267681-171267681 on Build GRCh38
rs769051184 Chr.1: 171267666-171267666 on Build GRCh38
rs139749571 Chr.1: 171267683-171267683 on Build GRCh38
rs767561051 Chr.1: 171267737-171267737 on Build GRCh38
rs748517871 Chr.1: 171267783-171267783 on Build GRCh38
rs913247531 Chr.1: 171267814-171267814 on Build GRCh38
rs758343045 Chr.1: 171267498-171267498 on Build GRCh38
rs750459977 Chr.1: 171267739-171267739 on Build GRCh38
rs749129852 Chr.1: 171267527-171267527 on Build GRCh38
rs377033267 Chr.1: 171267587-171267587 on Build GRCh38
rs751584026 Chr.1: 171267619-171267619 on Build GRCh38
rs749632600 Chr.1: 171267656-171267656 on Build GRCh38
rs1037141792 Chr.1: 171267540-171267540 on Build GRCh38
rs375401980 Chr.1: 171267508-171267508 on Build GRCh38
rs779190772 Chr.1: 171267779-171267779 on Build GRCh38
rs567578525 Chr.1: 171267474-171267474 on Build GRCh38
rs774890789 Chr.1: 171267557-171267557 on Build GRCh38
rs142359838 Chr.1: 171267576-171267576 on Build GRCh38
rs538710309 Chr.1: 171267598-171267598 on Build GRCh38
rs988820148 Chr.1: 171267784-171267784 on Build GRCh38
rs751675766 Chr.1: 171267504-171267504 on Build GRCh38
rs780384679 Chr.1: 171267651-171267651 on Build GRCh38
rs769141001 Chr.1: 171267555-171267555 on Build GRCh38
rs202109403 Chr.1: 171267570-171267570 on Build GRCh38
rs1007893426 Chr.1: 171267523-171267523 on Build GRCh38
rs1037983826 Chr.1: 171267721-171267721 on Build GRCh38
rs757288392 Chr.1: 171267633-171267633 on Build GRCh38
rs750525854 Chr.1: 171267639-171267639 on Build GRCh38
rs1001330616 Chr.1: 171267750-171267750 on Build GRCh38
rs746033149 Chr.1: 171267521-171267521 on Build GRCh38
rs757459270 Chr.1: 171267512-171267512 on Build GRCh38
rs968766576 Chr.1: 171267824-171267824 on Build GRCh38
rs761781331 Chr.1: 171267735-171267735 on Build GRCh38
rs762897404 Chr.1: 171267611-171267611 on Build GRCh38
rs928994827 Chr.1: 171267725-171267725 on Build GRCh38
rs766482936 Chr.1: 171267755-171267755 on Build GRCh38
rs748607745 Chr.1: 171267679-171267679 on Build GRCh38
rs756269271 Chr.1: 171267751-171267751 on Build GRCh38
rs772582247 Chr.1: 171267680-171267680 on Build GRCh38
rs565799648 Chr.1: 171267709-171267709 on Build GRCh38
rs531922207 Chr.1: 171267503-171267503 on Build GRCh38

View Full Product Details