Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 171283034-171283575 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FMO1
Entrez Gene ID
2326
Gene Name
flavin containing monooxygenase 1
Gene Aliases
-
UniGene
Hs.1424
Gene Chromosome Location
Chr.1: 171248471-171285978 on Build GRCh38
Transcripts
NM_001282692.1 NM_001282693.1 NM_001282694.1 NM_002021.2 XM_005245037.3 XM_005245038.3 XM_006711241.3 XM_006711242.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2074525 c.1213A>G p.M405V Chr.1: 171283173-171283173 on Build GRCh38
3478291 c.1235G>A p.R412K Chr.1: 171283195-171283195 on Build GRCh38
1689173 c.1222G>A p.E408K Chr.1: 171283182-171283182 on Build GRCh38
4025057 c.1198C>T p.P400S Chr.1: 171283158-171283158 on Build GRCh38
6122758 c.1186G>T p.V396L Chr.1: 171283146-171283146 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs746894799 Chr.1: 171283226-171283226 on Build GRCh38
rs765337869 Chr.1: 171283266-171283282 on Build GRCh38
rs374681982 Chr.1: 171283320-171283320 on Build GRCh38
rs59755275 Chr.1: 171283295-171283297 on Build GRCh38
rs779739003 Chr.1: 171283099-171283099 on Build GRCh38
rs1001159467 Chr.1: 171283489-171283489 on Build GRCh38
rs775423112 Chr.1: 171283241-171283241 on Build GRCh38
rs745840342 Chr.1: 171283236-171283237 on Build GRCh38
rs764503595 Chr.1: 171283257-171283257 on Build GRCh38
rs11589290 Chr.1: 171283430-171283430 on Build GRCh38
rs754487395 Chr.1: 171283267-171283267 on Build GRCh38
rs562976131 Chr.1: 171283420-171283420 on Build GRCh38
rs970402170 Chr.1: 171283298-171283298 on Build GRCh38
rs577487000 Chr.1: 171283209-171283209 on Build GRCh38
rs116209269 Chr.1: 171283452-171283452 on Build GRCh38
rs770514618 Chr.1: 171283231-171283232 on Build GRCh38
rs772028445 Chr.1: 171283133-171283133 on Build GRCh38
rs756768001 Chr.1: 171283492-171283492 on Build GRCh38
rs796848636 Chr.1: 171283290-171283292 on Build GRCh38
rs761910511 Chr.1: 171283266-171283278 on Build GRCh38
rs11589287 Chr.1: 171283378-171283378 on Build GRCh38
rs1049525892 Chr.1: 171283415-171283415 on Build GRCh38
rs925228971 Chr.1: 171283375-171283375 on Build GRCh38
rs564041104 Chr.1: 171283494-171283494 on Build GRCh38
rs763266614 Chr.1: 171283255-171283255 on Build GRCh38
rs867249493 Chr.1: 171283284-171283284 on Build GRCh38
rs371464520 Chr.1: 171283344-171283344 on Build GRCh38
rs28360426 Chr.1: 171283310-171283311 on Build GRCh38
rs750476074 Chr.1: 171283266-171283277 on Build GRCh38
rs34983496 Chr.1: 171283073-171283074 on Build GRCh38
rs768198578 Chr.1: 171283264-171283264 on Build GRCh38
rs373075012 Chr.1: 171283282-171283283 on Build GRCh38
rs774460814 Chr.1: 171283235-171283236 on Build GRCh38
rs753467526 Chr.1: 171283200-171283200 on Build GRCh38
rs774867753 Chr.1: 171283259-171283259 on Build GRCh38
rs866287781 Chr.1: 171283295-171283295 on Build GRCh38
rs773174188 Chr.1: 171283137-171283137 on Build GRCh38
rs1036281424 Chr.1: 171283430-171283431 on Build GRCh38
rs775609862 Chr.1: 171283248-171283248 on Build GRCh38
rs12092985 Chr.1: 171283398-171283398 on Build GRCh38
rs978180313 Chr.1: 171283299-171283299 on Build GRCh38
rs778479559 Chr.1: 171283204-171283204 on Build GRCh38
rs369795138 Chr.1: 171283231-171283231 on Build GRCh38
rs374900858 Chr.1: 171283246-171283246 on Build GRCh38
rs371841951 Chr.1: 171283214-171283214 on Build GRCh38
rs375889373 Chr.1: 171283298-171283304 on Build GRCh38
rs906137947 Chr.1: 171283535-171283535 on Build GRCh38
rs979317927 Chr.1: 171283366-171283366 on Build GRCh38
rs762341607 Chr.1: 171283263-171283263 on Build GRCh38
rs764630922 Chr.1: 171283178-171283178 on Build GRCh38
rs754647737 Chr.1: 171283201-171283201 on Build GRCh38
rs7522793 Chr.1: 171283303-171283303 on Build GRCh38
rs370891332 Chr.1: 171283310-171283310 on Build GRCh38
rs369656596 Chr.1: 171283303-171283304 on Build GRCh38
rs768656093 Chr.1: 171283469-171283469 on Build GRCh38
rs369826926 Chr.1: 171283096-171283096 on Build GRCh38
rs559359446 Chr.1: 171283317-171283317 on Build GRCh38
rs58911039 Chr.1: 171283297-171283297 on Build GRCh38
rs936581973 Chr.1: 171283304-171283304 on Build GRCh38
rs376198073 Chr.1: 171283092-171283092 on Build GRCh38
rs576788028 Chr.1: 171283104-171283104 on Build GRCh38
rs752072022 Chr.1: 171283181-171283181 on Build GRCh38
rs748819235 Chr.1: 171283230-171283242 on Build GRCh38
rs753088959 Chr.1: 171283162-171283162 on Build GRCh38
rs764391746 Chr.1: 171283266-171283280 on Build GRCh38
rs149541428 Chr.1: 171283485-171283485 on Build GRCh38
rs574489946 Chr.1: 171283386-171283386 on Build GRCh38
rs571381164 Chr.1: 171283113-171283113 on Build GRCh38
rs869047317 Chr.1: 171283320-171283321 on Build GRCh38
rs397844228 Chr.1: 171283319-171283320 on Build GRCh38
rs376888994 Chr.1: 171283283-171283284 on Build GRCh38
rs59966758 Chr.1: 171283294-171283297 on Build GRCh38
rs376008142 Chr.1: 171283298-171283299 on Build GRCh38
rs765441758 Chr.1: 171283161-171283161 on Build GRCh38
rs559308229 Chr.1: 171283166-171283166 on Build GRCh38
rs3765336 Chr.1: 171283313-171283313 on Build GRCh38
rs765927391 Chr.1: 171283192-171283192 on Build GRCh38
rs1126692 Chr.1: 171283148-171283148 on Build GRCh38
rs931268154 Chr.1: 171283358-171283358 on Build GRCh38
rs747954466 Chr.1: 171283127-171283127 on Build GRCh38
rs368453140 Chr.1: 171283298-171283310 on Build GRCh38
rs35331306 Chr.1: 171283266-171283266 on Build GRCh38
rs1029552581 Chr.1: 171283067-171283067 on Build GRCh38
rs776707882 Chr.1: 171283158-171283158 on Build GRCh38
rs768203959 Chr.1: 171283107-171283107 on Build GRCh38
rs760641362 Chr.1: 171283145-171283145 on Build GRCh38
rs760321978 Chr.1: 171283266-171283281 on Build GRCh38
rs944101359 Chr.1: 171283466-171283466 on Build GRCh38
rs771890990 Chr.1: 171283240-171283241 on Build GRCh38
rs866025094 Chr.1: 171283180-171283180 on Build GRCh38
rs12057955 Chr.1: 171283548-171283548 on Build GRCh38
rs867465772 Chr.1: 171283285-171283285 on Build GRCh38
rs540085380 Chr.1: 171283285-171283297 on Build GRCh38
rs770031590 Chr.1: 171283239-171283239 on Build GRCh38
rs150824879 Chr.1: 171283085-171283085 on Build GRCh38
rs796130161 Chr.1: 171283293-171283304 on Build GRCh38
rs796481227 Chr.1: 171283299-171283310 on Build GRCh38
rs770862345 Chr.1: 171283227-171283227 on Build GRCh38
rs757860489 Chr.1: 171283191-171283191 on Build GRCh38
rs777570410 Chr.1: 171283216-171283216 on Build GRCh38
rs185338607 Chr.1: 171283534-171283534 on Build GRCh38
rs753377595 Chr.1: 171283266-171283266 on Build GRCh38
rs755721540 Chr.1: 171283093-171283093 on Build GRCh38
rs772906428 Chr.1: 171283550-171283550 on Build GRCh38
rs945040897 Chr.1: 171283345-171283345 on Build GRCh38
rs868057676 Chr.1: 171283175-171283175 on Build GRCh38
rs980641436 Chr.1: 171283327-171283328 on Build GRCh38
rs1050646435 Chr.1: 171283483-171283483 on Build GRCh38
rs776787526 Chr.1: 171283266-171283279 on Build GRCh38
rs759685080 Chr.1: 171283160-171283160 on Build GRCh38
rs56208728 Chr.1: 171283291-171283297 on Build GRCh38

View Full Product Details