Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 93166653-93167157 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C8orf87
Entrez Gene ID
389676
Gene Name
chromosome 8 open reading frame 87
Gene Aliases
-
UniGene
Hs.386711
Gene Chromosome Location
Chr.8: 93134095-93166850 on Build GRCh38
Transcripts
NM_001242668.1

SNP Detail

SNP ID Chromosome Location
rs918255934 Chr.8: 93166751-93166751 on Build GRCh38
rs72662238 Chr.8: 93166991-93166991 on Build GRCh38
rs764050001 Chr.8: 93166918-93166918 on Build GRCh38
rs902956531 Chr.8: 93167112-93167112 on Build GRCh38
rs368794531 Chr.8: 93166722-93166722 on Build GRCh38
rs554638798 Chr.8: 93166801-93166801 on Build GRCh38
rs965163506 Chr.8: 93166965-93166965 on Build GRCh38
rs759081146 Chr.8: 93166879-93166879 on Build GRCh38
rs193058916 Chr.8: 93167114-93167114 on Build GRCh38
rs986007833 Chr.8: 93166823-93166823 on Build GRCh38
rs148248007 Chr.8: 93166953-93166953 on Build GRCh38
rs986273995 Chr.8: 93166707-93166707 on Build GRCh38
rs376314616 Chr.8: 93166684-93166684 on Build GRCh38
rs974139336 Chr.8: 93166866-93166866 on Build GRCh38
rs147159696 Chr.8: 93166912-93166912 on Build GRCh38
rs775022519 Chr.8: 93166788-93166788 on Build GRCh38
rs558270412 Chr.8: 93166687-93166687 on Build GRCh38
rs760364167 Chr.8: 93166820-93166820 on Build GRCh38
rs753693572 Chr.8: 93166923-93166923 on Build GRCh38
rs980609816 Chr.8: 93167129-93167129 on Build GRCh38
rs571294985 Chr.8: 93166735-93166735 on Build GRCh38
rs1011218596 Chr.8: 93167021-93167021 on Build GRCh38
rs184756640 Chr.8: 93166872-93166872 on Build GRCh38
rs553698692 Chr.8: 93166791-93166791 on Build GRCh38
rs186779927 Chr.8: 93166720-93166720 on Build GRCh38
rs939434805 Chr.8: 93166818-93166818 on Build GRCh38
rs965091285 Chr.8: 93166724-93166724 on Build GRCh38
rs577644972 Chr.8: 93167053-93167053 on Build GRCh38
rs771548195 Chr.8: 93166764-93166764 on Build GRCh38
rs374946912 Chr.8: 93166908-93166909 on Build GRCh38
rs370096199 Chr.8: 93167097-93167097 on Build GRCh38
rs188790160 Chr.8: 93167005-93167005 on Build GRCh38
rs778899172 Chr.8: 93166756-93166756 on Build GRCh38
rs749053565 Chr.8: 93166713-93166713 on Build GRCh38
rs911224129 Chr.8: 93166862-93166862 on Build GRCh38
rs191591431 Chr.8: 93166763-93166763 on Build GRCh38
rs762758078 Chr.8: 93166697-93166697 on Build GRCh38
rs976859286 Chr.8: 93166730-93166730 on Build GRCh38
rs1052885120 Chr.8: 93166837-93166837 on Build GRCh38
rs768388615 Chr.8: 93166802-93166802 on Build GRCh38
rs984170854 Chr.8: 93167018-93167018 on Build GRCh38
rs951094659 Chr.8: 93167009-93167009 on Build GRCh38
rs1020050740 Chr.8: 93167039-93167039 on Build GRCh38
rs751139846 Chr.8: 93166863-93166863 on Build GRCh38
rs7817523 Chr.8: 93167008-93167008 on Build GRCh38
rs923112265 Chr.8: 93166766-93166766 on Build GRCh38
rs947909662 Chr.8: 93166787-93166787 on Build GRCh38
rs937618523 Chr.8: 93166969-93166969 on Build GRCh38

View Full Product Details