Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 103042188-103042600 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATP6V1C1
Entrez Gene ID
528
Gene Name
ATPase H+ transporting V1 subunit C1
Gene Aliases
ATP6C, ATP6D, VATC, Vma5
UniGene
Hs.86905
Gene Chromosome Location
Chr.8: 103021020-103073057 on Build GRCh38
Transcripts
NM_001695.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1094450 c.140C>G p.T47R Chr.8: 103042347-103042347 on Build GRCh38
421804 c.175G>C p.A59P Chr.8: 103042382-103042382 on Build GRCh38
1094451 c.200+2T>C p.? Chr.8: 103042409-103042409 on Build GRCh38
5885781 c.144_145delGG p.D49fs*8 Chr.8: 103042351-103042352 on Build GRCh38
5501102 c.189A>G p.A63A Chr.8: 103042396-103042396 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs189143176 Chr.8: 103042538-103042538 on Build GRCh38
rs28566904 Chr.8: 103042297-103042297 on Build GRCh38
rs984253338 Chr.8: 103042486-103042486 on Build GRCh38
rs974618218 Chr.8: 103042533-103042533 on Build GRCh38
rs943270325 Chr.8: 103042284-103042284 on Build GRCh38
rs767884054 Chr.8: 103042303-103042303 on Build GRCh38
rs745694688 Chr.8: 103042333-103042333 on Build GRCh38
rs1012126886 Chr.8: 103042396-103042396 on Build GRCh38
rs371738834 Chr.8: 103042424-103042424 on Build GRCh38
rs752902037 Chr.8: 103042306-103042306 on Build GRCh38
rs771811020 Chr.8: 103042336-103042336 on Build GRCh38
rs909035172 Chr.8: 103042511-103042511 on Build GRCh38
rs1042067154 Chr.8: 103042571-103042571 on Build GRCh38
rs757188353 Chr.8: 103042320-103042320 on Build GRCh38
rs774683419 Chr.8: 103042412-103042412 on Build GRCh38
rs760073408 Chr.8: 103042455-103042457 on Build GRCh38
rs1017269750 Chr.8: 103042402-103042402 on Build GRCh38
rs555394441 Chr.8: 103042233-103042233 on Build GRCh38
rs763423415 Chr.8: 103042382-103042382 on Build GRCh38
rs1038965897 Chr.8: 103042285-103042285 on Build GRCh38
rs767051501 Chr.8: 103042389-103042389 on Build GRCh38
rs1026382914 Chr.8: 103042418-103042418 on Build GRCh38
rs755250667 Chr.8: 103042299-103042299 on Build GRCh38
rs753931988 Chr.8: 103042431-103042431 on Build GRCh38
rs766697928 Chr.8: 103042296-103042296 on Build GRCh38
rs750124163 Chr.8: 103042378-103042378 on Build GRCh38
rs750259406 Chr.8: 103042434-103042434 on Build GRCh38
rs567012208 Chr.8: 103042309-103042309 on Build GRCh38
rs901496768 Chr.8: 103042285-103042285 on Build GRCh38
rs577318073 Chr.8: 103042312-103042312 on Build GRCh38
rs771495111 Chr.8: 103042401-103042401 on Build GRCh38
rs371500951 Chr.8: 103042285-103042286 on Build GRCh38
rs746705001 Chr.8: 103042450-103042450 on Build GRCh38
rs113025588 Chr.8: 103042465-103042465 on Build GRCh38
rs764305377 Chr.8: 103042429-103042429 on Build GRCh38
rs371680106 Chr.8: 103042345-103042345 on Build GRCh38
rs567405795 Chr.8: 103042281-103042281 on Build GRCh38
rs974993668 Chr.8: 103042523-103042523 on Build GRCh38
rs543279211 Chr.8: 103042539-103042539 on Build GRCh38
rs767932946 Chr.8: 103042420-103042420 on Build GRCh38
rs761042446 Chr.8: 103042297-103042298 on Build GRCh38
rs3779908 Chr.8: 103042298-103042298 on Build GRCh38
rs746653614 Chr.8: 103042351-103042351 on Build GRCh38
rs202036945 Chr.8: 103042437-103042437 on Build GRCh38
rs796444277 Chr.8: 103042297-103042298 on Build GRCh38
rs963342444 Chr.8: 103042411-103042411 on Build GRCh38
rs946398382 Chr.8: 103042261-103042261 on Build GRCh38
rs7009365 Chr.8: 103042375-103042375 on Build GRCh38
rs994856084 Chr.8: 103042415-103042415 on Build GRCh38
rs369016872 Chr.8: 103042421-103042421 on Build GRCh38
rs1039698212 Chr.8: 103042218-103042218 on Build GRCh38
rs952912461 Chr.8: 103042476-103042476 on Build GRCh38
rs368123012 Chr.8: 103042325-103042325 on Build GRCh38
rs1042098071 Chr.8: 103042286-103042286 on Build GRCh38
rs142055270 Chr.8: 103042298-103042298 on Build GRCh38
rs879096617 Chr.8: 103042286-103042286 on Build GRCh38
rs373645213 Chr.8: 103042433-103042433 on Build GRCh38
rs375980479 Chr.8: 103042357-103042357 on Build GRCh38
rs779822181 Chr.8: 103042448-103042448 on Build GRCh38

View Full Product Details