Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 103298894-103299392 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FZD6
Entrez Gene ID
8323
Gene Name
frizzled class receptor 6
Gene Aliases
FZ-6, FZ6, HFZ6, NDNC10
UniGene
Hs.591863
Gene Chromosome Location
Chr.8: 103298433-103332866 on Build GRCh38
Transcripts
NM_001164615.1 NM_001164616.1 NM_001317796.1 NM_003506.3 NR_133921.1

SNP Detail

SNP ID Chromosome Location
rs1030917138 Chr.8: 103299242-103299242 on Build GRCh38
rs972232778 Chr.8: 103299006-103299006 on Build GRCh38
rs112989704 Chr.8: 103299128-103299128 on Build GRCh38
rs896613669 Chr.8: 103299121-103299121 on Build GRCh38
rs1053686715 Chr.8: 103299083-103299083 on Build GRCh38
rs564354572 Chr.8: 103299124-103299124 on Build GRCh38
rs1055041627 Chr.8: 103299091-103299091 on Build GRCh38
rs749858790 Chr.8: 103299308-103299308 on Build GRCh38
rs1016457798 Chr.8: 103299211-103299211 on Build GRCh38
rs2272097 Chr.8: 103299036-103299036 on Build GRCh38
rs1006421719 Chr.8: 103298987-103298987 on Build GRCh38
rs372052056 Chr.8: 103299146-103299146 on Build GRCh38
rs779762210 Chr.8: 103299241-103299241 on Build GRCh38
rs769301425 Chr.8: 103299366-103299366 on Build GRCh38
rs985772036 Chr.8: 103299056-103299056 on Build GRCh38
rs945657329 Chr.8: 103299012-103299012 on Build GRCh38
rs1002853331 Chr.8: 103298944-103298944 on Build GRCh38
rs979551562 Chr.8: 103299354-103299354 on Build GRCh38
rs962166833 Chr.8: 103298996-103298996 on Build GRCh38
rs827526 Chr.8: 103299295-103299295 on Build GRCh38
rs907246206 Chr.8: 103299131-103299131 on Build GRCh38
rs937898855 Chr.8: 103299365-103299365 on Build GRCh38
rs1016510157 Chr.8: 103298988-103298988 on Build GRCh38
rs74949239 Chr.8: 103299294-103299294 on Build GRCh38
rs1034768240 Chr.8: 103298954-103298954 on Build GRCh38
rs771410381 Chr.8: 103299316-103299316 on Build GRCh38
rs989161354 Chr.8: 103299311-103299311 on Build GRCh38
rs927780525 Chr.8: 103299348-103299348 on Build GRCh38
rs936391309 Chr.8: 103299080-103299080 on Build GRCh38
rs527982295 Chr.8: 103298940-103298940 on Build GRCh38
rs780871977 Chr.8: 103299252-103299252 on Build GRCh38
rs1020368908 Chr.8: 103298920-103298920 on Build GRCh38
rs779329870 Chr.8: 103299359-103299359 on Build GRCh38
rs827525 Chr.8: 103299248-103299248 on Build GRCh38
rs938037170 Chr.8: 103299086-103299086 on Build GRCh38
rs922994216 Chr.8: 103299033-103299033 on Build GRCh38
rs945279082 Chr.8: 103299321-103299321 on Build GRCh38
rs910215779 Chr.8: 103299061-103299061 on Build GRCh38
rs560063836 Chr.8: 103299200-103299200 on Build GRCh38
rs866148259 Chr.8: 103298967-103298967 on Build GRCh38
rs182414246 Chr.8: 103299187-103299187 on Build GRCh38
rs1014047616 Chr.8: 103299123-103299123 on Build GRCh38
rs550032401 Chr.8: 103299204-103299204 on Build GRCh38
rs1003378552 Chr.8: 103299137-103299137 on Build GRCh38
rs962283231 Chr.8: 103299215-103299215 on Build GRCh38
rs560662665 Chr.8: 103298912-103298912 on Build GRCh38
rs1055918698 Chr.8: 103299161-103299161 on Build GRCh38
rs913851966 Chr.8: 103299110-103299110 on Build GRCh38
rs955155638 Chr.8: 103299249-103299249 on Build GRCh38
rs1046229472 Chr.8: 103299322-103299322 on Build GRCh38
rs894463531 Chr.8: 103299178-103299178 on Build GRCh38
rs566728282 Chr.8: 103299343-103299343 on Build GRCh38

View Full Product Details