Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 103299872-103300387 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FZD6
Entrez Gene ID
8323
Gene Name
frizzled class receptor 6
Gene Aliases
FZ-6, FZ6, HFZ6, NDNC10
UniGene
Hs.591863
Gene Chromosome Location
Chr.8: 103298433-103332866 on Build GRCh38
Transcripts
NM_001164615.1 NM_001164616.1 NM_001317796.1 NM_003506.3 NR_133921.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6549582 c.52G>T p.G18W Chr.8: 103300159-103300159 on Build GRCh38
1623446 c.39A>G p.L13L Chr.8: 103300146-103300146 on Build GRCh38
6013234 c.68C>T p.T23I Chr.8: 103300175-103300175 on Build GRCh38
6668172 c.159T>C p.I53I Chr.8: 103300266-103300266 on Build GRCh38
6668177 c.19T>G p.L7V Chr.8: 103300126-103300126 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs111920622 Chr.8: 103300208-103300208 on Build GRCh38
rs752048729 Chr.8: 103300202-103300202 on Build GRCh38
rs973457330 Chr.8: 103300356-103300357 on Build GRCh38
rs3736046 Chr.8: 103300303-103300303 on Build GRCh38
rs370879185 Chr.8: 103300327-103300327 on Build GRCh38
rs755276890 Chr.8: 103300106-103300106 on Build GRCh38
rs144185033 Chr.8: 103300272-103300272 on Build GRCh38
rs773146626 Chr.8: 103300236-103300236 on Build GRCh38
rs1032484941 Chr.8: 103300122-103300122 on Build GRCh38
rs552528993 Chr.8: 103300045-103300045 on Build GRCh38
rs553335677 Chr.8: 103300238-103300238 on Build GRCh38
rs748362787 Chr.8: 103300114-103300114 on Build GRCh38
rs374556380 Chr.8: 103300206-103300206 on Build GRCh38
rs919364870 Chr.8: 103299916-103299916 on Build GRCh38
rs372702981 Chr.8: 103300165-103300165 on Build GRCh38
rs827528 Chr.8: 103300204-103300204 on Build GRCh38
rs770928662 Chr.8: 103300138-103300138 on Build GRCh38
rs376581731 Chr.8: 103300082-103300082 on Build GRCh38
rs752212598 Chr.8: 103300164-103300164 on Build GRCh38
rs527570952 Chr.8: 103300005-103300005 on Build GRCh38
rs1020611585 Chr.8: 103300047-103300047 on Build GRCh38
rs754581593 Chr.8: 103300133-103300133 on Build GRCh38
rs773784932 Chr.8: 103300319-103300319 on Build GRCh38
rs768184791 Chr.8: 103300242-103300242 on Build GRCh38
rs201580513 Chr.8: 103300191-103300191 on Build GRCh38
rs766063780 Chr.8: 103299941-103299941 on Build GRCh38
rs757230055 Chr.8: 103300262-103300262 on Build GRCh38
rs763564162 Chr.8: 103300176-103300176 on Build GRCh38
rs150839394 Chr.8: 103300192-103300192 on Build GRCh38
rs751952479 Chr.8: 103300087-103300087 on Build GRCh38
rs1039727243 Chr.8: 103300011-103300011 on Build GRCh38
rs777884052 Chr.8: 103300131-103300131 on Build GRCh38
rs771982148 Chr.8: 103300161-103300161 on Build GRCh38
rs753953103 Chr.8: 103300261-103300261 on Build GRCh38
rs932570091 Chr.8: 103299910-103299910 on Build GRCh38
rs779974159 Chr.8: 103300307-103300307 on Build GRCh38
rs779056600 Chr.8: 103300265-103300265 on Build GRCh38
rs568034251 Chr.8: 103300200-103300200 on Build GRCh38
rs943642777 Chr.8: 103300007-103300007 on Build GRCh38
rs963465921 Chr.8: 103300289-103300289 on Build GRCh38
rs1051793032 Chr.8: 103299989-103299989 on Build GRCh38
rs375688505 Chr.8: 103300166-103300166 on Build GRCh38
rs539738840 Chr.8: 103300332-103300332 on Build GRCh38
rs3736047 Chr.8: 103300113-103300113 on Build GRCh38
rs117477069 Chr.8: 103300153-103300153 on Build GRCh38
rs760374585 Chr.8: 103300175-103300175 on Build GRCh38
rs761443232 Chr.8: 103300333-103300333 on Build GRCh38
rs559803467 Chr.8: 103299915-103299915 on Build GRCh38
rs571735437 Chr.8: 103300271-103300271 on Build GRCh38
rs756185937 Chr.8: 103300119-103300119 on Build GRCh38
rs746328732 Chr.8: 103300218-103300218 on Build GRCh38
rs893976907 Chr.8: 103300019-103300019 on Build GRCh38
rs768386963 Chr.8: 103300324-103300324 on Build GRCh38
rs755552434 Chr.8: 103300170-103300170 on Build GRCh38
rs970993467 Chr.8: 103300207-103300207 on Build GRCh38
rs777814122 Chr.8: 103300250-103300250 on Build GRCh38
rs746734173 Chr.8: 103300313-103300313 on Build GRCh38
rs774093482 Chr.8: 103300150-103300150 on Build GRCh38
rs752962628 Chr.8: 103300220-103300220 on Build GRCh38

View Full Product Details